Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33666394-33666892 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTL10
Entrez Gene ID
170487
Gene Name
actin like 10
Gene Aliases
C20orf134, dJ63M2.2
UniGene
Hs.592151
Gene Chromosome Location
Chr.20: 33666498-33668525 on Build GRCh38
Transcripts
NM_001024675.1 NM_031231.3 NM_031232.3 XM_005260510.1 XM_011528992.2 XM_017028015.1

Gene Symbol
NECAB3
Entrez Gene ID
63941
Gene Name
N-terminal EF-hand calcium binding protein 3
Gene Aliases
APBA2BP, EFCBP3, NIP1, STIP3, SYTIP2, XB51, dJ63M2.4, dJ63M2.5
UniGene
Hs.516986
Gene Chromosome Location
Chr.20: 33657087-33674458 on Build GRCh38
Transcripts
NM_001024675.1 NM_031231.3 NM_031232.3 XM_005260510.1 XM_011528992.2 XM_017028015.1

SNP Detail

SNP ID Chromosome Location
rs550250630 Chr.20: 33666765-33666765 on Build GRCh38
rs926010007 Chr.20: 33666690-33666690 on Build GRCh38
rs927374268 Chr.20: 33666715-33666715 on Build GRCh38
rs995876575 Chr.20: 33666769-33666769 on Build GRCh38
rs764214533 Chr.20: 33666537-33666537 on Build GRCh38
rs372869529 Chr.20: 33666510-33666510 on Build GRCh38
rs1010813615 Chr.20: 33666759-33666759 on Build GRCh38
rs755918148 Chr.20: 33666632-33666632 on Build GRCh38
rs563650218 Chr.20: 33666515-33666515 on Build GRCh38
rs980106129 Chr.20: 33666863-33666863 on Build GRCh38
rs78347838 Chr.20: 33666771-33666771 on Build GRCh38
rs1024909524 Chr.20: 33666855-33666855 on Build GRCh38
rs754844980 Chr.20: 33666835-33666835 on Build GRCh38
rs1005963628 Chr.20: 33666816-33666816 on Build GRCh38
rs916134362 Chr.20: 33666640-33666640 on Build GRCh38
rs1014817587 Chr.20: 33666841-33666841 on Build GRCh38
rs907680838 Chr.20: 33666538-33666538 on Build GRCh38
rs763290850 Chr.20: 33666461-33666461 on Build GRCh38
rs370149268 Chr.20: 33666768-33666768 on Build GRCh38
rs191559365 Chr.20: 33666621-33666621 on Build GRCh38
rs984604046 Chr.20: 33666508-33666508 on Build GRCh38
rs971823917 Chr.20: 33666545-33666545 on Build GRCh38
rs892282051 Chr.20: 33666760-33666760 on Build GRCh38
rs1052259663 Chr.20: 33666427-33666427 on Build GRCh38
rs778535064 Chr.20: 33666847-33666847 on Build GRCh38
rs531681422 Chr.20: 33666740-33666740 on Build GRCh38
rs376991547 Chr.20: 33666613-33666613 on Build GRCh38
rs936036306 Chr.20: 33666720-33666720 on Build GRCh38
rs979168064 Chr.20: 33666675-33666675 on Build GRCh38
rs1019612688 Chr.20: 33666430-33666430 on Build GRCh38
rs1026031085 Chr.20: 33666824-33666824 on Build GRCh38
rs961931980 Chr.20: 33666840-33666840 on Build GRCh38
rs79959038 Chr.20: 33666636-33666636 on Build GRCh38
rs996232403 Chr.20: 33666459-33666459 on Build GRCh38
rs933154559 Chr.20: 33666761-33666761 on Build GRCh38
rs181841703 Chr.20: 33666468-33666468 on Build GRCh38
rs1044394439 Chr.20: 33666701-33666701 on Build GRCh38
rs1014617056 Chr.20: 33666818-33666818 on Build GRCh38
rs1026400168 Chr.20: 33666605-33666605 on Build GRCh38
rs950101422 Chr.20: 33666660-33666660 on Build GRCh38
rs969680342 Chr.20: 33666859-33666859 on Build GRCh38
rs186701833 Chr.20: 33666455-33666455 on Build GRCh38
rs1016338726 Chr.20: 33666790-33666790 on Build GRCh38
rs1028590005 Chr.20: 33666770-33666770 on Build GRCh38
rs1015957097 Chr.20: 33666517-33666517 on Build GRCh38
rs963097717 Chr.20: 33666527-33666527 on Build GRCh38
rs949021615 Chr.20: 33666641-33666641 on Build GRCh38
rs959809077 Chr.20: 33666831-33666831 on Build GRCh38
rs780166568 Chr.20: 33666689-33666689 on Build GRCh38
rs898835998 Chr.20: 33666806-33666806 on Build GRCh38
rs927028861 Chr.20: 33666712-33666712 on Build GRCh38
rs951777160 Chr.20: 33666486-33666486 on Build GRCh38
rs888760261 Chr.20: 33666473-33666473 on Build GRCh38
rs1040658908 Chr.20: 33666764-33666764 on Build GRCh38
rs892311841 Chr.20: 33666439-33666439 on Build GRCh38
rs1054630166 Chr.20: 33666738-33666738 on Build GRCh38

View Full Product Details