Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44160072-44160571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JPH2
Entrez Gene ID
57158
Gene Name
junctophilin 2
Gene Aliases
CMH17, JP-2, JP2
UniGene
Hs.441737
Gene Chromosome Location
Chr.20: 44111697-44187578 on Build GRCh38
Transcripts
NM_020433.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7265019 c.413G>A p.R138H Chr.20: 44160374-44160374 on Build GRCh38
6689704 c.385T>C p.Y129H Chr.20: 44160402-44160402 on Build GRCh38
7248223 c.500G>A p.R167H Chr.20: 44160287-44160287 on Build GRCh38
4098673 c.415C>A p.H139N Chr.20: 44160372-44160372 on Build GRCh38
5429599 c.546G>A p.S182S Chr.20: 44160241-44160241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038484873 Chr.20: 44160537-44160537 on Build GRCh38
rs977513162 Chr.20: 44160539-44160539 on Build GRCh38
rs772816984 Chr.20: 44160116-44160116 on Build GRCh38
rs730880254 Chr.20: 44160222-44160222 on Build GRCh38
rs767328866 Chr.20: 44160150-44160150 on Build GRCh38
rs914203878 Chr.20: 44160207-44160207 on Build GRCh38
rs952147058 Chr.20: 44160146-44160146 on Build GRCh38
rs762464432 Chr.20: 44160349-44160349 on Build GRCh38
rs953581894 Chr.20: 44160186-44160186 on Build GRCh38
rs779167207 Chr.20: 44160427-44160427 on Build GRCh38
rs561135147 Chr.20: 44160491-44160491 on Build GRCh38
rs387906898 Chr.20: 44160293-44160293 on Build GRCh38
rs587782951 Chr.20: 44160305-44160305 on Build GRCh38
rs894217949 Chr.20: 44160309-44160309 on Build GRCh38
rs864309612 Chr.20: 44160270-44160271 on Build GRCh38
rs922166422 Chr.20: 44160175-44160175 on Build GRCh38
rs559155501 Chr.20: 44160425-44160425 on Build GRCh38
rs768914995 Chr.20: 44160448-44160448 on Build GRCh38
rs557042688 Chr.20: 44160331-44160331 on Build GRCh38
rs777856415 Chr.20: 44160402-44160402 on Build GRCh38
rs938498462 Chr.20: 44160240-44160240 on Build GRCh38
rs772526418 Chr.20: 44160317-44160317 on Build GRCh38
rs1029141147 Chr.20: 44160483-44160483 on Build GRCh38
rs772582964 Chr.20: 44160344-44160344 on Build GRCh38
rs772851568 Chr.20: 44160457-44160457 on Build GRCh38
rs140538587 Chr.20: 44160382-44160382 on Build GRCh38
rs776045429 Chr.20: 44160329-44160329 on Build GRCh38
rs587780956 Chr.20: 44160145-44160145 on Build GRCh38
rs759488753 Chr.20: 44160373-44160373 on Build GRCh38
rs764629322 Chr.20: 44160112-44160112 on Build GRCh38
rs945931469 Chr.20: 44160443-44160443 on Build GRCh38
rs886038834 Chr.20: 44160311-44160311 on Build GRCh38
rs373715686 Chr.20: 44160439-44160439 on Build GRCh38
rs543732492 Chr.20: 44160321-44160321 on Build GRCh38
rs779505448 Chr.20: 44160287-44160287 on Build GRCh38
rs747722861 Chr.20: 44160330-44160330 on Build GRCh38
rs767281714 Chr.20: 44160381-44160381 on Build GRCh38
rs572602608 Chr.20: 44160456-44160456 on Build GRCh38
rs761591158 Chr.20: 44160095-44160095 on Build GRCh38
rs976272461 Chr.20: 44160167-44160167 on Build GRCh38
rs565885872 Chr.20: 44160205-44160205 on Build GRCh38
rs894196248 Chr.20: 44160527-44160527 on Build GRCh38
rs972983859 Chr.20: 44160403-44160403 on Build GRCh38
rs757693348 Chr.20: 44160270-44160270 on Build GRCh38
rs759012012 Chr.20: 44160444-44160444 on Build GRCh38
rs746384802 Chr.20: 44160304-44160304 on Build GRCh38
rs1055875145 Chr.20: 44160520-44160520 on Build GRCh38
rs534491843 Chr.20: 44160208-44160208 on Build GRCh38
rs554853074 Chr.20: 44160215-44160215 on Build GRCh38
rs1012335909 Chr.20: 44160348-44160348 on Build GRCh38
rs766395991 Chr.20: 44160122-44160122 on Build GRCh38
rs1002693181 Chr.20: 44160093-44160093 on Build GRCh38
rs201197277 Chr.20: 44160413-44160413 on Build GRCh38
rs987288946 Chr.20: 44160419-44160419 on Build GRCh38
rs553395930 Chr.20: 44160319-44160319 on Build GRCh38
rs869025440 Chr.20: 44160228-44160228 on Build GRCh38
rs1055580234 Chr.20: 44160247-44160247 on Build GRCh38
rs574746149 Chr.20: 44160225-44160225 on Build GRCh38
rs762794427 Chr.20: 44160119-44160119 on Build GRCh38
rs765874503 Chr.20: 44160363-44160363 on Build GRCh38
rs387906897 Chr.20: 44160366-44160366 on Build GRCh38
rs767560074 Chr.20: 44160462-44160462 on Build GRCh38
rs747464638 Chr.20: 44160446-44160446 on Build GRCh38
rs886038779 Chr.20: 44160250-44160250 on Build GRCh38
rs762346600 Chr.20: 44160428-44160428 on Build GRCh38
rs398124358 Chr.20: 44160163-44160163 on Build GRCh38
rs753525812 Chr.20: 44160329-44160330 on Build GRCh38
rs759548679 Chr.20: 44160432-44160434 on Build GRCh38
rs111987307 Chr.20: 44160416-44160416 on Build GRCh38
rs989955838 Chr.20: 44160437-44160437 on Build GRCh38
rs869025439 Chr.20: 44160285-44160285 on Build GRCh38
rs1005944157 Chr.20: 44160528-44160528 on Build GRCh38
rs868619268 Chr.20: 44160332-44160332 on Build GRCh38
rs796219248 Chr.20: 44160139-44160139 on Build GRCh38
rs942789550 Chr.20: 44160431-44160431 on Build GRCh38
rs12480648 Chr.20: 44160512-44160512 on Build GRCh38
rs914841890 Chr.20: 44160524-44160524 on Build GRCh38
rs876657835 Chr.20: 44160290-44160290 on Build GRCh38
rs746329738 Chr.20: 44160441-44160441 on Build GRCh38
rs541224104 Chr.20: 44160474-44160474 on Build GRCh38
rs953527871 Chr.20: 44160505-44160505 on Build GRCh38
rs558770240 Chr.20: 44160126-44160126 on Build GRCh38

View Full Product Details