Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 59006220-59006711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSZ
Entrez Gene ID
1522
Gene Name
cathepsin Z
Gene Aliases
CTSX
UniGene
Hs.252549
Gene Chromosome Location
Chr.20: 58995187-59007254 on Build GRCh38
Transcripts
NM_001336.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5618904 c.192G>A p.K64K Chr.20: 59006437-59006437 on Build GRCh38
5881224 c.274T>C p.C92R Chr.20: 59006355-59006355 on Build GRCh38
2765000 c.214G>C p.D72H Chr.20: 59006415-59006415 on Build GRCh38
1028538 c.202T>A p.W68R Chr.20: 59006427-59006427 on Build GRCh38
2765002 c.154C>T p.R52W Chr.20: 59006475-59006475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761643208 Chr.20: 59006495-59006495 on Build GRCh38
rs769000655 Chr.20: 59006362-59006362 on Build GRCh38
rs956042402 Chr.20: 59006266-59006266 on Build GRCh38
rs776414207 Chr.20: 59006422-59006422 on Build GRCh38
rs757883716 Chr.20: 59006433-59006433 on Build GRCh38
rs541770537 Chr.20: 59006300-59006300 on Build GRCh38
rs753235038 Chr.20: 59006529-59006529 on Build GRCh38
rs755559076 Chr.20: 59006457-59006457 on Build GRCh38
rs756915928 Chr.20: 59006339-59006339 on Build GRCh38
rs918868200 Chr.20: 59006547-59006547 on Build GRCh38
rs748592572 Chr.20: 59006461-59006461 on Build GRCh38
rs377705741 Chr.20: 59006307-59006307 on Build GRCh38
rs911364993 Chr.20: 59006357-59006357 on Build GRCh38
rs143686598 Chr.20: 59006507-59006507 on Build GRCh38
rs751089751 Chr.20: 59006510-59006510 on Build GRCh38
rs771023014 Chr.20: 59006398-59006398 on Build GRCh38
rs531348521 Chr.20: 59006546-59006546 on Build GRCh38
rs770148459 Chr.20: 59006386-59006386 on Build GRCh38
rs757910431 Chr.20: 59006349-59006349 on Build GRCh38
rs1039046428 Chr.20: 59006374-59006374 on Build GRCh38
rs750064793 Chr.20: 59006432-59006432 on Build GRCh38
rs756802063 Chr.20: 59006569-59006569 on Build GRCh38
rs774512938 Chr.20: 59006479-59006479 on Build GRCh38
rs374624286 Chr.20: 59006503-59006503 on Build GRCh38
rs763001927 Chr.20: 59006388-59006388 on Build GRCh38
rs768646415 Chr.20: 59006482-59006482 on Build GRCh38
rs1056165806 Chr.20: 59006562-59006562 on Build GRCh38
rs775329763 Chr.20: 59006415-59006415 on Build GRCh38
rs756536654 Chr.20: 59006462-59006462 on Build GRCh38
rs947677471 Chr.20: 59006630-59006630 on Build GRCh38
rs186380851 Chr.20: 59006454-59006454 on Build GRCh38
rs140246463 Chr.20: 59006442-59006442 on Build GRCh38
rs374531729 Chr.20: 59006279-59006279 on Build GRCh38
rs1023353788 Chr.20: 59006456-59006456 on Build GRCh38
rs769039356 Chr.20: 59006272-59006272 on Build GRCh38
rs74350367 Chr.20: 59006468-59006468 on Build GRCh38
rs916307553 Chr.20: 59006298-59006298 on Build GRCh38
rs773567578 Chr.20: 59006387-59006387 on Build GRCh38
rs763025483 Chr.20: 59006296-59006296 on Build GRCh38
rs764984182 Chr.20: 59006498-59006498 on Build GRCh38
rs764926142 Chr.20: 59006431-59006431 on Build GRCh38
rs898663425 Chr.20: 59006467-59006467 on Build GRCh38
rs1023427201 Chr.20: 59006533-59006533 on Build GRCh38
rs780701364 Chr.20: 59006600-59006600 on Build GRCh38
rs942918275 Chr.20: 59006684-59006684 on Build GRCh38
rs754533124 Chr.20: 59006513-59006513 on Build GRCh38
rs770268470 Chr.20: 59006688-59006688 on Build GRCh38
rs780471774 Chr.20: 59006355-59006355 on Build GRCh38
rs768200449 Chr.20: 59006519-59006519 on Build GRCh38
rs776534185 Chr.20: 59006486-59006486 on Build GRCh38
rs763960952 Chr.20: 59006420-59006420 on Build GRCh38
rs760699500 Chr.20: 59006418-59006418 on Build GRCh38
rs781710040 Chr.20: 59006368-59006368 on Build GRCh38
rs968302966 Chr.20: 59006544-59006544 on Build GRCh38
rs747515854 Chr.20: 59006361-59006361 on Build GRCh38
rs897367507 Chr.20: 59006384-59006384 on Build GRCh38
rs892403870 Chr.20: 59006660-59006660 on Build GRCh38
rs749046807 Chr.20: 59006452-59006453 on Build GRCh38
rs999912152 Chr.20: 59006320-59006320 on Build GRCh38
rs749496207 Chr.20: 59006473-59006473 on Build GRCh38
rs756589555 Chr.20: 59006536-59006536 on Build GRCh38
rs368487798 Chr.20: 59006343-59006343 on Build GRCh38
rs748470022 Chr.20: 59006381-59006381 on Build GRCh38
rs930209287 Chr.20: 59006554-59006554 on Build GRCh38
rs111756806 Chr.20: 59006439-59006439 on Build GRCh38
rs528315317 Chr.20: 59006635-59006635 on Build GRCh38
rs771018508 Chr.20: 59006478-59006478 on Build GRCh38
rs772838602 Chr.20: 59006500-59006500 on Build GRCh38
rs762153494 Chr.20: 59006280-59006280 on Build GRCh38
rs1041531145 Chr.20: 59006304-59006304 on Build GRCh38
rs775163478 Chr.20: 59006300-59006303 on Build GRCh38
rs57276474 Chr.20: 59006654-59006654 on Build GRCh38
rs774331858 Chr.20: 59006475-59006475 on Build GRCh38
rs1052270576 Chr.20: 59006659-59006659 on Build GRCh38
rs974236883 Chr.20: 59006543-59006543 on Build GRCh38
rs995581034 Chr.20: 59006475-59006475 on Build GRCh38
rs753591068 Chr.20: 59006338-59006338 on Build GRCh38
rs770097234 Chr.20: 59006283-59006283 on Build GRCh38
rs1056135344 Chr.20: 59006423-59006423 on Build GRCh38
rs903567675 Chr.20: 59006643-59006643 on Build GRCh38
rs781739366 Chr.20: 59006458-59006458 on Build GRCh38
rs1000664531 Chr.20: 59006499-59006499 on Build GRCh38
rs768543182 Chr.20: 59006457-59006457 on Build GRCh38
rs955483601 Chr.20: 59006511-59006511 on Build GRCh38
rs369011055 Chr.20: 59006324-59006324 on Build GRCh38
rs745398273 Chr.20: 59006622-59006622 on Build GRCh38
rs912238471 Chr.20: 59006673-59006675 on Build GRCh38
rs766672443 Chr.20: 59006344-59006344 on Build GRCh38
rs771143249 Chr.20: 59006298-59006299 on Build GRCh38
rs763846487 Chr.20: 59006335-59006335 on Build GRCh38
rs990689996 Chr.20: 59006244-59006244 on Build GRCh38
rs141636434 Chr.20: 59006293-59006293 on Build GRCh38
rs201197341 Chr.20: 59006435-59006435 on Build GRCh38

View Full Product Details