Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44104410-44104908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102724200
Entrez Gene ID
102724200
Gene Name
trafficking protein particle complex subunit 10-like
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.21: 5155503-5165755 on Build GRCh38
Transcripts
XM_011546082.2 NM_003274.4 XM_005261168.4 XM_011529714.2 XM_011529715.2 XM_011529716.2 XM_011529719.2 XM_011529721.2 XM_011529722.2 XM_011529723.2 XM_011529727.2 XM_011529728.2 XM_011529729.2 XM_017028454.1 XM_017028455.1

Gene Symbol
TRAPPC10
Entrez Gene ID
7109
Gene Name
trafficking protein particle complex 10
Gene Aliases
EHOC-1, EHOC1, GT334, TMEM1, TRS130, TRS30
UniGene
Hs.126221
Gene Chromosome Location
Chr.21: 44012325-44106552 on Build GRCh38
Transcripts
XM_011546082.2 NM_003274.4 XM_005261168.4 XM_011529714.2 XM_011529715.2 XM_011529716.2 XM_011529719.2 XM_011529721.2 XM_011529722.2 XM_011529723.2 XM_011529727.2 XM_011529728.2 XM_011529729.2 XM_017028454.1 XM_017028455.1

SNP Detail

SNP ID Chromosome Location
rs1017376400 Chr.21: 44104826-44104826 on Build GRCh38
rs969782154 Chr.21: 44104881-44104881 on Build GRCh38
rs997383905 Chr.21: 44104855-44104855 on Build GRCh38
rs1007321282 Chr.21: 44104767-44104767 on Build GRCh38
rs931340026 Chr.21: 44104735-44104735 on Build GRCh38
rs914334615 Chr.21: 44104620-44104620 on Build GRCh38
rs1027106510 Chr.21: 44104456-44104456 on Build GRCh38
rs924251057 Chr.21: 44104583-44104583 on Build GRCh38
rs143099716 Chr.21: 44104877-44104877 on Build GRCh38
rs375469118 Chr.21: 44104546-44104546 on Build GRCh38
rs760754654 Chr.21: 44104685-44104685 on Build GRCh38
rs938897555 Chr.21: 44104700-44104700 on Build GRCh38
rs1037326786 Chr.21: 44104709-44104709 on Build GRCh38
rs951939345 Chr.21: 44104486-44104486 on Build GRCh38
rs989834082 Chr.21: 44104868-44104868 on Build GRCh38
rs955770369 Chr.21: 44104588-44104588 on Build GRCh38
rs975998376 Chr.21: 44104565-44104565 on Build GRCh38
rs897141121 Chr.21: 44104718-44104718 on Build GRCh38
rs879820597 Chr.21: 44104833-44104833 on Build GRCh38
rs1017453581 Chr.21: 44104545-44104545 on Build GRCh38
rs149054837 Chr.21: 44104541-44104541 on Build GRCh38
rs767295181 Chr.21: 44104616-44104619 on Build GRCh38
rs901705936 Chr.21: 44104832-44104832 on Build GRCh38
rs889897857 Chr.21: 44104738-44104738 on Build GRCh38
rs561641204 Chr.21: 44104691-44104691 on Build GRCh38
rs535729514 Chr.21: 44104570-44104570 on Build GRCh38
rs181505185 Chr.21: 44104856-44104856 on Build GRCh38
rs990296693 Chr.21: 44104611-44104612 on Build GRCh38
rs952424577 Chr.21: 44104848-44104848 on Build GRCh38
rs554135966 Chr.21: 44104736-44104736 on Build GRCh38
rs948826201 Chr.21: 44104621-44104621 on Build GRCh38
rs955863185 Chr.21: 44104867-44104867 on Build GRCh38
rs980284359 Chr.21: 44104689-44104689 on Build GRCh38

View Full Product Details