Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38871739-38872230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX6
Entrez Gene ID
23466
Gene Name
chromobox 6
Gene Aliases
-
UniGene
Hs.592201
Gene Chromosome Location
Chr.22: 38861427-38872334 on Build GRCh38
Transcripts
NM_001303494.1 NM_014292.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391447 c.37G>A p.A13T Chr.22: 38872154-38872154 on Build GRCh38
6401084 c.14C>T p.A5V Chr.22: 38872177-38872177 on Build GRCh38
4104162 c.129G>A p.E43E Chr.22: 38871742-38871742 on Build GRCh38
6409096 c.25C>T p.R9W Chr.22: 38872166-38872166 on Build GRCh38
6409139 c.1A>G p.M1V Chr.22: 38872190-38872190 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs916827977 Chr.22: 38872082-38872082 on Build GRCh38
rs767538044 Chr.22: 38871907-38871907 on Build GRCh38
rs548732588 Chr.22: 38871792-38871792 on Build GRCh38
rs377030092 Chr.22: 38872201-38872201 on Build GRCh38
rs747463255 Chr.22: 38872146-38872146 on Build GRCh38
rs369727544 Chr.22: 38872171-38872171 on Build GRCh38
rs374109380 Chr.22: 38872182-38872182 on Build GRCh38
rs776037927 Chr.22: 38871801-38871801 on Build GRCh38
rs1055802885 Chr.22: 38871887-38871887 on Build GRCh38
rs756154140 Chr.22: 38871964-38871964 on Build GRCh38
rs1019291471 Chr.22: 38871974-38871974 on Build GRCh38
rs200823973 Chr.22: 38871871-38871871 on Build GRCh38
rs376837850 Chr.22: 38871860-38871860 on Build GRCh38
rs770476621 Chr.22: 38872176-38872176 on Build GRCh38
rs1031026291 Chr.22: 38872064-38872064 on Build GRCh38
rs764098190 Chr.22: 38872205-38872205 on Build GRCh38
rs757355510 Chr.22: 38872081-38872081 on Build GRCh38
rs978135728 Chr.22: 38872049-38872049 on Build GRCh38
rs772567928 Chr.22: 38871799-38871799 on Build GRCh38
rs769161449 Chr.22: 38872154-38872154 on Build GRCh38
rs762587207 Chr.22: 38872167-38872167 on Build GRCh38
rs546864798 Chr.22: 38872048-38872048 on Build GRCh38
rs767382636 Chr.22: 38872195-38872195 on Build GRCh38
rs772798557 Chr.22: 38871809-38871809 on Build GRCh38
rs769251388 Chr.22: 38871808-38871808 on Build GRCh38
rs990121551 Chr.22: 38871821-38871821 on Build GRCh38
rs997975787 Chr.22: 38871987-38871987 on Build GRCh38
rs1002852763 Chr.22: 38871874-38871874 on Build GRCh38
rs756309660 Chr.22: 38871761-38871761 on Build GRCh38
rs757419554 Chr.22: 38872206-38872206 on Build GRCh38
rs746208791 Chr.22: 38872119-38872119 on Build GRCh38
rs965116557 Chr.22: 38871976-38871976 on Build GRCh38
rs749433053 Chr.22: 38871764-38871764 on Build GRCh38
rs779265846 Chr.22: 38872105-38872105 on Build GRCh38
rs761293439 Chr.22: 38871805-38871805 on Build GRCh38
rs369275015 Chr.22: 38871760-38871760 on Build GRCh38
rs753965456 Chr.22: 38872077-38872077 on Build GRCh38
rs760632368 Chr.22: 38872203-38872203 on Build GRCh38
rs1003902881 Chr.22: 38871969-38871969 on Build GRCh38
rs1046569414 Chr.22: 38871847-38871847 on Build GRCh38
rs752719918 Chr.22: 38871962-38871962 on Build GRCh38
rs995809365 Chr.22: 38871954-38871954 on Build GRCh38
rs375620926 Chr.22: 38871770-38871770 on Build GRCh38
rs888090245 Chr.22: 38871948-38871948 on Build GRCh38
rs1041853610 Chr.22: 38871822-38871822 on Build GRCh38
rs772525943 Chr.22: 38872155-38872155 on Build GRCh38
rs772478012 Chr.22: 38872132-38872132 on Build GRCh38
rs948196379 Chr.22: 38871841-38871841 on Build GRCh38
rs899725366 Chr.22: 38871983-38871983 on Build GRCh38
rs777952127 Chr.22: 38872074-38872074 on Build GRCh38
rs946111348 Chr.22: 38871819-38871819 on Build GRCh38
rs369765424 Chr.22: 38871777-38871777 on Build GRCh38
rs1053995192 Chr.22: 38871796-38871796 on Build GRCh38
rs528397800 Chr.22: 38871888-38871888 on Build GRCh38
rs778112526 Chr.22: 38871763-38871763 on Build GRCh38
rs780509720 Chr.22: 38872134-38872134 on Build GRCh38
rs1029426854 Chr.22: 38872001-38872003 on Build GRCh38
rs751307195 Chr.22: 38871883-38871883 on Build GRCh38
rs571201104 Chr.22: 38872065-38872065 on Build GRCh38
rs369674368 Chr.22: 38871773-38871773 on Build GRCh38

View Full Product Details