Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39965783-39966281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRAP2
Entrez Gene ID
9402
Gene Name
GRB2-related adaptor protein 2
Gene Aliases
GADS, GRAP-2, GRB2L, GRBLG, GRID, GRPL, GrbX, Grf40, Mona, P38
UniGene
Hs.517499
Gene Chromosome Location
Chr.22: 39901073-39973343 on Build GRCh38
Transcripts
NM_001291824.1 NM_001291825.1 NM_001291826.1 NM_001291828.1 NM_004810.3 XM_006724376.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4753433 c.236T>C p.L79P Chr.22: 39966133-39966133 on Build GRCh38
234765 c.408C>T p.S136S Chr.22: 39966107-39966107 on Build GRCh38
4961003 c.399G>A p.R133R Chr.22: 39966098-39966098 on Build GRCh38
6840768 c.139G>C p.G47R Chr.22: 39966036-39966036 on Build GRCh38
4753432 c.434T>C p.L145P Chr.22: 39966133-39966133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs6001712 Chr.22: 39965933-39965933 on Build GRCh38
rs752379967 Chr.22: 39966164-39966164 on Build GRCh38
rs866441319 Chr.22: 39966070-39966070 on Build GRCh38
rs140074469 Chr.22: 39966148-39966148 on Build GRCh38
rs188608044 Chr.22: 39965921-39965921 on Build GRCh38
rs775865688 Chr.22: 39966106-39966106 on Build GRCh38
rs772467342 Chr.22: 39965975-39965975 on Build GRCh38
rs575041912 Chr.22: 39965859-39965859 on Build GRCh38
rs531056755 Chr.22: 39966178-39966178 on Build GRCh38
rs766948780 Chr.22: 39966163-39966163 on Build GRCh38
rs776685904 Chr.22: 39966184-39966184 on Build GRCh38
rs546986327 Chr.22: 39966239-39966239 on Build GRCh38
rs1009685469 Chr.22: 39966147-39966147 on Build GRCh38
rs201333393 Chr.22: 39966207-39966207 on Build GRCh38
rs1052674350 Chr.22: 39965883-39965883 on Build GRCh38
rs139269286 Chr.22: 39966145-39966145 on Build GRCh38
rs267606253 Chr.22: 39966136-39966136 on Build GRCh38
rs768711558 Chr.22: 39966161-39966162 on Build GRCh38
rs367728516 Chr.22: 39966075-39966075 on Build GRCh38
rs144299118 Chr.22: 39966065-39966065 on Build GRCh38
rs372341383 Chr.22: 39966181-39966181 on Build GRCh38
rs769173032 Chr.22: 39966138-39966138 on Build GRCh38
rs968196034 Chr.22: 39965906-39965906 on Build GRCh38
rs527914191 Chr.22: 39966226-39966226 on Build GRCh38
rs543434560 Chr.22: 39965868-39965868 on Build GRCh38
rs959853233 Chr.22: 39965928-39965928 on Build GRCh38
rs773774988 Chr.22: 39966200-39966200 on Build GRCh38
rs370974940 Chr.22: 39965945-39965945 on Build GRCh38
rs765826788 Chr.22: 39966040-39966040 on Build GRCh38
rs561341837 Chr.22: 39966220-39966220 on Build GRCh38
rs769826010 Chr.22: 39966242-39966242 on Build GRCh38
rs536016771 Chr.22: 39965885-39965885 on Build GRCh38
rs200044407 Chr.22: 39966002-39966002 on Build GRCh38
rs904066689 Chr.22: 39965835-39965835 on Build GRCh38
rs372740629 Chr.22: 39966034-39966034 on Build GRCh38
rs754398099 Chr.22: 39966025-39966025 on Build GRCh38
rs991180549 Chr.22: 39965930-39965930 on Build GRCh38
rs749209183 Chr.22: 39966082-39966082 on Build GRCh38
rs753712017 Chr.22: 39966173-39966173 on Build GRCh38
rs775847200 Chr.22: 39965984-39965984 on Build GRCh38
rs1009625468 Chr.22: 39965903-39965903 on Build GRCh38
rs764493218 Chr.22: 39966003-39966003 on Build GRCh38
rs115997833 Chr.22: 39965953-39965953 on Build GRCh38
rs182871357 Chr.22: 39965905-39965905 on Build GRCh38
rs763676052 Chr.22: 39966103-39966103 on Build GRCh38
rs1039884888 Chr.22: 39965830-39965830 on Build GRCh38
rs748004836 Chr.22: 39965939-39965939 on Build GRCh38
rs187242644 Chr.22: 39965820-39965820 on Build GRCh38
rs778689725 Chr.22: 39966090-39966090 on Build GRCh38
rs1023990699 Chr.22: 39965937-39965937 on Build GRCh38
rs979715715 Chr.22: 39965912-39965912 on Build GRCh38
rs759516733 Chr.22: 39965966-39965966 on Build GRCh38
rs755717851 Chr.22: 39966166-39966166 on Build GRCh38
rs149610155 Chr.22: 39966041-39966041 on Build GRCh38
rs775441624 Chr.22: 39966068-39966068 on Build GRCh38
rs375513433 Chr.22: 39966198-39966198 on Build GRCh38
rs376214658 Chr.22: 39966183-39966183 on Build GRCh38
rs563770584 Chr.22: 39966177-39966177 on Build GRCh38
rs778895360 Chr.22: 39965901-39965901 on Build GRCh38
rs916873850 Chr.22: 39965931-39965931 on Build GRCh38
rs777479486 Chr.22: 39965917-39965917 on Build GRCh38
rs141561569 Chr.22: 39965915-39965915 on Build GRCh38
rs374234748 Chr.22: 39965956-39965956 on Build GRCh38
rs757270442 Chr.22: 39966083-39966083 on Build GRCh38
rs1001126436 Chr.22: 39965842-39965842 on Build GRCh38
rs780881602 Chr.22: 39966058-39966058 on Build GRCh38
rs747157900 Chr.22: 39966107-39966107 on Build GRCh38
rs754414010 Chr.22: 39966048-39966048 on Build GRCh38
rs745674279 Chr.22: 39966017-39966017 on Build GRCh38
rs751130456 Chr.22: 39966045-39966045 on Build GRCh38
rs772261912 Chr.22: 39966101-39966101 on Build GRCh38
rs372514119 Chr.22: 39965954-39965954 on Build GRCh38
rs189744680 Chr.22: 39965862-39965862 on Build GRCh38
rs759098932 Chr.22: 39966152-39966152 on Build GRCh38
rs755910417 Chr.22: 39966067-39966067 on Build GRCh38
rs371510448 Chr.22: 39966099-39966099 on Build GRCh38
rs1007555387 Chr.22: 39965840-39965840 on Build GRCh38
rs1001741379 Chr.22: 39965962-39965962 on Build GRCh38
rs761805277 Chr.22: 39966180-39966180 on Build GRCh38
rs369236185 Chr.22: 39966185-39966185 on Build GRCh38
rs945691756 Chr.22: 39966246-39966246 on Build GRCh38

View Full Product Details