Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43946073-43946581 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNPLA3
Entrez Gene ID
80339
Gene Name
patatin like phospholipase domain containing 3
Gene Aliases
ADPN, C22orf20, iPLA(2)epsilon
UniGene
Hs.654800
Gene Chromosome Location
Chr.22: 43923739-43947568 on Build GRCh38
Transcripts
NM_025225.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
276830 c.1220C>T p.S407F Chr.22: 43946156-43946156 on Build GRCh38
5816218 c.1254A>G p.P418P Chr.22: 43946190-43946190 on Build GRCh38
3740458 c.1417C>T p.P473S Chr.22: 43946353-43946353 on Build GRCh38
4716232 c.1296C>A p.S432S Chr.22: 43946232-43946232 on Build GRCh38
5393808 c.1382A>G p.K461R Chr.22: 43946318-43946318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780726605 Chr.22: 43946220-43946220 on Build GRCh38
rs747490781 Chr.22: 43946224-43946224 on Build GRCh38
rs768161377 Chr.22: 43946329-43946329 on Build GRCh38
rs762674387 Chr.22: 43946147-43946147 on Build GRCh38
rs6006460 Chr.22: 43946294-43946294 on Build GRCh38
rs745504551 Chr.22: 43946308-43946308 on Build GRCh38
rs200019431 Chr.22: 43946153-43946153 on Build GRCh38
rs141084368 Chr.22: 43946270-43946270 on Build GRCh38
rs781298809 Chr.22: 43946157-43946157 on Build GRCh38
rs750469596 Chr.22: 43946203-43946203 on Build GRCh38
rs774075869 Chr.22: 43946344-43946344 on Build GRCh38
rs200503449 Chr.22: 43946122-43946122 on Build GRCh38
rs145821885 Chr.22: 43946238-43946238 on Build GRCh38
rs371569586 Chr.22: 43946287-43946287 on Build GRCh38
rs749470981 Chr.22: 43946115-43946115 on Build GRCh38
rs769195391 Chr.22: 43946235-43946235 on Build GRCh38
rs767510066 Chr.22: 43946378-43946379 on Build GRCh38
rs763225620 Chr.22: 43946370-43946370 on Build GRCh38
rs1027790067 Chr.22: 43946486-43946486 on Build GRCh38
rs1057199550 Chr.22: 43946546-43946546 on Build GRCh38
rs745563266 Chr.22: 43946416-43946416 on Build GRCh38
rs1046784151 Chr.22: 43946557-43946557 on Build GRCh38
rs569866825 Chr.22: 43946129-43946129 on Build GRCh38
rs760581974 Chr.22: 43946271-43946271 on Build GRCh38
rs373173858 Chr.22: 43946412-43946412 on Build GRCh38
rs111933596 Chr.22: 43946154-43946154 on Build GRCh38
rs147289545 Chr.22: 43946276-43946276 on Build GRCh38
rs753209557 Chr.22: 43946398-43946398 on Build GRCh38
rs1041140276 Chr.22: 43946101-43946101 on Build GRCh38
rs115043594 Chr.22: 43946514-43946514 on Build GRCh38
rs766637347 Chr.22: 43946375-43946375 on Build GRCh38
rs546817766 Chr.22: 43946225-43946225 on Build GRCh38
rs774966241 Chr.22: 43946315-43946315 on Build GRCh38
rs747015303 Chr.22: 43946320-43946320 on Build GRCh38
rs575255100 Chr.22: 43946120-43946120 on Build GRCh38
rs374810102 Chr.22: 43946200-43946200 on Build GRCh38
rs749439408 Chr.22: 43946293-43946293 on Build GRCh38
rs149925560 Chr.22: 43946174-43946174 on Build GRCh38
rs2294918 Chr.22: 43946236-43946236 on Build GRCh38
rs773041966 Chr.22: 43946132-43946132 on Build GRCh38
rs557722575 Chr.22: 43946452-43946452 on Build GRCh38
rs927963508 Chr.22: 43946368-43946368 on Build GRCh38
rs768584834 Chr.22: 43946332-43946332 on Build GRCh38
rs377420917 Chr.22: 43946119-43946119 on Build GRCh38
rs138667813 Chr.22: 43946239-43946239 on Build GRCh38
rs773357812 Chr.22: 43946362-43946362 on Build GRCh38
rs759020903 Chr.22: 43946164-43946164 on Build GRCh38
rs138696154 Chr.22: 43946099-43946099 on Build GRCh38
rs866388723 Chr.22: 43946541-43946541 on Build GRCh38
rs2294917 Chr.22: 43946106-43946106 on Build GRCh38
rs763503205 Chr.22: 43946547-43946547 on Build GRCh38
rs927153737 Chr.22: 43946532-43946532 on Build GRCh38
rs753625728 Chr.22: 43946274-43946274 on Build GRCh38
rs758444417 Chr.22: 43946216-43946216 on Build GRCh38
rs76510336 Chr.22: 43946295-43946295 on Build GRCh38
rs754106300 Chr.22: 43946197-43946197 on Build GRCh38
rs144014738 Chr.22: 43946114-43946114 on Build GRCh38
rs771618618 Chr.22: 43946312-43946312 on Build GRCh38
rs368899768 Chr.22: 43946397-43946397 on Build GRCh38
rs766547732 Chr.22: 43946284-43946284 on Build GRCh38
rs539980744 Chr.22: 43946487-43946487 on Build GRCh38
rs757396903 Chr.22: 43946116-43946116 on Build GRCh38
rs906592148 Chr.22: 43946185-43946190 on Build GRCh38
rs569255289 Chr.22: 43946401-43946401 on Build GRCh38
rs765777650 Chr.22: 43946103-43946103 on Build GRCh38
rs758992807 Chr.22: 43946251-43946251 on Build GRCh38
rs907815767 Chr.22: 43946133-43946133 on Build GRCh38
rs773997072 Chr.22: 43946248-43946248 on Build GRCh38
rs1005088043 Chr.22: 43946529-43946529 on Build GRCh38
rs376208148 Chr.22: 43946355-43946355 on Build GRCh38
rs913043512 Chr.22: 43946476-43946476 on Build GRCh38
rs367919387 Chr.22: 43946267-43946267 on Build GRCh38
rs201650776 Chr.22: 43946158-43946158 on Build GRCh38
rs752584334 Chr.22: 43946188-43946188 on Build GRCh38
rs886057601 Chr.22: 43946453-43946453 on Build GRCh38
rs993971795 Chr.22: 43946333-43946333 on Build GRCh38
rs974540521 Chr.22: 43946110-43946110 on Build GRCh38
rs976740888 Chr.22: 43946243-43946243 on Build GRCh38
rs56680842 Chr.22: 43946121-43946121 on Build GRCh38
rs557141181 Chr.22: 43946300-43946300 on Build GRCh38
rs769128146 Chr.22: 43946269-43946270 on Build GRCh38
rs760758882 Chr.22: 43946555-43946555 on Build GRCh38
rs147312179 Chr.22: 43946301-43946303 on Build GRCh38
rs763844316 Chr.22: 43946190-43946190 on Build GRCh38
rs751698167 Chr.22: 43946386-43946386 on Build GRCh38
rs774779678 Chr.22: 43946299-43946300 on Build GRCh38
rs2294919 Chr.22: 43946445-43946445 on Build GRCh38
rs143404144 Chr.22: 43946275-43946275 on Build GRCh38
rs6006461 Chr.22: 43946433-43946433 on Build GRCh38
rs140909509 Chr.22: 43946390-43946392 on Build GRCh38
rs144968908 Chr.22: 43946189-43946189 on Build GRCh38
rs752494490 Chr.22: 43946107-43946107 on Build GRCh38
rs773733628 Chr.22: 43946161-43946161 on Build GRCh38
rs757406991 Chr.22: 43946198-43946198 on Build GRCh38
rs532304239 Chr.22: 43946266-43946266 on Build GRCh38
rs201268858 Chr.22: 43946273-43946273 on Build GRCh38
rs755847307 Chr.22: 43946108-43946108 on Build GRCh38
rs886057602 Chr.22: 43946523-43946523 on Build GRCh38
rs911263670 Chr.22: 43946097-43946097 on Build GRCh38
rs947173899 Chr.22: 43946493-43946493 on Build GRCh38
rs760429303 Chr.22: 43946455-43946455 on Build GRCh38
rs957001096 Chr.22: 43946453-43946453 on Build GRCh38
rs953074677 Chr.22: 43946228-43946228 on Build GRCh38
rs761531694 Chr.22: 43946354-43946354 on Build GRCh38
rs995874006 Chr.22: 43946465-43946465 on Build GRCh38
rs570274837 Chr.22: 43946552-43946552 on Build GRCh38
rs758435999 Chr.22: 43946279-43946279 on Build GRCh38
rs778463214 Chr.22: 43946302-43946302 on Build GRCh38
rs751568967 Chr.22: 43946104-43946104 on Build GRCh38
rs769739852 Chr.22: 43946212-43946212 on Build GRCh38

View Full Product Details