Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45195205-45195710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0930
Entrez Gene ID
23313
Gene Name
KIAA0930
Gene Aliases
C22orf9, LSC3
UniGene
Hs.592207
Gene Chromosome Location
Chr.22: 45192242-45240769 on Build GRCh38
Transcripts
NM_001009880.1 NM_015264.1 XM_011530017.2 XM_017028672.1

SNP Detail

SNP ID Chromosome Location
rs148259746 Chr.22: 45195562-45195562 on Build GRCh38
rs930150350 Chr.22: 45195635-45195635 on Build GRCh38
rs574722156 Chr.22: 45195671-45195671 on Build GRCh38
rs895973428 Chr.22: 45195634-45195634 on Build GRCh38
rs879218344 Chr.22: 45195670-45195670 on Build GRCh38
rs5845698 Chr.22: 45195672-45195673 on Build GRCh38
rs960618100 Chr.22: 45195572-45195572 on Build GRCh38
rs1052343212 Chr.22: 45195285-45195285 on Build GRCh38
rs528280544 Chr.22: 45195331-45195331 on Build GRCh38
rs540341730 Chr.22: 45195319-45195319 on Build GRCh38
rs1038119399 Chr.22: 45195669-45195669 on Build GRCh38
rs888706737 Chr.22: 45195639-45195639 on Build GRCh38
rs923501343 Chr.22: 45195652-45195657 on Build GRCh38
rs911317636 Chr.22: 45195236-45195236 on Build GRCh38
rs1038550647 Chr.22: 45195555-45195555 on Build GRCh38
rs115040787 Chr.22: 45195633-45195633 on Build GRCh38
rs574893523 Chr.22: 45195283-45195283 on Build GRCh38
rs935254221 Chr.22: 45195277-45195277 on Build GRCh38
rs922440158 Chr.22: 45195533-45195533 on Build GRCh38
rs559616162 Chr.22: 45195322-45195322 on Build GRCh38
rs114184899 Chr.22: 45195534-45195534 on Build GRCh38
rs1051144636 Chr.22: 45195653-45195653 on Build GRCh38
rs1018329214 Chr.22: 45195510-45195510 on Build GRCh38
rs1047319837 Chr.22: 45195636-45195636 on Build GRCh38
rs926359933 Chr.22: 45195580-45195580 on Build GRCh38
rs1005771864 Chr.22: 45195326-45195326 on Build GRCh38
rs397775556 Chr.22: 45195677-45195678 on Build GRCh38
rs994413472 Chr.22: 45195476-45195476 on Build GRCh38
rs567115957 Chr.22: 45195521-45195521 on Build GRCh38
rs968378140 Chr.22: 45195677-45195677 on Build GRCh38
rs147381538 Chr.22: 45195600-45195600 on Build GRCh38
rs993964914 Chr.22: 45195674-45195675 on Build GRCh38
rs977001807 Chr.22: 45195255-45195255 on Build GRCh38
rs142728186 Chr.22: 45195591-45195591 on Build GRCh38
rs976570916 Chr.22: 45195520-45195520 on Build GRCh38
rs1005983096 Chr.22: 45195506-45195506 on Build GRCh38
rs115943311 Chr.22: 45195414-45195414 on Build GRCh38
rs773751922 Chr.22: 45195325-45195325 on Build GRCh38
rs946404554 Chr.22: 45195307-45195307 on Build GRCh38
rs1049421908 Chr.22: 45195543-45195543 on Build GRCh38
rs1012638638 Chr.22: 45195588-45195588 on Build GRCh38
rs997839870 Chr.22: 45195282-45195282 on Build GRCh38
rs149545544 Chr.22: 45195353-45195353 on Build GRCh38
rs1005286891 Chr.22: 45195549-45195549 on Build GRCh38
rs946380782 Chr.22: 45195567-45195567 on Build GRCh38
rs761463870 Chr.22: 45195332-45195332 on Build GRCh38
rs914903094 Chr.22: 45195290-45195290 on Build GRCh38
rs561087767 Chr.22: 45195261-45195261 on Build GRCh38
rs1040634676 Chr.22: 45195256-45195256 on Build GRCh38
rs553856626 Chr.22: 45195664-45195665 on Build GRCh38
rs184234402 Chr.22: 45195642-45195642 on Build GRCh38
rs532086472 Chr.22: 45195260-45195260 on Build GRCh38
rs1034976498 Chr.22: 45195367-45195367 on Build GRCh38
rs768235530 Chr.22: 45195251-45195251 on Build GRCh38
rs932263071 Chr.22: 45195652-45195652 on Build GRCh38
rs964481119 Chr.22: 45195515-45195515 on Build GRCh38
rs575273426 Chr.22: 45195249-45195249 on Build GRCh38
rs554971860 Chr.22: 45195665-45195665 on Build GRCh38
rs988006812 Chr.22: 45195539-45195539 on Build GRCh38
rs889503310 Chr.22: 45195547-45195547 on Build GRCh38
rs1029732682 Chr.22: 45195667-45195667 on Build GRCh38
rs879608346 Chr.22: 45195291-45195291 on Build GRCh38
rs1045144213 Chr.22: 45195568-45195568 on Build GRCh38
rs1001683025 Chr.22: 45195405-45195405 on Build GRCh38
rs963886415 Chr.22: 45195248-45195248 on Build GRCh38
rs550197949 Chr.22: 45195643-45195643 on Build GRCh38
rs952723193 Chr.22: 45195484-45195484 on Build GRCh38
rs546265643 Chr.22: 45195561-45195561 on Build GRCh38
rs1022185397 Chr.22: 45195675-45195675 on Build GRCh38
rs200631815 Chr.22: 45195680-45195680 on Build GRCh38
rs899872077 Chr.22: 45195659-45195659 on Build GRCh38
rs941064678 Chr.22: 45195660-45195660 on Build GRCh38
rs530669543 Chr.22: 45195410-45195410 on Build GRCh38
rs896025873 Chr.22: 45195444-45195444 on Build GRCh38
rs1025929612 Chr.22: 45195479-45195479 on Build GRCh38
rs565175239 Chr.22: 45195393-45195393 on Build GRCh38
rs946840693 Chr.22: 45195447-45195447 on Build GRCh38

View Full Product Details