Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32502519-32503014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMTM6
Entrez Gene ID
54918
Gene Name
CKLF like MARVEL transmembrane domain containing 6
Gene Aliases
CKLFSF6, PRO2219
UniGene
Hs.380627
Gene Chromosome Location
Chr.3: 32481312-32502911 on Build GRCh38
Transcripts
NM_017801.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7329084 c.45G>T p.P15P Chr.3: 32502701-32502701 on Build GRCh38
1617483 c.76G>A p.A26T Chr.3: 32502670-32502670 on Build GRCh38
4116892 c.75C>T p.L25L Chr.3: 32502671-32502671 on Build GRCh38
5640743 c.32C>A p.T11K Chr.3: 32502714-32502714 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs929232234 Chr.3: 32502976-32502976 on Build GRCh38
rs755589271 Chr.3: 32502791-32502791 on Build GRCh38
rs760787827 Chr.3: 32502721-32502721 on Build GRCh38
rs746401911 Chr.3: 32502711-32502711 on Build GRCh38
rs747810007 Chr.3: 32502648-32502648 on Build GRCh38
rs200575664 Chr.3: 32502667-32502667 on Build GRCh38
rs765141219 Chr.3: 32502635-32502635 on Build GRCh38
rs769365885 Chr.3: 32502651-32502651 on Build GRCh38
rs201598355 Chr.3: 32502655-32502655 on Build GRCh38
rs1039306152 Chr.3: 32502984-32502984 on Build GRCh38
rs771321238 Chr.3: 32502704-32502704 on Build GRCh38
rs556719622 Chr.3: 32502716-32502716 on Build GRCh38
rs943514359 Chr.3: 32502972-32502972 on Build GRCh38
rs751392119 Chr.3: 32502719-32502719 on Build GRCh38
rs768216145 Chr.3: 32502575-32502575 on Build GRCh38
rs956569134 Chr.3: 32502628-32502628 on Build GRCh38
rs1033556985 Chr.3: 32502845-32502845 on Build GRCh38
rs527713087 Chr.3: 32502636-32502636 on Build GRCh38
rs967618036 Chr.3: 32502884-32502884 on Build GRCh38
rs766629780 Chr.3: 32502555-32502555 on Build GRCh38
rs977537450 Chr.3: 32502900-32502900 on Build GRCh38
rs749653928 Chr.3: 32502702-32502702 on Build GRCh38
rs780681502 Chr.3: 32502695-32502695 on Build GRCh38
rs890730931 Chr.3: 32502752-32502752 on Build GRCh38
rs548662937 Chr.3: 32502566-32502566 on Build GRCh38
rs779344571 Chr.3: 32502707-32502707 on Build GRCh38
rs759545061 Chr.3: 32502777-32502777 on Build GRCh38
rs766869852 Chr.3: 32502586-32502586 on Build GRCh38
rs758194560 Chr.3: 32502686-32502686 on Build GRCh38
rs745791021 Chr.3: 32502563-32502563 on Build GRCh38
rs112518454 Chr.3: 32502578-32502578 on Build GRCh38
rs957134056 Chr.3: 32502917-32502917 on Build GRCh38
rs763454560 Chr.3: 32502581-32502581 on Build GRCh38
rs766207744 Chr.3: 32502773-32502773 on Build GRCh38
rs939063133 Chr.3: 32502893-32502893 on Build GRCh38
rs929094714 Chr.3: 32502822-32502822 on Build GRCh38
rs998145500 Chr.3: 32502859-32502859 on Build GRCh38
rs539878478 Chr.3: 32502905-32502905 on Build GRCh38
rs773593849 Chr.3: 32502654-32502654 on Build GRCh38
rs949993232 Chr.3: 32502811-32502811 on Build GRCh38
rs1025698966 Chr.3: 32502611-32502611 on Build GRCh38
rs1048942045 Chr.3: 32502843-32502844 on Build GRCh38
rs760984074 Chr.3: 32502681-32502681 on Build GRCh38
rs779062964 Chr.3: 32502558-32502558 on Build GRCh38
rs767669810 Chr.3: 32502616-32502616 on Build GRCh38
rs775744829 Chr.3: 32502717-32502717 on Build GRCh38
rs762726629 Chr.3: 32502764-32502764 on Build GRCh38
rs773505998 Chr.3: 32502579-32502579 on Build GRCh38
rs1029837668 Chr.3: 32502867-32502867 on Build GRCh38
rs980691650 Chr.3: 32502844-32502844 on Build GRCh38
rs897752713 Chr.3: 32502815-32502815 on Build GRCh38
rs1039294821 Chr.3: 32502799-32502799 on Build GRCh38
rs568520192 Chr.3: 32502783-32502783 on Build GRCh38
rs776326488 Chr.3: 32502576-32502576 on Build GRCh38
rs774314029 Chr.3: 32502587-32502587 on Build GRCh38
rs981116871 Chr.3: 32502966-32502966 on Build GRCh38
rs993332569 Chr.3: 32502590-32502590 on Build GRCh38
rs899485198 Chr.3: 32502543-32502543 on Build GRCh38
rs769052520 Chr.3: 32502751-32502751 on Build GRCh38
rs919362523 Chr.3: 32502861-32502861 on Build GRCh38
rs777381010 Chr.3: 32502699-32502699 on Build GRCh38
rs926656879 Chr.3: 32502677-32502677 on Build GRCh38
rs892179474 Chr.3: 32502824-32502824 on Build GRCh38
rs779439090 Chr.3: 32502564-32502564 on Build GRCh38
rs759559567 Chr.3: 32502694-32502694 on Build GRCh38
rs746487933 Chr.3: 32502570-32502570 on Build GRCh38
rs899871447 Chr.3: 32502857-32502857 on Build GRCh38
rs1025250216 Chr.3: 32502832-32502832 on Build GRCh38
rs755781406 Chr.3: 32502698-32502698 on Build GRCh38
rs1006079554 Chr.3: 32502848-32502848 on Build GRCh38
rs759634931 Chr.3: 32502610-32502610 on Build GRCh38
rs1033622097 Chr.3: 32502915-32502915 on Build GRCh38
rs867658185 Chr.3: 32502897-32502897 on Build GRCh38
rs949666258 Chr.3: 32502958-32502958 on Build GRCh38
rs368361590 Chr.3: 32502641-32502641 on Build GRCh38
rs199588531 Chr.3: 32502644-32502644 on Build GRCh38
rs776885415 Chr.3: 32502754-32502754 on Build GRCh38
rs963352933 Chr.3: 32502854-32502854 on Build GRCh38
rs764372570 Chr.3: 32502684-32502684 on Build GRCh38
rs915665865 Chr.3: 32502948-32502948 on Build GRCh38
rs536210258 Chr.3: 32502638-32502638 on Build GRCh38
rs759598730 Chr.3: 32502671-32502671 on Build GRCh38
rs765523861 Chr.3: 32502650-32502651 on Build GRCh38
rs899538064 Chr.3: 32502807-32502807 on Build GRCh38
rs758298649 Chr.3: 32502637-32502637 on Build GRCh38
rs912098138 Chr.3: 32502928-32502928 on Build GRCh38
rs991684784 Chr.3: 32502938-32502938 on Build GRCh38
rs987555960 Chr.3: 32502869-32502869 on Build GRCh38
rs1009419139 Chr.3: 32502829-32502829 on Build GRCh38
rs929359933 Chr.3: 32502862-32502862 on Build GRCh38
rs752961094 Chr.3: 32502625-32502625 on Build GRCh38
rs1032097688 Chr.3: 32502823-32502823 on Build GRCh38
rs535956550 Chr.3: 32502789-32502789 on Build GRCh38
rs928814938 Chr.3: 32502889-32502889 on Build GRCh38
rs970524144 Chr.3: 32502843-32502843 on Build GRCh38
rs980264693 Chr.3: 32502883-32502883 on Build GRCh38
rs775655055 Chr.3: 32502679-32502679 on Build GRCh38
rs572582645 Chr.3: 32502855-32502855 on Build GRCh38
rs554214710 Chr.3: 32502812-32502812 on Build GRCh38
rs757179723 Chr.3: 32502631-32502631 on Build GRCh38
rs751243628 Chr.3: 32502776-32502776 on Build GRCh38
rs750270505 Chr.3: 32502685-32502685 on Build GRCh38
rs762155822 Chr.3: 32502646-32502650 on Build GRCh38
rs970593113 Chr.3: 32502649-32502649 on Build GRCh38
rs747614297 Chr.3: 32502697-32502697 on Build GRCh38
rs892094534 Chr.3: 32502911-32502911 on Build GRCh38
rs113000765 Chr.3: 32502827-32502827 on Build GRCh38
rs767624070 Chr.3: 32502673-32502673 on Build GRCh38

View Full Product Details