Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47845620-47846120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX30
Entrez Gene ID
22907
Gene Name
DEAH-box helicase 30
Gene Aliases
DDX30, RETCOR
UniGene
Hs.517948
Gene Chromosome Location
Chr.3: 47802909-47850196 on Build GRCh38
Transcripts
NM_014966.3 NM_138615.2 XM_006713033.1 XM_011533490.1 XM_011533491.1 XM_011533492.2 XM_011533493.1 XM_011533494.2 XM_011533495.1 XM_011533496.1 XM_011533497.2 XM_011533498.1 XM_017005914.1 XM_017005915.1 XM_017005916.1 XM_017005917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3069705 c.972G>A p.P324P Chr.3: 47845732-47845732 on Build GRCh38
6279315 c.1043T>C p.I348T Chr.3: 47845803-47845803 on Build GRCh38
1670144 c.971C>T p.P324L Chr.3: 47845731-47845731 on Build GRCh38
7426175 c.1049T>C p.V350A Chr.3: 47845809-47845809 on Build GRCh38
3069703 c.966C>T p.N322N Chr.3: 47845726-47845726 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974711417 Chr.3: 47845681-47845681 on Build GRCh38
rs566786160 Chr.3: 47845826-47845826 on Build GRCh38
rs781207480 Chr.3: 47845964-47845964 on Build GRCh38
rs764406446 Chr.3: 47845834-47845834 on Build GRCh38
rs752225413 Chr.3: 47845950-47845950 on Build GRCh38
rs377728207 Chr.3: 47845901-47845901 on Build GRCh38
rs563983650 Chr.3: 47845758-47845758 on Build GRCh38
rs774630669 Chr.3: 47845791-47845791 on Build GRCh38
rs879082872 Chr.3: 47846016-47846016 on Build GRCh38
rs746927412 Chr.3: 47845741-47845741 on Build GRCh38
rs781022286 Chr.3: 47845763-47845763 on Build GRCh38
rs545956398 Chr.3: 47846050-47846050 on Build GRCh38
rs749516789 Chr.3: 47845771-47845771 on Build GRCh38
rs575490259 Chr.3: 47845981-47845981 on Build GRCh38
rs180914153 Chr.3: 47845951-47845951 on Build GRCh38
rs529468329 Chr.3: 47845670-47845670 on Build GRCh38
rs777049115 Chr.3: 47845838-47845838 on Build GRCh38
rs753191163 Chr.3: 47845688-47845688 on Build GRCh38
rs755916078 Chr.3: 47845895-47845895 on Build GRCh38
rs568710065 Chr.3: 47845649-47845649 on Build GRCh38
rs780919567 Chr.3: 47845882-47845882 on Build GRCh38
rs144616702 Chr.3: 47845813-47845813 on Build GRCh38
rs979130723 Chr.3: 47845931-47845931 on Build GRCh38
rs982909961 Chr.3: 47845694-47845694 on Build GRCh38
rs370881771 Chr.3: 47845852-47845852 on Build GRCh38
rs367847892 Chr.3: 47845732-47845732 on Build GRCh38
rs542138275 Chr.3: 47845916-47845916 on Build GRCh38
rs903063850 Chr.3: 47845893-47845893 on Build GRCh38
rs1018299432 Chr.3: 47845947-47845947 on Build GRCh38
rs79691392 Chr.3: 47846087-47846087 on Build GRCh38
rs766786959 Chr.3: 47845880-47845880 on Build GRCh38
rs756870874 Chr.3: 47845691-47845691 on Build GRCh38
rs1008252488 Chr.3: 47845940-47845940 on Build GRCh38
rs569099891 Chr.3: 47845719-47845719 on Build GRCh38
rs200116835 Chr.3: 47845884-47845884 on Build GRCh38
rs759703914 Chr.3: 47845795-47845795 on Build GRCh38
rs951585746 Chr.3: 47846091-47846091 on Build GRCh38
rs947366878 Chr.3: 47845877-47845877 on Build GRCh38
rs548954335 Chr.3: 47845932-47845932 on Build GRCh38
rs974385575 Chr.3: 47846059-47846059 on Build GRCh38
rs550722288 Chr.3: 47845674-47845674 on Build GRCh38
rs138599337 Chr.3: 47845738-47845738 on Build GRCh38
rs757843620 Chr.3: 47845726-47845726 on Build GRCh38
rs771274801 Chr.3: 47845787-47845787 on Build GRCh38
rs778458349 Chr.3: 47845693-47845693 on Build GRCh38
rs376062811 Chr.3: 47845690-47845690 on Build GRCh38
rs200777275 Chr.3: 47845839-47845839 on Build GRCh38
rs113176570 Chr.3: 47845651-47845651 on Build GRCh38
rs772213163 Chr.3: 47845804-47845804 on Build GRCh38
rs201596740 Chr.3: 47845747-47845747 on Build GRCh38
rs760188602 Chr.3: 47845678-47845678 on Build GRCh38
rs771188861 Chr.3: 47845672-47845672 on Build GRCh38
rs964388442 Chr.3: 47846018-47846018 on Build GRCh38
rs764879143 Chr.3: 47845840-47845840 on Build GRCh38
rs373090001 Chr.3: 47845861-47845861 on Build GRCh38
rs751503282 Chr.3: 47845865-47845865 on Build GRCh38
rs768127027 Chr.3: 47845686-47845686 on Build GRCh38
rs964354219 Chr.3: 47845671-47845671 on Build GRCh38
rs750008150 Chr.3: 47845843-47845843 on Build GRCh38
rs773774044 Chr.3: 47845754-47845754 on Build GRCh38

View Full Product Details