Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 108823447-108823990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKNAD1
Entrez Gene ID
254268
Gene Name
AKNA domain containing 1
Gene Aliases
C1orf62
UniGene
Hs.729441
Gene Chromosome Location
Chr.1: 108815898-108858242 on Build GRCh38
Transcripts
NM_152763.4 NR_049760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5341357 c.1181-8C>A p.? Chr.1: 108823485-108823485 on Build GRCh38
6849352 c.1103T>A p.M368K Chr.1: 108823643-108823643 on Build GRCh38
462534 c.2020C>T p.P674S Chr.1: 108823605-108823605 on Build GRCh38
7128365 c.1966G>C p.D656H Chr.1: 108823659-108823659 on Build GRCh38
3470741 c.2043G>A p.R681R Chr.1: 108823582-108823582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs924060778 Chr.1: 108823894-108823894 on Build GRCh38
rs757944789 Chr.1: 108823907-108823907 on Build GRCh38
rs893728731 Chr.1: 108823728-108823728 on Build GRCh38
rs7522260 Chr.1: 108823748-108823748 on Build GRCh38
rs764888120 Chr.1: 108823687-108823687 on Build GRCh38
rs201331558 Chr.1: 108823501-108823501 on Build GRCh38
rs578047945 Chr.1: 108823858-108823858 on Build GRCh38
rs751751513 Chr.1: 108823555-108823555 on Build GRCh38
rs768258990 Chr.1: 108823606-108823606 on Build GRCh38
rs922122327 Chr.1: 108823835-108823835 on Build GRCh38
rs761307678 Chr.1: 108823531-108823531 on Build GRCh38
rs544795464 Chr.1: 108823829-108823829 on Build GRCh38
rs769733436 Chr.1: 108823629-108823629 on Build GRCh38
rs986044647 Chr.1: 108823944-108823944 on Build GRCh38
rs747057495 Chr.1: 108823488-108823488 on Build GRCh38
rs372858211 Chr.1: 108823862-108823862 on Build GRCh38
rs766079377 Chr.1: 108823559-108823560 on Build GRCh38
rs866205366 Chr.1: 108823586-108823586 on Build GRCh38
rs770288962 Chr.1: 108823595-108823595 on Build GRCh38
rs375189165 Chr.1: 108823728-108823728 on Build GRCh38
rs570067338 Chr.1: 108823527-108823527 on Build GRCh38
rs758604466 Chr.1: 108823485-108823485 on Build GRCh38
rs762704903 Chr.1: 108823486-108823486 on Build GRCh38
rs7529592 Chr.1: 108823932-108823932 on Build GRCh38
rs563345431 Chr.1: 108823900-108823900 on Build GRCh38
rs749464471 Chr.1: 108823823-108823823 on Build GRCh38
rs774317628 Chr.1: 108823571-108823572 on Build GRCh38
rs988020500 Chr.1: 108823893-108823893 on Build GRCh38
rs879165246 Chr.1: 108823495-108823495 on Build GRCh38
rs555768237 Chr.1: 108823972-108823972 on Build GRCh38
rs1003728100 Chr.1: 108823767-108823767 on Build GRCh38
rs1005224808 Chr.1: 108823812-108823812 on Build GRCh38
rs748320153 Chr.1: 108823474-108823474 on Build GRCh38
rs373993251 Chr.1: 108823536-108823536 on Build GRCh38
rs777767037 Chr.1: 108823486-108823486 on Build GRCh38
rs1053755147 Chr.1: 108823921-108823921 on Build GRCh38
rs7522157 Chr.1: 108823664-108823664 on Build GRCh38
rs759372780 Chr.1: 108823609-108823610 on Build GRCh38
rs34261306 Chr.1: 108823697-108823698 on Build GRCh38
rs1022771195 Chr.1: 108823804-108823804 on Build GRCh38
rs369057849 Chr.1: 108823691-108823691 on Build GRCh38
rs911369139 Chr.1: 108823950-108823950 on Build GRCh38
rs539054570 Chr.1: 108823540-108823540 on Build GRCh38
rs1032747327 Chr.1: 108823827-108823827 on Build GRCh38
rs935375872 Chr.1: 108823896-108823896 on Build GRCh38
rs989089684 Chr.1: 108823532-108823532 on Build GRCh38
rs773772608 Chr.1: 108823529-108823529 on Build GRCh38
rs762537260 Chr.1: 108823524-108823524 on Build GRCh38
rs368846627 Chr.1: 108823549-108823549 on Build GRCh38
rs767613407 Chr.1: 108823615-108823615 on Build GRCh38
rs569635426 Chr.1: 108823654-108823654 on Build GRCh38
rs35042482 Chr.1: 108823622-108823623 on Build GRCh38
rs767500612 Chr.1: 108823534-108823534 on Build GRCh38
rs140828781 Chr.1: 108823504-108823504 on Build GRCh38
rs1025411459 Chr.1: 108823784-108823784 on Build GRCh38
rs555304765 Chr.1: 108823719-108823719 on Build GRCh38
rs771295327 Chr.1: 108823813-108823813 on Build GRCh38
rs906363416 Chr.1: 108823760-108823760 on Build GRCh38
rs376071319 Chr.1: 108823567-108823567 on Build GRCh38
rs558295181 Chr.1: 108823831-108823831 on Build GRCh38
rs373573870 Chr.1: 108823537-108823537 on Build GRCh38
rs746409597 Chr.1: 108823592-108823592 on Build GRCh38
rs1044912077 Chr.1: 108823749-108823749 on Build GRCh38
rs534849031 Chr.1: 108823796-108823796 on Build GRCh38
rs759366789 Chr.1: 108823681-108823681 on Build GRCh38
rs988470800 Chr.1: 108823865-108823865 on Build GRCh38
rs750511664 Chr.1: 108823722-108823722 on Build GRCh38
rs372310040 Chr.1: 108823659-108823659 on Build GRCh38
rs144702847 Chr.1: 108823585-108823585 on Build GRCh38
rs999973279 Chr.1: 108823815-108823815 on Build GRCh38
rs144674627 Chr.1: 108823510-108823510 on Build GRCh38
rs753645797 Chr.1: 108823675-108823675 on Build GRCh38
rs774039698 Chr.1: 108823640-108823640 on Build GRCh38
rs749631562 Chr.1: 108823596-108823596 on Build GRCh38
rs771636516 Chr.1: 108823490-108823490 on Build GRCh38
rs778054998 Chr.1: 108823551-108823551 on Build GRCh38
rs761393524 Chr.1: 108823646-108823646 on Build GRCh38
rs751835529 Chr.1: 108823689-108823689 on Build GRCh38

View Full Product Details