Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 68987304-68987806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EOGT
Entrez Gene ID
285203
Gene Name
EGF domain specific O-linked N-acetylglucosamine transferase
Gene Aliases
AER61, AOS4, C3orf64, EOGT1
UniGene
Hs.518059
Gene Chromosome Location
Chr.3: 68975212-69013961 on Build GRCh38
Transcripts
NM_001278689.1 NM_173654.2 NR_103826.1 XM_005264743.2 XM_011533599.1 XM_011533600.1 XM_011533602.1 XM_011533603.1 XM_017006204.1 XM_017006205.1 XM_017006206.1 XM_017006207.1 XM_017006209.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5399981 c.1128C>T p.Y376Y Chr.3: 68987469-68987469 on Build GRCh38
6836099 c.859G>A p.A287T Chr.3: 68987486-68987486 on Build GRCh38
6469083 c.1097G>A p.R366Q Chr.3: 68987500-68987500 on Build GRCh38
7316959 c.898G>A p.E300K Chr.3: 68987447-68987447 on Build GRCh38
5399978 c.877C>T p.R293W Chr.3: 68987468-68987468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs557801015 Chr.3: 68987738-68987738 on Build GRCh38
rs990013253 Chr.3: 68987641-68987641 on Build GRCh38
rs4855350 Chr.3: 68987631-68987631 on Build GRCh38
rs887236618 Chr.3: 68987629-68987629 on Build GRCh38
rs1025396890 Chr.3: 68987588-68987588 on Build GRCh38
rs1029720801 Chr.3: 68987700-68987700 on Build GRCh38
rs754383563 Chr.3: 68987392-68987392 on Build GRCh38
rs1038154571 Chr.3: 68987645-68987645 on Build GRCh38
rs939576033 Chr.3: 68987603-68987603 on Build GRCh38
rs200861184 Chr.3: 68987386-68987387 on Build GRCh38
rs943039027 Chr.3: 68987413-68987413 on Build GRCh38
rs4855542 Chr.3: 68987556-68987556 on Build GRCh38
rs922937753 Chr.3: 68987705-68987705 on Build GRCh38
rs536272903 Chr.3: 68987365-68987365 on Build GRCh38
rs955840820 Chr.3: 68987637-68987640 on Build GRCh38
rs763639987 Chr.3: 68987526-68987526 on Build GRCh38
rs1054393955 Chr.3: 68987384-68987385 on Build GRCh38
rs928254998 Chr.3: 68987579-68987579 on Build GRCh38
rs113070216 Chr.3: 68987562-68987562 on Build GRCh38
rs367720571 Chr.3: 68987543-68987543 on Build GRCh38
rs573156239 Chr.3: 68987445-68987445 on Build GRCh38
rs186870285 Chr.3: 68987483-68987483 on Build GRCh38
rs772839750 Chr.3: 68987777-68987777 on Build GRCh38
rs578006231 Chr.3: 68987744-68987744 on Build GRCh38
rs4855540 Chr.3: 68987527-68987527 on Build GRCh38
rs749798424 Chr.3: 68987477-68987477 on Build GRCh38
rs552195809 Chr.3: 68987779-68987779 on Build GRCh38
rs762670672 Chr.3: 68987731-68987731 on Build GRCh38
rs970800984 Chr.3: 68987591-68987591 on Build GRCh38
rs587776995 Chr.3: 68987467-68987467 on Build GRCh38
rs998739425 Chr.3: 68987732-68987732 on Build GRCh38
rs369359608 Chr.3: 68987361-68987361 on Build GRCh38
rs764931480 Chr.3: 68987497-68987497 on Build GRCh38
rs754434944 Chr.3: 68987557-68987557 on Build GRCh38
rs757889169 Chr.3: 68987501-68987501 on Build GRCh38
rs748404665 Chr.3: 68987486-68987486 on Build GRCh38
rs1047225362 Chr.3: 68987620-68987620 on Build GRCh38
rs755494763 Chr.3: 68987561-68987561 on Build GRCh38
rs560995851 Chr.3: 68987594-68987594 on Build GRCh38
rs763593027 Chr.3: 68987444-68987444 on Build GRCh38
rs899569492 Chr.3: 68987652-68987652 on Build GRCh38
rs559915653 Chr.3: 68987723-68987723 on Build GRCh38
rs759321842 Chr.3: 68987494-68987494 on Build GRCh38
rs756878687 Chr.3: 68987453-68987453 on Build GRCh38
rs752166441 Chr.3: 68987416-68987416 on Build GRCh38
rs768921770 Chr.3: 68987478-68987478 on Build GRCh38
rs77006427 Chr.3: 68987386-68987386 on Build GRCh38
rs377612780 Chr.3: 68987468-68987468 on Build GRCh38
rs996552158 Chr.3: 68987699-68987699 on Build GRCh38
rs766337013 Chr.3: 68987736-68987736 on Build GRCh38
rs891063290 Chr.3: 68987712-68987712 on Build GRCh38
rs768193075 Chr.3: 68987460-68987460 on Build GRCh38
rs4855541 Chr.3: 68987532-68987532 on Build GRCh38
rs540289397 Chr.3: 68987655-68987655 on Build GRCh38
rs113464671 Chr.3: 68987409-68987409 on Build GRCh38
rs747630324 Chr.3: 68987720-68987720 on Build GRCh38
rs924445828 Chr.3: 68987357-68987357 on Build GRCh38
rs369402395 Chr.3: 68987410-68987410 on Build GRCh38
rs76406029 Chr.3: 68987387-68987387 on Build GRCh38
rs1056603885 Chr.3: 68987336-68987337 on Build GRCh38
rs774552774 Chr.3: 68987485-68987485 on Build GRCh38
rs778595083 Chr.3: 68987550-68987565 on Build GRCh38
rs568513385 Chr.3: 68987374-68987375 on Build GRCh38
rs11331317 Chr.3: 68987374-68987374 on Build GRCh38
rs769011820 Chr.3: 68987396-68987396 on Build GRCh38
rs989975493 Chr.3: 68987755-68987755 on Build GRCh38
rs765155300 Chr.3: 68987415-68987415 on Build GRCh38
rs79339018 Chr.3: 68987385-68987385 on Build GRCh38
rs780551902 Chr.3: 68987473-68987473 on Build GRCh38
rs756582694 Chr.3: 68987469-68987469 on Build GRCh38
rs772379253 Chr.3: 68987489-68987489 on Build GRCh38
rs774771385 Chr.3: 68987398-68987398 on Build GRCh38
rs775566374 Chr.3: 68987500-68987500 on Build GRCh38
rs756865843 Chr.3: 68987541-68987542 on Build GRCh38
rs373777136 Chr.3: 68987431-68987431 on Build GRCh38
rs781429232 Chr.3: 68987457-68987457 on Build GRCh38
rs943321106 Chr.3: 68987660-68987660 on Build GRCh38
rs374985812 Chr.3: 68987399-68987399 on Build GRCh38
rs1012244937 Chr.3: 68987559-68987559 on Build GRCh38
rs370375193 Chr.3: 68987531-68987531 on Build GRCh38
rs370667366 Chr.3: 68987770-68987770 on Build GRCh38

View Full Product Details