Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 111885166-111885659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHLDB2
Entrez Gene ID
90102
Gene Name
pleckstrin homology like domain family B member 2
Gene Aliases
LL5b, LL5beta
UniGene
Hs.603252
Gene Chromosome Location
Chr.3: 111732480-111976517 on Build GRCh38
Transcripts
NM_001134437.1 NM_001134438.1 NM_001134439.1 NM_145753.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770521 c.1174G>A p.E392K Chr.3: 111885251-111885251 on Build GRCh38
81578 c.1179A>T p.A393A Chr.3: 111885256-111885256 on Build GRCh38
339137 c.1229G>A p.R410Q Chr.3: 111885306-111885306 on Build GRCh38
5397789 c.1224G>A p.R408R Chr.3: 111885220-111885220 on Build GRCh38
3585259 c.1315G>A p.E439K Chr.3: 111885392-111885392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777053959 Chr.3: 111885519-111885519 on Build GRCh38
rs767866327 Chr.3: 111885220-111885220 on Build GRCh38
rs768074664 Chr.3: 111885301-111885301 on Build GRCh38
rs749335252 Chr.3: 111885363-111885363 on Build GRCh38
rs769827679 Chr.3: 111885273-111885273 on Build GRCh38
rs774830644 Chr.3: 111885215-111885215 on Build GRCh38
rs577197997 Chr.3: 111885376-111885376 on Build GRCh38
rs777616487 Chr.3: 111885334-111885334 on Build GRCh38
rs764871118 Chr.3: 111885381-111885381 on Build GRCh38
rs535288937 Chr.3: 111885217-111885217 on Build GRCh38
rs928239102 Chr.3: 111885584-111885584 on Build GRCh38
rs989183428 Chr.3: 111885479-111885479 on Build GRCh38
rs267599544 Chr.3: 111885510-111885510 on Build GRCh38
rs767419226 Chr.3: 111885552-111885552 on Build GRCh38
rs559808708 Chr.3: 111885483-111885483 on Build GRCh38
rs745884447 Chr.3: 111885192-111885192 on Build GRCh38
rs752410885 Chr.3: 111885406-111885406 on Build GRCh38
rs1044600622 Chr.3: 111885437-111885437 on Build GRCh38
rs372210288 Chr.3: 111885198-111885198 on Build GRCh38
rs772379628 Chr.3: 111885368-111885368 on Build GRCh38
rs200217328 Chr.3: 111885197-111885197 on Build GRCh38
rs749566354 Chr.3: 111885290-111885290 on Build GRCh38
rs776803098 Chr.3: 111885188-111885188 on Build GRCh38
rs375225872 Chr.3: 111885366-111885366 on Build GRCh38
rs757192748 Chr.3: 111885346-111885346 on Build GRCh38
rs765534865 Chr.3: 111885428-111885428 on Build GRCh38
rs764819834 Chr.3: 111885322-111885322 on Build GRCh38
rs747896198 Chr.3: 111885253-111885253 on Build GRCh38
rs755177651 Chr.3: 111885458-111885458 on Build GRCh38
rs751333062 Chr.3: 111885422-111885422 on Build GRCh38
rs765447584 Chr.3: 111885626-111885626 on Build GRCh38
rs1027737017 Chr.3: 111885614-111885614 on Build GRCh38
rs775505480 Chr.3: 111885207-111885207 on Build GRCh38
rs939570359 Chr.3: 111885589-111885589 on Build GRCh38
rs146769860 Chr.3: 111885203-111885203 on Build GRCh38
rs756101034 Chr.3: 111885611-111885611 on Build GRCh38
rs145708919 Chr.3: 111885211-111885211 on Build GRCh38
rs772308395 Chr.3: 111885471-111885471 on Build GRCh38
rs548444031 Chr.3: 111885539-111885539 on Build GRCh38
rs750444525 Chr.3: 111885568-111885568 on Build GRCh38
rs988369230 Chr.3: 111885628-111885628 on Build GRCh38
rs370817906 Chr.3: 111885446-111885446 on Build GRCh38
rs747350608 Chr.3: 111885495-111885495 on Build GRCh38
rs201400841 Chr.3: 111885429-111885429 on Build GRCh38
rs752500653 Chr.3: 111885327-111885327 on Build GRCh38
rs759923986 Chr.3: 111885369-111885369 on Build GRCh38
rs774881867 Chr.3: 111885293-111885293 on Build GRCh38
rs778719861 Chr.3: 111885248-111885248 on Build GRCh38
rs776139862 Chr.3: 111885372-111885372 on Build GRCh38
rs763756227 Chr.3: 111885536-111885536 on Build GRCh38
rs866346015 Chr.3: 111885338-111885338 on Build GRCh38
rs771438959 Chr.3: 111885518-111885518 on Build GRCh38
rs769349319 Chr.3: 111885529-111885529 on Build GRCh38
rs745961915 Chr.3: 111885271-111885271 on Build GRCh38
rs753547789 Chr.3: 111885247-111885247 on Build GRCh38
rs773660300 Chr.3: 111885305-111885305 on Build GRCh38
rs889165572 Chr.3: 111885619-111885619 on Build GRCh38
rs202130294 Chr.3: 111885210-111885210 on Build GRCh38
rs755514961 Chr.3: 111885239-111885239 on Build GRCh38
rs749966444 Chr.3: 111885234-111885234 on Build GRCh38
rs771385895 Chr.3: 111885370-111885370 on Build GRCh38
rs762028335 Chr.3: 111885297-111885297 on Build GRCh38
rs765909957 Chr.3: 111885244-111885244 on Build GRCh38
rs200776359 Chr.3: 111885431-111885431 on Build GRCh38
rs575409620 Chr.3: 111885291-111885291 on Build GRCh38
rs751505308 Chr.3: 111885342-111885342 on Build GRCh38
rs758381184 Chr.3: 111885259-111885259 on Build GRCh38
rs562413511 Chr.3: 111885612-111885612 on Build GRCh38
rs562803042 Chr.3: 111885530-111885530 on Build GRCh38
rs763409044 Chr.3: 111885594-111885594 on Build GRCh38
rs569992771 Chr.3: 111885563-111885565 on Build GRCh38
rs201726032 Chr.3: 111885311-111885311 on Build GRCh38
rs777511475 Chr.3: 111885262-111885262 on Build GRCh38
rs201280788 Chr.3: 111885289-111885289 on Build GRCh38
rs779212442 Chr.3: 111885365-111885365 on Build GRCh38
rs148966541 Chr.3: 111885408-111885408 on Build GRCh38
rs758059422 Chr.3: 111885329-111885329 on Build GRCh38
rs563713509 Chr.3: 111885312-111885312 on Build GRCh38
rs760028436 Chr.3: 111885524-111885524 on Build GRCh38
rs748369031 Chr.3: 111885466-111885466 on Build GRCh38
rs779124420 Chr.3: 111885462-111885462 on Build GRCh38
rs969961718 Chr.3: 111885485-111885485 on Build GRCh38
rs762549500 Chr.3: 111885532-111885532 on Build GRCh38
rs897811796 Chr.3: 111885601-111885601 on Build GRCh38
rs368489081 Chr.3: 111885410-111885410 on Build GRCh38
rs1027768536 Chr.3: 111885341-111885341 on Build GRCh38
rs202073284 Chr.3: 111885189-111885189 on Build GRCh38
rs761232469 Chr.3: 111885309-111885309 on Build GRCh38
rs1004209008 Chr.3: 111885278-111885278 on Build GRCh38
rs780315916 Chr.3: 111885349-111885349 on Build GRCh38

View Full Product Details