Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122254107-122254618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASR
Entrez Gene ID
846
Gene Name
calcium sensing receptor
Gene Aliases
CAR, EIG8, FHH, FIH, GPRC2A, HHC, HHC1, HYPOC1, NSHPT, PCAR1
UniGene
Hs.435615
Gene Chromosome Location
Chr.3: 122183683-122286503 on Build GRCh38
Transcripts
NM_000388.3 NM_001178065.1 XM_005247837.2 XM_006713789.3 XM_017007324.1 XM_017007325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
140461 c.5C>A p.A2E Chr.3: 122254194-122254194 on Build GRCh38
5695148 c.16T>C p.C6R Chr.3: 122254205-122254205 on Build GRCh38
727903 c.67G>A p.D23N Chr.3: 122254256-122254256 on Build GRCh38
6888572 c.100C>T p.L34F Chr.3: 122254289-122254289 on Build GRCh38
4112732 c.91G>A p.D31N Chr.3: 122254280-122254280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558522564 Chr.3: 122254243-122254243 on Build GRCh38
rs767054957 Chr.3: 122254162-122254162 on Build GRCh38
rs769932724 Chr.3: 122254235-122254235 on Build GRCh38
rs199869938 Chr.3: 122254264-122254264 on Build GRCh38
rs753659949 Chr.3: 122254180-122254180 on Build GRCh38
rs200355690 Chr.3: 122254169-122254169 on Build GRCh38
rs201033211 Chr.3: 122254396-122254396 on Build GRCh38
rs200673016 Chr.3: 122254221-122254221 on Build GRCh38
rs1037138469 Chr.3: 122254400-122254400 on Build GRCh38
rs193922424 Chr.3: 122254355-122254355 on Build GRCh38
rs761576251 Chr.3: 122254246-122254246 on Build GRCh38
rs201564143 Chr.3: 122254249-122254249 on Build GRCh38
rs889629885 Chr.3: 122254461-122254461 on Build GRCh38
rs759033373 Chr.3: 122254399-122254399 on Build GRCh38
rs144604474 Chr.3: 122254285-122254285 on Build GRCh38
rs979596307 Chr.3: 122254259-122254259 on Build GRCh38
rs540394617 Chr.3: 122254452-122254452 on Build GRCh38
rs766213548 Chr.3: 122254527-122254527 on Build GRCh38
rs199515839 Chr.3: 122254229-122254229 on Build GRCh38
rs201869423 Chr.3: 122254384-122254384 on Build GRCh38
rs183425314 Chr.3: 122254567-122254567 on Build GRCh38
rs191165880 Chr.3: 122254416-122254416 on Build GRCh38
rs779995504 Chr.3: 122254323-122254323 on Build GRCh38
rs61733590 Chr.3: 122254303-122254303 on Build GRCh38
rs766308258 Chr.3: 122254172-122254172 on Build GRCh38
rs201731619 Chr.3: 122254226-122254226 on Build GRCh38
rs1007197554 Chr.3: 122254468-122254468 on Build GRCh38
rs193922420 Chr.3: 122254295-122254295 on Build GRCh38
rs769021483 Chr.3: 122254339-122254339 on Build GRCh38
rs141880581 Chr.3: 122254216-122254216 on Build GRCh38
rs764310311 Chr.3: 122254141-122254141 on Build GRCh38
rs886041823 Chr.3: 122254297-122254298 on Build GRCh38
rs104893717 Chr.3: 122254227-122254227 on Build GRCh38
rs202135908 Chr.3: 122254153-122254153 on Build GRCh38
rs1015309336 Chr.3: 122254431-122254431 on Build GRCh38
rs756322119 Chr.3: 122254318-122254318 on Build GRCh38
rs762859143 Chr.3: 122254425-122254425 on Build GRCh38
rs200756928 Chr.3: 122254366-122254366 on Build GRCh38
rs772906030 Chr.3: 122254368-122254368 on Build GRCh38
rs568902441 Chr.3: 122254263-122254263 on Build GRCh38
rs960031739 Chr.3: 122254522-122254522 on Build GRCh38
rs368421439 Chr.3: 122254158-122254158 on Build GRCh38
rs121909265 Chr.3: 122254374-122254374 on Build GRCh38
rs754531742 Chr.3: 122254571-122254571 on Build GRCh38
rs200018529 Chr.3: 122254354-122254354 on Build GRCh38
rs988514233 Chr.3: 122254532-122254533 on Build GRCh38
rs371119165 Chr.3: 122254489-122254489 on Build GRCh38
rs758232331 Chr.3: 122254287-122254287 on Build GRCh38
rs778565441 Chr.3: 122254188-122254188 on Build GRCh38
rs997520806 Chr.3: 122254581-122254581 on Build GRCh38
rs201633414 Chr.3: 122254262-122254262 on Build GRCh38
rs867848185 Chr.3: 122254133-122254133 on Build GRCh38
rs996249687 Chr.3: 122254340-122254340 on Build GRCh38
rs866899753 Chr.3: 122254316-122254316 on Build GRCh38
rs397514729 Chr.3: 122254274-122254274 on Build GRCh38
rs199849269 Chr.3: 122254389-122254389 on Build GRCh38
rs765124679 Chr.3: 122254282-122254282 on Build GRCh38
rs200520980 Chr.3: 122254237-122254237 on Build GRCh38
rs763383822 Chr.3: 122254139-122254139 on Build GRCh38
rs966820424 Chr.3: 122254252-122254252 on Build GRCh38
rs775979608 Chr.3: 122254387-122254387 on Build GRCh38
rs199730938 Chr.3: 122254181-122254181 on Build GRCh38
rs121909262 Chr.3: 122254304-122254304 on Build GRCh38
rs931087409 Chr.3: 122254441-122254441 on Build GRCh38
rs201439187 Chr.3: 122254176-122254176 on Build GRCh38
rs1019531190 Chr.3: 122254521-122254521 on Build GRCh38
rs554770681 Chr.3: 122254379-122254379 on Build GRCh38
rs781573002 Chr.3: 122254297-122254297 on Build GRCh38
rs387907398 Chr.3: 122254288-122254288 on Build GRCh38
rs202129996 Chr.3: 122254404-122254404 on Build GRCh38
rs104893702 Chr.3: 122254330-122254330 on Build GRCh38
rs868678822 Chr.3: 122254276-122254276 on Build GRCh38
rs779428235 Chr.3: 122254342-122254342 on Build GRCh38
rs77852524 Chr.3: 122254267-122254267 on Build GRCh38
rs112042188 Chr.3: 122254195-122254195 on Build GRCh38
rs764556468 Chr.3: 122254198-122254198 on Build GRCh38
rs755580987 Chr.3: 122254163-122254163 on Build GRCh38
rs751002064 Chr.3: 122254442-122254442 on Build GRCh38
rs1055397172 Chr.3: 122254279-122254279 on Build GRCh38
rs866536776 Chr.3: 122254261-122254261 on Build GRCh38
rs749748004 Chr.3: 122254242-122254242 on Build GRCh38
rs142566660 Chr.3: 122254510-122254510 on Build GRCh38
rs886041154 Chr.3: 122254353-122254353 on Build GRCh38
rs961335370 Chr.3: 122254432-122254432 on Build GRCh38

View Full Product Details