Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 136152645-136153084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSL2
Entrez Gene ID
55167
Gene Name
male-specific lethal 2 homolog (Drosophila)
Gene Aliases
MSL-2, MSL2L1, RNF184
UniGene
Hs.18631
Gene Chromosome Location
Chr.3: 136148918-136197300 on Build GRCh38
Transcripts
NM_001145417.1 NM_018133.3 XM_005247571.3 XM_006713684.3 XM_011512949.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4113843 c.212A>C p.K71T Chr.3: 136152669-136152669 on Build GRCh38
6192771 c.170C>G p.P57R Chr.3: 136152711-136152711 on Build GRCh38
3123274 c.192T>C p.H64H Chr.3: 136152689-136152689 on Build GRCh38
1257988 c.230T>C p.M77T Chr.3: 136152651-136152651 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759508047 Chr.3: 136152776-136152776 on Build GRCh38
rs924827714 Chr.3: 136152870-136152870 on Build GRCh38
rs367847791 Chr.3: 136152786-136152786 on Build GRCh38
rs556790764 Chr.3: 136152908-136152908 on Build GRCh38
rs771238389 Chr.3: 136152708-136152708 on Build GRCh38
rs746352999 Chr.3: 136152725-136152725 on Build GRCh38
rs767955407 Chr.3: 136152909-136152909 on Build GRCh38
rs1012571370 Chr.3: 136152986-136152986 on Build GRCh38
rs1056189000 Chr.3: 136152942-136152942 on Build GRCh38
rs139226713 Chr.3: 136152671-136152671 on Build GRCh38
rs1015739520 Chr.3: 136152944-136152944 on Build GRCh38
rs548715971 Chr.3: 136152778-136152778 on Build GRCh38
rs374566316 Chr.3: 136152750-136152750 on Build GRCh38
rs534399804 Chr.3: 136152840-136152855 on Build GRCh38
rs199724419 Chr.3: 136152780-136152780 on Build GRCh38
rs373259101 Chr.3: 136152872-136152872 on Build GRCh38
rs749544519 Chr.3: 136152705-136152705 on Build GRCh38
rs776191592 Chr.3: 136152744-136152744 on Build GRCh38
rs556360184 Chr.3: 136152961-136152961 on Build GRCh38
rs751834718 Chr.3: 136152742-136152743 on Build GRCh38
rs755339765 Chr.3: 136152743-136152743 on Build GRCh38
rs893399891 Chr.3: 136152901-136152901 on Build GRCh38
rs1054527042 Chr.3: 136152891-136152891 on Build GRCh38
rs777905901 Chr.3: 136152676-136152676 on Build GRCh38
rs750842892 Chr.3: 136152920-136152920 on Build GRCh38
rs934848536 Chr.3: 136152888-136152888 on Build GRCh38
rs981921751 Chr.3: 136152856-136152856 on Build GRCh38
rs959247470 Chr.3: 136152791-136152791 on Build GRCh38
rs959994890 Chr.3: 136152815-136152815 on Build GRCh38
rs554381086 Chr.3: 136152911-136152911 on Build GRCh38
rs868520650 Chr.3: 136152696-136152696 on Build GRCh38
rs576429819 Chr.3: 136152971-136152971 on Build GRCh38
rs567778128 Chr.3: 136152927-136152927 on Build GRCh38
rs762714930 Chr.3: 136152761-136152761 on Build GRCh38
rs545063858 Chr.3: 136152991-136152991 on Build GRCh38
rs372333467 Chr.3: 136152803-136152803 on Build GRCh38
rs552993001 Chr.3: 136152955-136152955 on Build GRCh38
rs754374896 Chr.3: 136153048-136153048 on Build GRCh38
rs953349065 Chr.3: 136153015-136153015 on Build GRCh38
rs772519872 Chr.3: 136152728-136152728 on Build GRCh38
rs963626433 Chr.3: 136152976-136152976 on Build GRCh38
rs911725019 Chr.3: 136152817-136152817 on Build GRCh38
rs995031092 Chr.3: 136153004-136153004 on Build GRCh38
rs1026785014 Chr.3: 136152973-136152973 on Build GRCh38
rs571930421 Chr.3: 136152866-136152866 on Build GRCh38
rs761344261 Chr.3: 136152747-136152747 on Build GRCh38
rs533000067 Chr.3: 136152743-136152743 on Build GRCh38
rs950197205 Chr.3: 136152847-136152847 on Build GRCh38
rs780757789 Chr.3: 136152778-136152778 on Build GRCh38
rs201142509 Chr.3: 136152767-136152767 on Build GRCh38
rs534396410 Chr.3: 136152877-136152877 on Build GRCh38
rs111872749 Chr.3: 136152832-136152832 on Build GRCh38
rs762519839 Chr.3: 136152787-136152787 on Build GRCh38
rs528925835 Chr.3: 136152768-136152768 on Build GRCh38
rs959277858 Chr.3: 136152985-136152985 on Build GRCh38
rs1044782527 Chr.3: 136152907-136152907 on Build GRCh38
rs201684396 Chr.3: 136152759-136152759 on Build GRCh38
rs552094684 Chr.3: 136152834-136152834 on Build GRCh38
rs149929074 Chr.3: 136152718-136152718 on Build GRCh38
rs188841777 Chr.3: 136153011-136153011 on Build GRCh38

View Full Product Details