Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 136194892-136195379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSL2
Entrez Gene ID
55167
Gene Name
male-specific lethal 2 homolog (Drosophila)
Gene Aliases
MSL-2, MSL2L1, RNF184
UniGene
Hs.18631
Gene Chromosome Location
Chr.3: 136148918-136197300 on Build GRCh38
Transcripts
NM_018133.3 XM_006713684.3 XM_011512949.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1308634 c.88G>A p.E30K Chr.3: 136195026-136195026 on Build GRCh38
6715849 c.46C>T p.L16L Chr.3: 136195068-136195068 on Build GRCh38
1257990 c.122C>A p.S41Y Chr.3: 136194992-136194992 on Build GRCh38
419810 c.25C>T p.L9F Chr.3: 136195089-136195089 on Build GRCh38
479467 c.63C>T p.D21D Chr.3: 136195051-136195051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201616513 Chr.3: 136195040-136195040 on Build GRCh38
rs184843008 Chr.3: 136194976-136194976 on Build GRCh38
rs773380451 Chr.3: 136195159-136195159 on Build GRCh38
rs562803687 Chr.3: 136195257-136195257 on Build GRCh38
rs755754080 Chr.3: 136195031-136195033 on Build GRCh38
rs373922143 Chr.3: 136195107-136195107 on Build GRCh38
rs527990310 Chr.3: 136195210-136195210 on Build GRCh38
rs199950006 Chr.3: 136194924-136194924 on Build GRCh38
rs554018170 Chr.3: 136195008-136195008 on Build GRCh38
rs771826380 Chr.3: 136195136-136195136 on Build GRCh38
rs377395169 Chr.3: 136195045-136195045 on Build GRCh38
rs779331024 Chr.3: 136194925-136194925 on Build GRCh38
rs561169948 Chr.3: 136195296-136195296 on Build GRCh38
rs147762572 Chr.3: 136195087-136195087 on Build GRCh38
rs539025229 Chr.3: 136195102-136195102 on Build GRCh38
rs761727879 Chr.3: 136195066-136195066 on Build GRCh38
rs772498419 Chr.3: 136194939-136194939 on Build GRCh38
rs979950459 Chr.3: 136195069-136195069 on Build GRCh38
rs764130883 Chr.3: 136194986-136194986 on Build GRCh38
rs758694480 Chr.3: 136195083-136195083 on Build GRCh38
rs911218457 Chr.3: 136195050-136195050 on Build GRCh38
rs771615352 Chr.3: 136195084-136195084 on Build GRCh38
rs990667550 Chr.3: 136195052-136195052 on Build GRCh38
rs762692724 Chr.3: 136194954-136194954 on Build GRCh38
rs747825923 Chr.3: 136195028-136195029 on Build GRCh38
rs757454650 Chr.3: 136194991-136194991 on Build GRCh38
rs761440902 Chr.3: 136195058-136195058 on Build GRCh38
rs763075171 Chr.3: 136195165-136195165 on Build GRCh38
rs983362425 Chr.3: 136195324-136195324 on Build GRCh38
rs368825672 Chr.3: 136195134-136195134 on Build GRCh38
rs753983030 Chr.3: 136194987-136194987 on Build GRCh38
rs746745785 Chr.3: 136195143-136195143 on Build GRCh38
rs922712611 Chr.3: 136195244-136195244 on Build GRCh38
rs530347560 Chr.3: 136195344-136195344 on Build GRCh38
rs1009383804 Chr.3: 136195185-136195185 on Build GRCh38
rs980343978 Chr.3: 136195301-136195301 on Build GRCh38
rs368335816 Chr.3: 136195011-136195011 on Build GRCh38
rs201994928 Chr.3: 136195133-136195133 on Build GRCh38
rs199804963 Chr.3: 136195034-136195034 on Build GRCh38
rs891387655 Chr.3: 136195209-136195209 on Build GRCh38
rs776558734 Chr.3: 136195065-136195065 on Build GRCh38
rs900247843 Chr.3: 136195183-136195183 on Build GRCh38
rs372572175 Chr.3: 136195144-136195144 on Build GRCh38
rs938615719 Chr.3: 136195320-136195320 on Build GRCh38
rs531204762 Chr.3: 136195342-136195343 on Build GRCh38
rs982986774 Chr.3: 136195302-136195302 on Build GRCh38
rs745561575 Chr.3: 136195041-136195041 on Build GRCh38
rs747546180 Chr.3: 136194950-136194950 on Build GRCh38
rs754355230 Chr.3: 136195211-136195211 on Build GRCh38
rs767380911 Chr.3: 136194973-136194973 on Build GRCh38
rs144330176 Chr.3: 136195068-136195068 on Build GRCh38
rs941733424 Chr.3: 136195345-136195345 on Build GRCh38
rs142711366 Chr.3: 136195036-136195036 on Build GRCh38
rs112876762 Chr.3: 136194981-136194981 on Build GRCh38
rs559027819 Chr.3: 136195346-136195346 on Build GRCh38
rs754038944 Chr.3: 136194923-136194923 on Build GRCh38
rs760123413 Chr.3: 136195232-136195232 on Build GRCh38
rs377312944 Chr.3: 136195146-136195146 on Build GRCh38
rs367758510 Chr.3: 136194927-136194927 on Build GRCh38
rs752714503 Chr.3: 136195338-136195338 on Build GRCh38
rs1017899675 Chr.3: 136195219-136195219 on Build GRCh38
rs914643600 Chr.3: 136195057-136195057 on Build GRCh38
rs545026038 Chr.3: 136195175-136195175 on Build GRCh38
rs977130674 Chr.3: 136195241-136195241 on Build GRCh38
rs752029292 Chr.3: 136195105-136195105 on Build GRCh38
rs750662226 Chr.3: 136194997-136194997 on Build GRCh38
rs968404576 Chr.3: 136195193-136195193 on Build GRCh38
rs907781937 Chr.3: 136195331-136195331 on Build GRCh38
rs780355075 Chr.3: 136194945-136194945 on Build GRCh38
rs78742560 Chr.3: 136195113-136195113 on Build GRCh38
rs910041482 Chr.3: 136195306-136195306 on Build GRCh38
rs772896027 Chr.3: 136194952-136194952 on Build GRCh38
rs760657756 Chr.3: 136194985-136194985 on Build GRCh38
rs202239635 Chr.3: 136194966-136194966 on Build GRCh38
rs558672518 Chr.3: 136194951-136194951 on Build GRCh38
rs370918466 Chr.3: 136194936-136194936 on Build GRCh38
rs368091887 Chr.3: 136195121-136195121 on Build GRCh38
rs775250284 Chr.3: 136195142-136195142 on Build GRCh38
rs1022159813 Chr.3: 136195290-136195290 on Build GRCh38
rs775306319 Chr.3: 136195048-136195048 on Build GRCh38
rs764073442 Chr.3: 136195060-136195060 on Build GRCh38
rs770550343 Chr.3: 136194964-136194964 on Build GRCh38
rs189207100 Chr.3: 136195191-136195191 on Build GRCh38
rs531289091 Chr.3: 136195158-136195158 on Build GRCh38
rs760598672 Chr.3: 136195051-136195051 on Build GRCh38
rs777854377 Chr.3: 136194922-136194922 on Build GRCh38
rs892186510 Chr.3: 136195182-136195182 on Build GRCh38
rs766871304 Chr.3: 136195169-136195169 on Build GRCh38
rs990164365 Chr.3: 136195258-136195258 on Build GRCh38
rs547653380 Chr.3: 136195253-136195253 on Build GRCh38
rs758649999 Chr.3: 136195000-136195000 on Build GRCh38
rs773752002 Chr.3: 136195012-136195012 on Build GRCh38

View Full Product Details