Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 115651156-115651687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VANGL1
Entrez Gene ID
81839
Gene Name
VANGL planar cell polarity protein 1
Gene Aliases
KITENIN, LPP2, STB2, STBM2
UniGene
Hs.515130
Gene Chromosome Location
Chr.1: 115641953-115698224 on Build GRCh38
Transcripts
NM_001172411.1 NM_001172412.1 NM_138959.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5052426 c.10G>T p.E4* Chr.1: 115651423-115651423 on Build GRCh38
5735591 c.16A>T p.T6S Chr.1: 115651429-115651429 on Build GRCh38
7237004 c.65G>C p.R22T Chr.1: 115651478-115651478 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs376085770 Chr.1: 115651568-115651568 on Build GRCh38
rs74114601 Chr.1: 115651231-115651231 on Build GRCh38
rs766585358 Chr.1: 115651463-115651463 on Build GRCh38
rs114530176 Chr.1: 115651645-115651645 on Build GRCh38
rs575493003 Chr.1: 115651464-115651464 on Build GRCh38
rs752292713 Chr.1: 115651646-115651646 on Build GRCh38
rs61800295 Chr.1: 115651202-115651202 on Build GRCh38
rs567103171 Chr.1: 115651222-115651222 on Build GRCh38
rs199829701 Chr.1: 115651261-115651262 on Build GRCh38
rs181583261 Chr.1: 115651299-115651299 on Build GRCh38
rs77820456 Chr.1: 115651260-115651262 on Build GRCh38
rs749084675 Chr.1: 115651425-115651425 on Build GRCh38
rs757277483 Chr.1: 115651418-115651418 on Build GRCh38
rs922100276 Chr.1: 115651201-115651201 on Build GRCh38
rs75153165 Chr.1: 115651261-115651261 on Build GRCh38
rs946690867 Chr.1: 115651377-115651377 on Build GRCh38
rs375618149 Chr.1: 115651385-115651385 on Build GRCh38
rs886045118 Chr.1: 115651390-115651390 on Build GRCh38
rs763338932 Chr.1: 115651474-115651474 on Build GRCh38
rs773778906 Chr.1: 115651375-115651375 on Build GRCh38
rs563187708 Chr.1: 115651199-115651199 on Build GRCh38
rs190129771 Chr.1: 115651194-115651194 on Build GRCh38
rs17034095 Chr.1: 115651200-115651200 on Build GRCh38
rs1026787029 Chr.1: 115651361-115651361 on Build GRCh38
rs773571876 Chr.1: 115651252-115651253 on Build GRCh38
rs754624480 Chr.1: 115651430-115651430 on Build GRCh38
rs565243870 Chr.1: 115651408-115651408 on Build GRCh38
rs371629555 Chr.1: 115651449-115651449 on Build GRCh38
rs754747814 Chr.1: 115651530-115651530 on Build GRCh38
rs201664253 Chr.1: 115651253-115651253 on Build GRCh38
rs753604042 Chr.1: 115651522-115651522 on Build GRCh38
rs185393860 Chr.1: 115651396-115651396 on Build GRCh38
rs767649933 Chr.1: 115651492-115651492 on Build GRCh38
rs995677803 Chr.1: 115651331-115651331 on Build GRCh38
rs367781047 Chr.1: 115651504-115651504 on Build GRCh38
rs778582067 Chr.1: 115651433-115651433 on Build GRCh38
rs774497976 Chr.1: 115651482-115651482 on Build GRCh38
rs969792382 Chr.1: 115651245-115651245 on Build GRCh38
rs750294518 Chr.1: 115651422-115651422 on Build GRCh38
rs764242947 Chr.1: 115651477-115651477 on Build GRCh38
rs562165896 Chr.1: 115651589-115651589 on Build GRCh38
rs371100144 Chr.1: 115651382-115651382 on Build GRCh38
rs868067084 Chr.1: 115651244-115651244 on Build GRCh38
rs988708153 Chr.1: 115651362-115651362 on Build GRCh38
rs371198389 Chr.1: 115651533-115651533 on Build GRCh38
rs771644956 Chr.1: 115651439-115651439 on Build GRCh38
rs918262035 Chr.1: 115651586-115651586 on Build GRCh38
rs910547261 Chr.1: 115651383-115651383 on Build GRCh38
rs765588688 Chr.1: 115651392-115651392 on Build GRCh38
rs1005872532 Chr.1: 115651212-115651212 on Build GRCh38
rs912169929 Chr.1: 115651572-115651572 on Build GRCh38
rs958072438 Chr.1: 115651460-115651460 on Build GRCh38
rs560377079 Chr.1: 115651257-115651257 on Build GRCh38
rs761884414 Chr.1: 115651485-115651485 on Build GRCh38
rs747880582 Chr.1: 115651532-115651532 on Build GRCh38
rs540959231 Chr.1: 115651574-115651574 on Build GRCh38
rs753093036 Chr.1: 115651395-115651395 on Build GRCh38
rs779768443 Chr.1: 115651423-115651423 on Build GRCh38
rs532678140 Chr.1: 115651598-115651598 on Build GRCh38
rs57758037 Chr.1: 115651261-115651261 on Build GRCh38
rs936757009 Chr.1: 115651191-115651191 on Build GRCh38
rs771589773 Chr.1: 115651378-115651378 on Build GRCh38
rs1036724994 Chr.1: 115651268-115651268 on Build GRCh38
rs772420429 Chr.1: 115651195-115651195 on Build GRCh38
rs764348621 Chr.1: 115651397-115651397 on Build GRCh38
rs755658402 Chr.1: 115651659-115651659 on Build GRCh38
rs539378733 Chr.1: 115651531-115651531 on Build GRCh38
rs371591397 Chr.1: 115651359-115651359 on Build GRCh38
rs551980874 Chr.1: 115651205-115651205 on Build GRCh38
rs981604903 Chr.1: 115651266-115651267 on Build GRCh38
rs750484436 Chr.1: 115651496-115651496 on Build GRCh38
rs777222583 Chr.1: 115651380-115651380 on Build GRCh38
rs766305962 Chr.1: 115651514-115651514 on Build GRCh38
rs747678228 Chr.1: 115651437-115651437 on Build GRCh38
rs372307146 Chr.1: 115651573-115651573 on Build GRCh38
rs78368849 Chr.1: 115651376-115651376 on Build GRCh38
rs953708395 Chr.1: 115651550-115651550 on Build GRCh38
rs955873508 Chr.1: 115651357-115651357 on Build GRCh38
rs894963806 Chr.1: 115651628-115651628 on Build GRCh38
rs763408311 Chr.1: 115651315-115651315 on Build GRCh38
rs767455556 Chr.1: 115651421-115651421 on Build GRCh38
rs374630481 Chr.1: 115651480-115651480 on Build GRCh38
rs746435949 Chr.1: 115651458-115651458 on Build GRCh38
rs756052429 Chr.1: 115651506-115651506 on Build GRCh38

View Full Product Details