Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 39458734-39459228 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIAS
Entrez Gene ID
11019
Gene Name
lipoic acid synthetase
Gene Aliases
HGCLAS, HUSSY-01, LAS, LIP1, LS, PDHLD
UniGene
Hs.550502
Gene Chromosome Location
Chr.4: 39459022-39477653 on Build GRCh38
Transcripts
NM_001278590.1 NM_001278591.1 NM_001278592.1 NM_006859.3 NM_194451.2 XM_006713990.3 XM_017007665.1 XM_017007666.1 XM_017007667.1 NM_000661.4 NM_001024921.3

Gene Symbol
RPL9
Entrez Gene ID
6133
Gene Name
ribosomal protein L9
Gene Aliases
L9, NPC-A-16
UniGene
Hs.412370
Gene Chromosome Location
Chr.4: 39454124-39458948 on Build GRCh38
Transcripts
NM_001278590.1 NM_001278591.1 NM_001278592.1 NM_006859.3 NM_194451.2 XM_006713990.3 XM_017007665.1 XM_017007666.1 XM_017007667.1 NM_000661.4 NM_001024921.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6700968 c.12C>T p.R4R Chr.4: 39459129-39459129 on Build GRCh38
6700966 c.4T>C p.S2P Chr.4: 39459121-39459121 on Build GRCh38
3940871 c.7C>G p.L3V Chr.4: 39459124-39459124 on Build GRCh38
5039609 c.14G>A p.C5Y Chr.4: 39459131-39459131 on Build GRCh38
111733 c.14_19delGCGGGG p.C5_D7>Y Chr.4: 39459131-39459136 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753523124 Chr.4: 39458861-39458870 on Build GRCh38
rs187943580 Chr.4: 39458920-39458920 on Build GRCh38
rs909210627 Chr.4: 39459070-39459070 on Build GRCh38
rs935959267 Chr.4: 39458821-39458821 on Build GRCh38
rs577041802 Chr.4: 39459191-39459191 on Build GRCh38
rs761633225 Chr.4: 39459136-39459136 on Build GRCh38
rs192165699 Chr.4: 39458887-39458887 on Build GRCh38
rs147516123 Chr.4: 39459148-39459148 on Build GRCh38
rs367698915 Chr.4: 39458886-39458886 on Build GRCh38
rs752508692 Chr.4: 39458948-39458948 on Build GRCh38
rs761945224 Chr.4: 39458899-39458899 on Build GRCh38
rs139360916 Chr.4: 39459117-39459117 on Build GRCh38
rs751672446 Chr.4: 39459046-39459046 on Build GRCh38
rs145824152 Chr.4: 39458841-39458841 on Build GRCh38
rs770038370 Chr.4: 39459195-39459195 on Build GRCh38
rs948647185 Chr.4: 39458926-39458926 on Build GRCh38
rs371555159 Chr.4: 39458882-39458882 on Build GRCh38
rs766731883 Chr.4: 39458950-39458951 on Build GRCh38
rs748486869 Chr.4: 39459114-39459114 on Build GRCh38
rs948637163 Chr.4: 39458936-39458936 on Build GRCh38
rs532250115 Chr.4: 39458795-39458795 on Build GRCh38
rs575586178 Chr.4: 39459190-39459190 on Build GRCh38
rs763844132 Chr.4: 39458950-39458950 on Build GRCh38
rs977465197 Chr.4: 39458813-39458813 on Build GRCh38
rs748919573 Chr.4: 39459073-39459073 on Build GRCh38
rs976017247 Chr.4: 39458778-39458778 on Build GRCh38
rs760755406 Chr.4: 39459088-39459088 on Build GRCh38
rs999105881 Chr.4: 39459142-39459142 on Build GRCh38
rs774040503 Chr.4: 39459135-39459135 on Build GRCh38
rs752025916 Chr.4: 39458974-39458975 on Build GRCh38
rs750405548 Chr.4: 39458897-39458897 on Build GRCh38
rs370817832 Chr.4: 39459083-39459083 on Build GRCh38
rs192607174 Chr.4: 39459052-39459052 on Build GRCh38
rs3821989 Chr.4: 39458952-39458952 on Build GRCh38
rs2276889 Chr.4: 39458793-39458793 on Build GRCh38
rs371535593 Chr.4: 39458911-39458911 on Build GRCh38
rs1005864202 Chr.4: 39458893-39458893 on Build GRCh38
rs116433592 Chr.4: 39458919-39458919 on Build GRCh38
rs552022019 Chr.4: 39458834-39458834 on Build GRCh38
rs763714776 Chr.4: 39458866-39458866 on Build GRCh38
rs368281946 Chr.4: 39459089-39459089 on Build GRCh38
rs759743945 Chr.4: 39459092-39459092 on Build GRCh38
rs199924644 Chr.4: 39459100-39459100 on Build GRCh38
rs772474687 Chr.4: 39459115-39459115 on Build GRCh38
rs984797041 Chr.4: 39459066-39459066 on Build GRCh38
rs532232679 Chr.4: 39459005-39459005 on Build GRCh38
rs760399691 Chr.4: 39458921-39458921 on Build GRCh38
rs891318932 Chr.4: 39459004-39459004 on Build GRCh38
rs557550245 Chr.4: 39458888-39458888 on Build GRCh38
rs1008214601 Chr.4: 39458978-39458978 on Build GRCh38
rs779690207 Chr.4: 39459108-39459108 on Build GRCh38
rs553110956 Chr.4: 39459144-39459144 on Build GRCh38
rs774055448 Chr.4: 39458898-39458898 on Build GRCh38
rs1040413348 Chr.4: 39458861-39458861 on Build GRCh38
rs11549995 Chr.4: 39458794-39458794 on Build GRCh38
rs756447054 Chr.4: 39458807-39458807 on Build GRCh38
rs551024345 Chr.4: 39459095-39459095 on Build GRCh38
rs983283954 Chr.4: 39459069-39459069 on Build GRCh38
rs573005019 Chr.4: 39459149-39459149 on Build GRCh38
rs368786581 Chr.4: 39459137-39459137 on Build GRCh38
rs766250791 Chr.4: 39458943-39458943 on Build GRCh38
rs776010886 Chr.4: 39458922-39458922 on Build GRCh38
rs776875932 Chr.4: 39459090-39459090 on Build GRCh38
rs187504573 Chr.4: 39458878-39458878 on Build GRCh38
rs950788561 Chr.4: 39459062-39459062 on Build GRCh38
rs78325971 Chr.4: 39459086-39459086 on Build GRCh38
rs1023394284 Chr.4: 39459034-39459034 on Build GRCh38
rs755281327 Chr.4: 39459193-39459194 on Build GRCh38
rs559528179 Chr.4: 39459043-39459043 on Build GRCh38
rs754671895 Chr.4: 39459112-39459112 on Build GRCh38
rs924615964 Chr.4: 39458819-39458819 on Build GRCh38
rs17553177 Chr.4: 39458943-39458943 on Build GRCh38
rs767605728 Chr.4: 39458905-39458905 on Build GRCh38
rs755745456 Chr.4: 39458968-39458968 on Build GRCh38
rs775825722 Chr.4: 39459201-39459201 on Build GRCh38
rs575423476 Chr.4: 39458838-39458838 on Build GRCh38
rs779423793 Chr.4: 39459113-39459113 on Build GRCh38
rs767549350 Chr.4: 39458945-39458945 on Build GRCh38
rs753034980 Chr.4: 39459159-39459159 on Build GRCh38
rs184218786 Chr.4: 39459161-39459161 on Build GRCh38
rs764276027 Chr.4: 39459017-39459017 on Build GRCh38
rs900128559 Chr.4: 39458990-39458990 on Build GRCh38
rs776691180 Chr.4: 39459118-39459118 on Build GRCh38
rs761674455 Chr.4: 39459099-39459099 on Build GRCh38
rs1012715855 Chr.4: 39458877-39458877 on Build GRCh38
rs948689461 Chr.4: 39459082-39459082 on Build GRCh38
rs1051325640 Chr.4: 39458995-39458995 on Build GRCh38
rs889461254 Chr.4: 39459180-39459180 on Build GRCh38
rs144566412 Chr.4: 39458885-39458885 on Build GRCh38
rs779814468 Chr.4: 39459072-39459072 on Build GRCh38
rs981782668 Chr.4: 39459055-39459055 on Build GRCh38
rs878995907 Chr.4: 39459192-39459192 on Build GRCh38
rs370273736 Chr.4: 39459053-39459053 on Build GRCh38
rs563585827 Chr.4: 39458963-39458963 on Build GRCh38
rs769193742 Chr.4: 39459076-39459076 on Build GRCh38
rs915459167 Chr.4: 39458791-39458791 on Build GRCh38
rs183248215 Chr.4: 39458894-39458894 on Build GRCh38
rs778655996 Chr.4: 39459156-39459156 on Build GRCh38
rs376007055 Chr.4: 39458867-39458867 on Build GRCh38
rs2276890 Chr.4: 39458870-39458870 on Build GRCh38
rs771992768 Chr.4: 39458865-39458865 on Build GRCh38
rs779309308 Chr.4: 39458946-39458946 on Build GRCh38
rs771363108 Chr.4: 39459174-39459174 on Build GRCh38
rs540003300 Chr.4: 39458904-39458904 on Build GRCh38
rs2687959 Chr.4: 39459177-39459177 on Build GRCh38
rs941783632 Chr.4: 39458975-39458975 on Build GRCh38
rs909158364 Chr.4: 39458929-39458929 on Build GRCh38
rs772165767 Chr.4: 39458915-39458915 on Build GRCh38
rs747620289 Chr.4: 39459172-39459172 on Build GRCh38
rs747611391 Chr.4: 39458913-39458913 on Build GRCh38
rs775039588 Chr.4: 39458932-39458932 on Build GRCh38
rs753600472 Chr.4: 39459109-39459109 on Build GRCh38
rs997516565 Chr.4: 39458991-39458991 on Build GRCh38
rs947116408 Chr.4: 39459151-39459151 on Build GRCh38
rs750658575 Chr.4: 39458906-39458906 on Build GRCh38
rs371534347 Chr.4: 39458847-39458847 on Build GRCh38
rs561700411 Chr.4: 39458923-39458923 on Build GRCh38
rs767927654 Chr.4: 39459060-39459060 on Build GRCh38
rs772875462 Chr.4: 39459204-39459204 on Build GRCh38
rs780576068 Chr.4: 39458954-39458954 on Build GRCh38
rs552344295 Chr.4: 39459021-39459021 on Build GRCh38
rs988402230 Chr.4: 39458755-39458755 on Build GRCh38
rs1030248171 Chr.4: 39458903-39458903 on Build GRCh38
rs757217335 Chr.4: 39458953-39458953 on Build GRCh38
rs574414526 Chr.4: 39458947-39458947 on Build GRCh38
rs749933078 Chr.4: 39459101-39459101 on Build GRCh38
rs143267758 Chr.4: 39459145-39459145 on Build GRCh38
rs757009355 Chr.4: 39458873-39458873 on Build GRCh38
rs750426614 Chr.4: 39459141-39459141 on Build GRCh38
rs372288500 Chr.4: 39459122-39459122 on Build GRCh38
rs548211367 Chr.4: 39459056-39459056 on Build GRCh38
rs763200777 Chr.4: 39458959-39458959 on Build GRCh38
rs1017819678 Chr.4: 39458979-39458979 on Build GRCh38
rs906991221 Chr.4: 39458884-39458884 on Build GRCh38
rs769950134 Chr.4: 39459133-39459133 on Build GRCh38
rs769320032 Chr.4: 39458925-39458925 on Build GRCh38
rs755691859 Chr.4: 39459102-39459102 on Build GRCh38
rs1052946160 Chr.4: 39459147-39459147 on Build GRCh38
rs763067097 Chr.4: 39459203-39459203 on Build GRCh38

View Full Product Details