Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70768460-70768887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RUFY3
Entrez Gene ID
22902
Gene Name
RUN and FYVE domain containing 3
Gene Aliases
RIPX, SINGAR1, ZFYVE30
UniGene
Hs.740904
Gene Chromosome Location
Chr.4: 70704204-70808622 on Build GRCh38
Transcripts
NM_001037442.3 NM_001130709.1 NM_001291993.1 NM_001291994.1 NM_014961.4 XM_005265656.2 XM_005265658.1 XM_011531750.2 XM_011531751.2 XM_011531752.2 XM_011531753.2 XM_011531754.2 XM_011531755.2 XM_011531756.2 XM_017007890.1 XM_017007891.1 XM_017007892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4411820 c.773C>T p.A258V Chr.4: 70768558-70768558 on Build GRCh38
7308205 c.792A>G p.E264E Chr.4: 70768577-70768577 on Build GRCh38
5553081 c.681A>G p.E227E Chr.4: 70768646-70768646 on Build GRCh38
7121409 c.679G>C p.E227Q Chr.4: 70768644-70768644 on Build GRCh38
4411822 c.545C>T p.A182V Chr.4: 70768558-70768558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748783645 Chr.4: 70768577-70768577 on Build GRCh38
rs919709394 Chr.4: 70768695-70768695 on Build GRCh38
rs369491730 Chr.4: 70768516-70768516 on Build GRCh38
rs200374107 Chr.4: 70768704-70768704 on Build GRCh38
rs368488073 Chr.4: 70768622-70768622 on Build GRCh38
rs768204417 Chr.4: 70768580-70768580 on Build GRCh38
rs1038043240 Chr.4: 70768811-70768811 on Build GRCh38
rs199654265 Chr.4: 70768555-70768555 on Build GRCh38
rs912242333 Chr.4: 70768677-70768677 on Build GRCh38
rs1057345124 Chr.4: 70768832-70768832 on Build GRCh38
rs776561491 Chr.4: 70768689-70768689 on Build GRCh38
rs896826260 Chr.4: 70768833-70768833 on Build GRCh38
rs374725713 Chr.4: 70768560-70768560 on Build GRCh38
rs372261834 Chr.4: 70768628-70768628 on Build GRCh38
rs1003089560 Chr.4: 70768509-70768509 on Build GRCh38
rs142016934 Chr.4: 70768507-70768507 on Build GRCh38
rs771848845 Chr.4: 70768635-70768635 on Build GRCh38
rs1003093690 Chr.4: 70768810-70768810 on Build GRCh38
rs778789786 Chr.4: 70768606-70768606 on Build GRCh38
rs1044964838 Chr.4: 70768729-70768729 on Build GRCh38
rs1010180116 Chr.4: 70768530-70768530 on Build GRCh38
rs959424956 Chr.4: 70768529-70768529 on Build GRCh38
rs911001742 Chr.4: 70768726-70768726 on Build GRCh38
rs753724137 Chr.4: 70768850-70768850 on Build GRCh38
rs375551015 Chr.4: 70768691-70768691 on Build GRCh38
rs201458860 Chr.4: 70768672-70768672 on Build GRCh38
rs1035673354 Chr.4: 70768514-70768514 on Build GRCh38
rs371685617 Chr.4: 70768643-70768643 on Build GRCh38
rs569418744 Chr.4: 70768845-70768845 on Build GRCh38
rs763189392 Chr.4: 70768533-70768533 on Build GRCh38
rs73824891 Chr.4: 70768860-70768860 on Build GRCh38
rs770134125 Chr.4: 70768682-70768682 on Build GRCh38
rs763970049 Chr.4: 70768782-70768782 on Build GRCh38
rs571709411 Chr.4: 70768631-70768631 on Build GRCh38
rs979710113 Chr.4: 70768550-70768550 on Build GRCh38
rs200931412 Chr.4: 70768607-70768607 on Build GRCh38
rs986690732 Chr.4: 70768595-70768595 on Build GRCh38
rs752157688 Chr.4: 70768548-70768548 on Build GRCh38
rs764681198 Chr.4: 70768547-70768547 on Build GRCh38
rs1033781491 Chr.4: 70768525-70768525 on Build GRCh38
rs772895482 Chr.4: 70768644-70768644 on Build GRCh38
rs182772740 Chr.4: 70768566-70768566 on Build GRCh38
rs201677724 Chr.4: 70768568-70768568 on Build GRCh38
rs764305064 Chr.4: 70768705-70768705 on Build GRCh38
rs145431698 Chr.4: 70768559-70768559 on Build GRCh38
rs1021690891 Chr.4: 70768535-70768535 on Build GRCh38
rs149653894 Chr.4: 70768549-70768549 on Build GRCh38
rs146339077 Chr.4: 70768803-70768803 on Build GRCh38
rs866920541 Chr.4: 70768836-70768836 on Build GRCh38

View Full Product Details