Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 119478564-119479080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B4
Entrez Gene ID
3295
Gene Name
hydroxysteroid 17-beta dehydrogenase 4
Gene Aliases
DBP, MFE-2, MPF-2, PRLTS1, SDR8C1
UniGene
Hs.406861
Gene Chromosome Location
Chr.5: 119452443-119542335 on Build GRCh38
Transcripts
NM_000414.3 NM_001199291.2 NM_001199292.1 NM_001292027.1 NM_001292028.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6322724 c.541G>T p.E181* Chr.5: 119478940-119478940 on Build GRCh38
3134572 c.532C>T p.L178F Chr.5: 119478931-119478931 on Build GRCh38
6101313 c.549G>A p.R183R Chr.5: 119478948-119478948 on Build GRCh38
1310463 c.514C>G p.L172V Chr.5: 119478913-119478913 on Build GRCh38
4840068 c.527A>C p.N176T Chr.5: 119478926-119478926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750582359 Chr.5: 119478984-119478984 on Build GRCh38
rs770301555 Chr.5: 119478918-119478918 on Build GRCh38
rs748937758 Chr.5: 119478987-119478987 on Build GRCh38
rs1038445943 Chr.5: 119478914-119478914 on Build GRCh38
rs1018258580 Chr.5: 119478782-119478782 on Build GRCh38
rs956947896 Chr.5: 119478950-119478950 on Build GRCh38
rs767080082 Chr.5: 119478817-119478817 on Build GRCh38
rs950293554 Chr.5: 119478764-119478764 on Build GRCh38
rs779756356 Chr.5: 119478855-119478855 on Build GRCh38
rs550705310 Chr.5: 119478986-119478986 on Build GRCh38
rs770261826 Chr.5: 119478996-119478996 on Build GRCh38
rs753646023 Chr.5: 119478973-119478973 on Build GRCh38
rs199755823 Chr.5: 119478955-119478955 on Build GRCh38
rs1045053143 Chr.5: 119478779-119478779 on Build GRCh38
rs775766910 Chr.5: 119478925-119478925 on Build GRCh38
rs762551539 Chr.5: 119478788-119478788 on Build GRCh38
rs756080976 Chr.5: 119478827-119478827 on Build GRCh38
rs925430446 Chr.5: 119478600-119478600 on Build GRCh38
rs778048565 Chr.5: 119478879-119478879 on Build GRCh38
rs371272788 Chr.5: 119478804-119478804 on Build GRCh38
rs1038246380 Chr.5: 119479039-119479039 on Build GRCh38
rs370080881 Chr.5: 119478819-119478819 on Build GRCh38
rs536841970 Chr.5: 119478679-119478679 on Build GRCh38
rs536487449 Chr.5: 119479026-119479026 on Build GRCh38
rs767169728 Chr.5: 119478961-119478961 on Build GRCh38
rs776060692 Chr.5: 119478999-119478999 on Build GRCh38
rs954683559 Chr.5: 119478590-119478590 on Build GRCh38
rs754803105 Chr.5: 119478977-119478977 on Build GRCh38
rs753064337 Chr.5: 119478787-119478787 on Build GRCh38
rs749090911 Chr.5: 119478858-119478858 on Build GRCh38
rs1045376697 Chr.5: 119478703-119478703 on Build GRCh38
rs763363391 Chr.5: 119478935-119478935 on Build GRCh38
rs776640310 Chr.5: 119478900-119478900 on Build GRCh38
rs758061281 Chr.5: 119478831-119478832 on Build GRCh38
rs867848133 Chr.5: 119478685-119478685 on Build GRCh38
rs758074832 Chr.5: 119478698-119478698 on Build GRCh38
rs764299947 Chr.5: 119478949-119478949 on Build GRCh38
rs761601392 Chr.5: 119479013-119479013 on Build GRCh38
rs777556109 Chr.5: 119479023-119479042 on Build GRCh38
rs751088170 Chr.5: 119478805-119478805 on Build GRCh38
rs781288160 Chr.5: 119478994-119478994 on Build GRCh38
rs192036834 Chr.5: 119478719-119478719 on Build GRCh38
rs765174171 Chr.5: 119478841-119478841 on Build GRCh38
rs772919216 Chr.5: 119479025-119479025 on Build GRCh38
rs183972735 Chr.5: 119478708-119478708 on Build GRCh38
rs934810724 Chr.5: 119478729-119478729 on Build GRCh38
rs897274090 Chr.5: 119478976-119478976 on Build GRCh38
rs987539899 Chr.5: 119478650-119478650 on Build GRCh38
rs142141053 Chr.5: 119478995-119478995 on Build GRCh38
rs935520686 Chr.5: 119478625-119478625 on Build GRCh38
rs750409693 Chr.5: 119478821-119478821 on Build GRCh38
rs752455947 Chr.5: 119479040-119479040 on Build GRCh38
rs201280286 Chr.5: 119479036-119479036 on Build GRCh38
rs559620592 Chr.5: 119478730-119478730 on Build GRCh38
rs746404368 Chr.5: 119478906-119478906 on Build GRCh38
rs955449465 Chr.5: 119478760-119478760 on Build GRCh38
rs545729272 Chr.5: 119478709-119478709 on Build GRCh38
rs771009588 Chr.5: 119478881-119478881 on Build GRCh38
rs766330442 Chr.5: 119478969-119478969 on Build GRCh38
rs774818626 Chr.5: 119479007-119479007 on Build GRCh38
rs774066654 Chr.5: 119478954-119478954 on Build GRCh38
rs759362780 Chr.5: 119478725-119478725 on Build GRCh38
rs1057516310 Chr.5: 119479006-119479009 on Build GRCh38
rs949850525 Chr.5: 119478691-119478691 on Build GRCh38
rs765736425 Chr.5: 119479035-119479035 on Build GRCh38
rs765610180 Chr.5: 119478784-119478784 on Build GRCh38
rs372914814 Chr.5: 119478965-119478965 on Build GRCh38
rs749700774 Chr.5: 119479003-119479003 on Build GRCh38
rs193193124 Chr.5: 119478707-119478707 on Build GRCh38
rs541880475 Chr.5: 119478758-119478758 on Build GRCh38
rs1008741520 Chr.5: 119478780-119478780 on Build GRCh38
rs376738933 Chr.5: 119479037-119479037 on Build GRCh38
rs374797053 Chr.5: 119478808-119478808 on Build GRCh38
rs1053278816 Chr.5: 119478660-119478660 on Build GRCh38
rs767259858 Chr.5: 119479024-119479024 on Build GRCh38
rs910180381 Chr.5: 119478653-119478653 on Build GRCh38
rs75814129 Chr.5: 119478732-119478732 on Build GRCh38
rs747162503 Chr.5: 119478880-119478880 on Build GRCh38

View Full Product Details