Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 159316593-159317086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12B
Entrez Gene ID
3593
Gene Name
interleukin 12B
Gene Aliases
CLMF, CLMF2, IL-12B, IMD28, IMD29, NKSF, NKSF2
UniGene
Hs.674
Gene Chromosome Location
Chr.5: 159314783-159330473 on Build GRCh38
Transcripts
NM_002187.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5975489 c.856-1G>C p.? Chr.5: 159316817-159316817 on Build GRCh38
5082424 c.856-4G>T p.? Chr.5: 159316820-159316820 on Build GRCh38
5081186 c.856-3C>A p.? Chr.5: 159316819-159316819 on Build GRCh38
1642666 c.864A>G p.R288R Chr.5: 159316808-159316808 on Build GRCh38
2152033 c.925C>T p.R309W Chr.5: 159316747-159316747 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764288572 Chr.5: 159316848-159316848 on Build GRCh38
rs750452400 Chr.5: 159316774-159316774 on Build GRCh38
rs762969963 Chr.5: 159316826-159316826 on Build GRCh38
rs201161608 Chr.5: 159316676-159316676 on Build GRCh38
rs757678113 Chr.5: 159316791-159316791 on Build GRCh38
rs572087899 Chr.5: 159316888-159316888 on Build GRCh38
rs1053434834 Chr.5: 159316675-159316675 on Build GRCh38
rs150902695 Chr.5: 159316775-159316775 on Build GRCh38
rs377511395 Chr.5: 159316770-159316770 on Build GRCh38
rs768523225 Chr.5: 159316751-159316751 on Build GRCh38
rs913646189 Chr.5: 159316905-159316905 on Build GRCh38
rs370904274 Chr.5: 159316809-159316809 on Build GRCh38
rs866583843 Chr.5: 159317047-159317047 on Build GRCh38
rs376860098 Chr.5: 159316858-159316858 on Build GRCh38
rs371280399 Chr.5: 159316735-159316735 on Build GRCh38
rs754181801 Chr.5: 159316790-159316790 on Build GRCh38
rs778129509 Chr.5: 159316728-159316728 on Build GRCh38
rs760598277 Chr.5: 159316647-159316648 on Build GRCh38
rs760718653 Chr.5: 159316847-159316847 on Build GRCh38
rs898968596 Chr.5: 159316760-159316760 on Build GRCh38
rs890103497 Chr.5: 159317040-159317040 on Build GRCh38
rs1045624249 Chr.5: 159316619-159316619 on Build GRCh38
rs934330712 Chr.5: 159316707-159316707 on Build GRCh38
rs186003445 Chr.5: 159316781-159316781 on Build GRCh38
rs779642242 Chr.5: 159316806-159316806 on Build GRCh38
rs571398445 Chr.5: 159316739-159316739 on Build GRCh38
rs890924951 Chr.5: 159316953-159316953 on Build GRCh38
rs943880210 Chr.5: 159316959-159316959 on Build GRCh38
rs776936485 Chr.5: 159316736-159316736 on Build GRCh38
rs56404680 Chr.5: 159316682-159316682 on Build GRCh38
rs150030340 Chr.5: 159316798-159316798 on Build GRCh38
rs563410998 Chr.5: 159316659-159316659 on Build GRCh38
rs138991749 Chr.5: 159316691-159316691 on Build GRCh38
rs978311869 Chr.5: 159316857-159316857 on Build GRCh38
rs879408394 Chr.5: 159316710-159316710 on Build GRCh38
rs368783048 Chr.5: 159316649-159316649 on Build GRCh38
rs144054146 Chr.5: 159316808-159316808 on Build GRCh38
rs11574790 Chr.5: 159316838-159316838 on Build GRCh38
rs763955123 Chr.5: 159316664-159316664 on Build GRCh38
rs374661285 Chr.5: 159316782-159316782 on Build GRCh38
rs760387348 Chr.5: 159316663-159316663 on Build GRCh38
rs3213119 Chr.5: 159316780-159316780 on Build GRCh38
rs936802322 Chr.5: 159316876-159316876 on Build GRCh38
rs377524637 Chr.5: 159316734-159316734 on Build GRCh38
rs776601045 Chr.5: 159316758-159316758 on Build GRCh38
rs759845005 Chr.5: 159316844-159316844 on Build GRCh38
rs11574791 Chr.5: 159316662-159316662 on Build GRCh38
rs143277765 Chr.5: 159316690-159316690 on Build GRCh38
rs191112662 Chr.5: 159316874-159316874 on Build GRCh38
rs139163137 Chr.5: 159316802-159316802 on Build GRCh38
rs752964137 Chr.5: 159316712-159316712 on Build GRCh38
rs776481806 Chr.5: 159316810-159316810 on Build GRCh38
rs771683283 Chr.5: 159316747-159316747 on Build GRCh38
rs772964813 Chr.5: 159316818-159316818 on Build GRCh38
rs201512006 Chr.5: 159316711-159316711 on Build GRCh38
rs914269376 Chr.5: 159316892-159316892 on Build GRCh38
rs926770092 Chr.5: 159316638-159316638 on Build GRCh38
rs761865432 Chr.5: 159316685-159316685 on Build GRCh38
rs879167745 Chr.5: 159316960-159316960 on Build GRCh38
rs146408881 Chr.5: 159316812-159316812 on Build GRCh38
rs11574789 Chr.5: 159316885-159316885 on Build GRCh38
rs775607044 Chr.5: 159316843-159316843 on Build GRCh38
rs757551412 Chr.5: 159316865-159316865 on Build GRCh38
rs1049510351 Chr.5: 159316929-159316929 on Build GRCh38
rs776548694 Chr.5: 159316821-159316821 on Build GRCh38
rs183259271 Chr.5: 159316920-159316920 on Build GRCh38
rs756372086 Chr.5: 159316724-159316724 on Build GRCh38
rs759089990 Chr.5: 159316647-159316647 on Build GRCh38
rs56064925 Chr.5: 159316699-159316699 on Build GRCh38
rs780789325 Chr.5: 159316771-159316771 on Build GRCh38
rs371057558 Chr.5: 159317022-159317022 on Build GRCh38
rs374523632 Chr.5: 159316784-159316784 on Build GRCh38
rs989229217 Chr.5: 159316887-159316887 on Build GRCh38
rs754126843 Chr.5: 159316854-159316854 on Build GRCh38
rs769713143 Chr.5: 159316817-159316817 on Build GRCh38
rs144356274 Chr.5: 159316799-159316799 on Build GRCh38
rs773879958 Chr.5: 159316640-159316640 on Build GRCh38
rs373042362 Chr.5: 159316750-159316750 on Build GRCh38
rs149957653 Chr.5: 159317039-159317039 on Build GRCh38
rs757104946 Chr.5: 159316674-159316675 on Build GRCh38
rs367828574 Chr.5: 159316800-159316800 on Build GRCh38

View Full Product Details