Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 168385119-168385488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WWC1
Entrez Gene ID
23286
Gene Name
WW and C2 domain containing 1
Gene Aliases
HBEBP3, HBEBP36, KIBRA, MEMRYQTL, PPP1R168
UniGene
Hs.484047
Gene Chromosome Location
Chr.5: 168292060-168472303 on Build GRCh38
Transcripts
NM_001161661.1 NM_001161662.1 NM_015238.2 XM_005265850.1 XM_005265853.2 XM_011534485.1 XM_011534486.1 XM_011534487.1 XM_011534488.1 XM_011534489.1 XM_011534490.1 XM_011534491.1 XM_017009276.1 XM_017009277.1 XM_017009278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3338493 c.253C>T p.R85* Chr.5: 168385234-168385234 on Build GRCh38
5356711 c.241A>T p.I81F Chr.5: 168385222-168385222 on Build GRCh38
4541011 c.292G>A p.D98N Chr.5: 168385273-168385273 on Build GRCh38
4788911 c.400G>T p.V134F Chr.5: 168385381-168385381 on Build GRCh38
4775725 c.400G>A p.V134I Chr.5: 168385381-168385381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750622325 Chr.5: 168385199-168385199 on Build GRCh38
rs959265913 Chr.5: 168385356-168385356 on Build GRCh38
rs371306643 Chr.5: 168385408-168385408 on Build GRCh38
rs141065175 Chr.5: 168385329-168385329 on Build GRCh38
rs576183356 Chr.5: 168385278-168385278 on Build GRCh38
rs756712761 Chr.5: 168385453-168385453 on Build GRCh38
rs564853802 Chr.5: 168385381-168385381 on Build GRCh38
rs191310466 Chr.5: 168385153-168385153 on Build GRCh38
rs748965666 Chr.5: 168385269-168385269 on Build GRCh38
rs1048387296 Chr.5: 168385143-168385143 on Build GRCh38
rs753417043 Chr.5: 168385445-168385445 on Build GRCh38
rs144891291 Chr.5: 168385347-168385347 on Build GRCh38
rs746904400 Chr.5: 168385376-168385376 on Build GRCh38
rs545162323 Chr.5: 168385247-168385247 on Build GRCh38
rs774422436 Chr.5: 168385399-168385399 on Build GRCh38
rs373994122 Chr.5: 168385169-168385169 on Build GRCh38
rs144904124 Chr.5: 168385267-168385267 on Build GRCh38
rs140349506 Chr.5: 168385380-168385380 on Build GRCh38
rs200222307 Chr.5: 168385318-168385318 on Build GRCh38
rs781547786 Chr.5: 168385246-168385246 on Build GRCh38
rs558259236 Chr.5: 168385325-168385325 on Build GRCh38
rs752028192 Chr.5: 168385316-168385316 on Build GRCh38
rs540923375 Chr.5: 168385345-168385345 on Build GRCh38
rs750677394 Chr.5: 168385307-168385307 on Build GRCh38
rs761949336 Chr.5: 168385167-168385167 on Build GRCh38
rs375573316 Chr.5: 168385339-168385339 on Build GRCh38
rs753230603 Chr.5: 168385438-168385438 on Build GRCh38
rs780398674 Chr.5: 168385235-168385235 on Build GRCh38
rs770086119 Chr.5: 168385252-168385252 on Build GRCh38
rs756427059 Chr.5: 168385249-168385249 on Build GRCh38
rs763093926 Chr.5: 168385395-168385395 on Build GRCh38
rs771709179 Chr.5: 168385350-168385350 on Build GRCh38
rs777428232 Chr.5: 168385254-168385254 on Build GRCh38
rs140660160 Chr.5: 168385290-168385290 on Build GRCh38
rs778643617 Chr.5: 168385454-168385454 on Build GRCh38
rs990160836 Chr.5: 168385279-168385279 on Build GRCh38
rs764558324 Chr.5: 168385442-168385442 on Build GRCh38
rs758246001 Chr.5: 168385463-168385463 on Build GRCh38
rs992045157 Chr.5: 168385446-168385446 on Build GRCh38
rs775264335 Chr.5: 168385292-168385292 on Build GRCh38
rs777201013 Chr.5: 168385333-168385333 on Build GRCh38
rs1048819674 Chr.5: 168385460-168385460 on Build GRCh38
rs955187984 Chr.5: 168385239-168385239 on Build GRCh38
rs374384593 Chr.5: 168385431-168385431 on Build GRCh38
rs770661252 Chr.5: 168385340-168385340 on Build GRCh38
rs766690072 Chr.5: 168385398-168385398 on Build GRCh38
rs758863684 Chr.5: 168385215-168385215 on Build GRCh38
rs763209673 Chr.5: 168385308-168385308 on Build GRCh38
rs145834613 Chr.5: 168385291-168385291 on Build GRCh38
rs368286797 Chr.5: 168385456-168385456 on Build GRCh38
rs759957091 Chr.5: 168385417-168385417 on Build GRCh38
rs888430617 Chr.5: 168385466-168385466 on Build GRCh38
rs557621965 Chr.5: 168385250-168385250 on Build GRCh38
rs781748133 Chr.5: 168385319-168385319 on Build GRCh38
rs149869064 Chr.5: 168385242-168385242 on Build GRCh38
rs371599955 Chr.5: 168385258-168385258 on Build GRCh38
rs368164798 Chr.5: 168385238-168385238 on Build GRCh38
rs775504769 Chr.5: 168385364-168385364 on Build GRCh38
rs745326492 Chr.5: 168385396-168385396 on Build GRCh38
rs754012766 Chr.5: 168385165-168385165 on Build GRCh38
rs756560682 Chr.5: 168385441-168385441 on Build GRCh38
rs762004252 Chr.5: 168385305-168385305 on Build GRCh38
rs1004272041 Chr.5: 168385166-168385166 on Build GRCh38

View Full Product Details