Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 179617658-179618163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPH1
Entrez Gene ID
3187
Gene Name
heterogeneous nuclear ribonucleoprotein H1 (H)
Gene Aliases
HNRPH, HNRPH1, hnRNPH
UniGene
Hs.604001
Gene Chromosome Location
Chr.5: 179614178-179634784 on Build GRCh38
Transcripts
NM_001257293.1 NM_005520.2 XM_005265895.2 XM_005265896.2 XM_005265901.1 XM_005265902.4 XM_006714862.2 XM_006714863.2 XM_011534542.1 XM_011534544.1 XM_011534545.1 XM_011534546.1 XM_011534547.1 XM_017009414.1 XM_017009415.1 XM_017009416.1 XM_017009417.1 XM_017009418.1 XM_017009419.1 XM_017009420.1 XM_017009421.1 XM_017009422.1 XM_017009423.1 XM_017009424.1 XM_017009425.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6103684 c.706T>A p.Y236N Chr.5: 179618154-179618154 on Build GRCh38
6681307 c.62T>C p.V21A Chr.5: 179617848-179617848 on Build GRCh38
7169980 c.48A>T p.T16T Chr.5: 179617862-179617862 on Build GRCh38
7138861 c.94G>A p.E32K Chr.5: 179617816-179617816 on Build GRCh38
3011557 c.805delT p.S269fs*47 Chr.5: 179617915-179617915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772688424 Chr.5: 179618093-179618093 on Build GRCh38
rs374187855 Chr.5: 179618131-179618131 on Build GRCh38
rs1050716153 Chr.5: 179617706-179617706 on Build GRCh38
rs762182057 Chr.5: 179618119-179618119 on Build GRCh38
rs114713826 Chr.5: 179617886-179617886 on Build GRCh38
rs1042621530 Chr.5: 179617697-179617697 on Build GRCh38
rs750355470 Chr.5: 179617693-179617693 on Build GRCh38
rs758687811 Chr.5: 179618026-179618026 on Build GRCh38
rs770037548 Chr.5: 179618069-179618070 on Build GRCh38
rs539627779 Chr.5: 179618097-179618097 on Build GRCh38
rs761418813 Chr.5: 179618068-179618069 on Build GRCh38
rs371144759 Chr.5: 179618067-179618067 on Build GRCh38
rs115489078 Chr.5: 179617999-179617999 on Build GRCh38
rs766063549 Chr.5: 179617741-179617741 on Build GRCh38
rs200553004 Chr.5: 179618076-179618076 on Build GRCh38
rs778196414 Chr.5: 179618114-179618114 on Build GRCh38
rs767529419 Chr.5: 179617931-179617931 on Build GRCh38
rs1010813561 Chr.5: 179617909-179617909 on Build GRCh38
rs779319718 Chr.5: 179618090-179618090 on Build GRCh38
rs879046509 Chr.5: 179617937-179617937 on Build GRCh38
rs768945177 Chr.5: 179618047-179618047 on Build GRCh38
rs199617940 Chr.5: 179617823-179617823 on Build GRCh38
rs566057819 Chr.5: 179618079-179618079 on Build GRCh38
rs755878408 Chr.5: 179617750-179617750 on Build GRCh38
rs749580517 Chr.5: 179617885-179617885 on Build GRCh38
rs140414006 Chr.5: 179618048-179618048 on Build GRCh38
rs754699121 Chr.5: 179617983-179617983 on Build GRCh38
rs747088298 Chr.5: 179618026-179618028 on Build GRCh38
rs753716331 Chr.5: 179617946-179617946 on Build GRCh38
rs373146708 Chr.5: 179617844-179617844 on Build GRCh38
rs370275246 Chr.5: 179618122-179618122 on Build GRCh38
rs775779304 Chr.5: 179618076-179618079 on Build GRCh38
rs112827407 Chr.5: 179617727-179617727 on Build GRCh38
rs900488696 Chr.5: 179617773-179617773 on Build GRCh38
rs942148816 Chr.5: 179617749-179617749 on Build GRCh38
rs878941703 Chr.5: 179617747-179617747 on Build GRCh38
rs370622280 Chr.5: 179617778-179617778 on Build GRCh38
rs191656062 Chr.5: 179618038-179618038 on Build GRCh38
rs889536500 Chr.5: 179617722-179617722 on Build GRCh38
rs879119150 Chr.5: 179617936-179617936 on Build GRCh38
rs776048911 Chr.5: 179617950-179617951 on Build GRCh38
rs907105051 Chr.5: 179617845-179617845 on Build GRCh38
rs771242480 Chr.5: 179618020-179618020 on Build GRCh38
rs767746065 Chr.5: 179617692-179617692 on Build GRCh38
rs371107241 Chr.5: 179617949-179617949 on Build GRCh38
rs747949329 Chr.5: 179617949-179617950 on Build GRCh38
rs373907497 Chr.5: 179618118-179618118 on Build GRCh38
rs746612505 Chr.5: 179617780-179617780 on Build GRCh38
rs764195917 Chr.5: 179617943-179617943 on Build GRCh38
rs761652664 Chr.5: 179617948-179617948 on Build GRCh38
rs757304534 Chr.5: 179617764-179617764 on Build GRCh38
rs754831641 Chr.5: 179617698-179617698 on Build GRCh38
rs747144550 Chr.5: 179618105-179618105 on Build GRCh38
rs765065517 Chr.5: 179618056-179618056 on Build GRCh38
rs201396285 Chr.5: 179617807-179617807 on Build GRCh38
rs367788432 Chr.5: 179618113-179618113 on Build GRCh38
rs769109224 Chr.5: 179617945-179617945 on Build GRCh38
rs753575343 Chr.5: 179617763-179617763 on Build GRCh38
rs771086884 Chr.5: 179618064-179618065 on Build GRCh38
rs757993508 Chr.5: 179617965-179617965 on Build GRCh38
rs778609998 Chr.5: 179617790-179617791 on Build GRCh38
rs779671967 Chr.5: 179617976-179617976 on Build GRCh38
rs750321620 Chr.5: 179617954-179617954 on Build GRCh38
rs761028285 Chr.5: 179617942-179617942 on Build GRCh38
rs572692674 Chr.5: 179618138-179618138 on Build GRCh38
rs761707756 Chr.5: 179618050-179618050 on Build GRCh38
rs373143741 Chr.5: 179617723-179617733 on Build GRCh38
rs759641757 Chr.5: 179617898-179617898 on Build GRCh38
rs770134368 Chr.5: 179618121-179618121 on Build GRCh38
rs374506740 Chr.5: 179618078-179618078 on Build GRCh38
rs4701143 Chr.5: 179618141-179618141 on Build GRCh38
rs200609447 Chr.5: 179617980-179617980 on Build GRCh38
rs567749303 Chr.5: 179618081-179618081 on Build GRCh38
rs777846035 Chr.5: 179617979-179617983 on Build GRCh38
rs538196122 Chr.5: 179618117-179618117 on Build GRCh38
rs757942493 Chr.5: 179617776-179617776 on Build GRCh38
rs755752852 Chr.5: 179618088-179618088 on Build GRCh38
rs774786323 Chr.5: 179618097-179618100 on Build GRCh38
rs766196278 Chr.5: 179618074-179618074 on Build GRCh38
rs188998161 Chr.5: 179617771-179617771 on Build GRCh38
rs775421749 Chr.5: 179617935-179617935 on Build GRCh38
rs559553978 Chr.5: 179617757-179617757 on Build GRCh38
rs779394978 Chr.5: 179618021-179618021 on Build GRCh38
rs1038110519 Chr.5: 179618102-179618102 on Build GRCh38
rs753437288 Chr.5: 179617697-179617698 on Build GRCh38
rs78795662 Chr.5: 179617892-179617892 on Build GRCh38
rs201483427 Chr.5: 179617852-179617852 on Build GRCh38
rs370278680 Chr.5: 179618106-179618106 on Build GRCh38
rs538803901 Chr.5: 179618105-179618105 on Build GRCh38
rs929093503 Chr.5: 179617705-179617705 on Build GRCh38
rs750228002 Chr.5: 179618069-179618069 on Build GRCh38
rs777026245 Chr.5: 179618068-179618074 on Build GRCh38
rs766036226 Chr.5: 179617698-179617703 on Build GRCh38
rs933350031 Chr.5: 179617880-179617880 on Build GRCh38
rs763050290 Chr.5: 179618073-179618073 on Build GRCh38
rs946971913 Chr.5: 179617684-179617687 on Build GRCh38
rs749682820 Chr.5: 179618011-179618011 on Build GRCh38
rs529513341 Chr.5: 179618029-179618029 on Build GRCh38
rs991842779 Chr.5: 179618052-179618052 on Build GRCh38
rs764593164 Chr.5: 179618089-179618089 on Build GRCh38
rs1003583023 Chr.5: 179617862-179617862 on Build GRCh38
rs917847872 Chr.5: 179617698-179617698 on Build GRCh38
rs759586603 Chr.5: 179617687-179617687 on Build GRCh38
rs763373878 Chr.5: 179618081-179618083 on Build GRCh38
rs778973080 Chr.5: 179617768-179617768 on Build GRCh38

View Full Product Details