Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151292530-151293035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PI4KB
Entrez Gene ID
5298
Gene Name
phosphatidylinositol 4-kinase beta
Gene Aliases
NPIK, PI4K-BETA, PI4K92, PI4KBETA, PI4KIIIBETA, PIK4CB
UniGene
Hs.632465
Gene Chromosome Location
Chr.1: 151291797-151327715 on Build GRCh38
Transcripts
NM_001198773.1 NM_001198774.1 NM_001198775.1 NM_002651.2 XM_005245264.3 XM_011509634.1 XM_011509635.1 XM_011509636.2 XM_011509637.2 XM_011509638.1 XM_017001486.1 XM_017001487.1 XM_017001488.1 XM_017001489.1 XM_017001490.1 XM_017001491.1 XM_017001492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6745830 c.2429G>A p.R810Q Chr.1: 151292910-151292910 on Build GRCh38
7117498 c.2356G>C p.E786Q Chr.1: 151292983-151292983 on Build GRCh38
895763 c.2481C>A p.I827I Chr.1: 151292858-151292858 on Build GRCh38
4480872 c.2461C>T p.Q821* Chr.1: 151292878-151292878 on Build GRCh38
1688519 c.2432C>T p.S811F Chr.1: 151292907-151292907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1043671425 Chr.1: 151292699-151292699 on Build GRCh38
rs759964689 Chr.1: 151292815-151292815 on Build GRCh38
rs587734969 Chr.1: 151292870-151292870 on Build GRCh38
rs141241084 Chr.1: 151292987-151292987 on Build GRCh38
rs1054346764 Chr.1: 151292727-151292727 on Build GRCh38
rs954042393 Chr.1: 151292649-151292651 on Build GRCh38
rs765415391 Chr.1: 151292864-151292864 on Build GRCh38
rs587673929 Chr.1: 151292702-151292702 on Build GRCh38
rs747764332 Chr.1: 151292645-151292645 on Build GRCh38
rs755533251 Chr.1: 151292956-151292956 on Build GRCh38
rs892824980 Chr.1: 151292728-151292728 on Build GRCh38
rs200780369 Chr.1: 151292848-151292848 on Build GRCh38
rs944573813 Chr.1: 151292684-151292684 on Build GRCh38
rs919938500 Chr.1: 151292624-151292624 on Build GRCh38
rs369267931 Chr.1: 151292749-151292760 on Build GRCh38
rs995727287 Chr.1: 151292837-151292844 on Build GRCh38
rs754878187 Chr.1: 151292900-151292900 on Build GRCh38
rs587600750 Chr.1: 151292828-151292828 on Build GRCh38
rs1044578917 Chr.1: 151292776-151292776 on Build GRCh38
rs778599401 Chr.1: 151293008-151293008 on Build GRCh38
rs748993864 Chr.1: 151292807-151292807 on Build GRCh38
rs1050926631 Chr.1: 151292627-151292627 on Build GRCh38
rs775958958 Chr.1: 151292587-151292587 on Build GRCh38
rs912627947 Chr.1: 151292663-151292663 on Build GRCh38
rs972688811 Chr.1: 151292614-151292614 on Build GRCh38
rs1023815557 Chr.1: 151292766-151292766 on Build GRCh38
rs376248933 Chr.1: 151292598-151292598 on Build GRCh38
rs192712769 Chr.1: 151292909-151292909 on Build GRCh38
rs1013752470 Chr.1: 151292752-151292752 on Build GRCh38
rs924404876 Chr.1: 151292709-151292709 on Build GRCh38
rs1025558522 Chr.1: 151292964-151292964 on Build GRCh38
rs1027224510 Chr.1: 151292845-151292845 on Build GRCh38
rs905571830 Chr.1: 151292767-151292767 on Build GRCh38
rs773394398 Chr.1: 151292971-151292971 on Build GRCh38
rs935356076 Chr.1: 151292714-151292714 on Build GRCh38
rs1804793 Chr.1: 151292667-151292667 on Build GRCh38
rs775986508 Chr.1: 151292827-151292827 on Build GRCh38
rs762070781 Chr.1: 151292849-151292849 on Build GRCh38
rs933808750 Chr.1: 151292620-151292620 on Build GRCh38
rs187446905 Chr.1: 151292719-151292719 on Build GRCh38
rs1015833784 Chr.1: 151292586-151292586 on Build GRCh38
rs962923953 Chr.1: 151292604-151292604 on Build GRCh38
rs138180526 Chr.1: 151292948-151292948 on Build GRCh38
rs1015719049 Chr.1: 151292823-151292823 on Build GRCh38
rs147013798 Chr.1: 151292597-151292597 on Build GRCh38
rs911301349 Chr.1: 151292655-151292655 on Build GRCh38
rs942798835 Chr.1: 151292676-151292676 on Build GRCh38
rs1002994376 Chr.1: 151292571-151292571 on Build GRCh38
rs766965683 Chr.1: 151292811-151292811 on Build GRCh38
rs370208380 Chr.1: 151292705-151292707 on Build GRCh38
rs746601111 Chr.1: 151292617-151292620 on Build GRCh38
rs770541827 Chr.1: 151292810-151292810 on Build GRCh38
rs776322518 Chr.1: 151292861-151292861 on Build GRCh38
rs748792388 Chr.1: 151292968-151292968 on Build GRCh38
rs372356107 Chr.1: 151292812-151292812 on Build GRCh38
rs777252576 Chr.1: 151292802-151292802 on Build GRCh38
rs777318720 Chr.1: 151292961-151292961 on Build GRCh38
rs878921131 Chr.1: 151292711-151292711 on Build GRCh38
rs771857158 Chr.1: 151292768-151292768 on Build GRCh38
rs368930115 Chr.1: 151292794-151292794 on Build GRCh38
rs970954148 Chr.1: 151292743-151292754 on Build GRCh38
rs1001303495 Chr.1: 151292784-151292784 on Build GRCh38
rs772558118 Chr.1: 151292822-151292822 on Build GRCh38
rs868579872 Chr.1: 151292745-151292745 on Build GRCh38
rs965459912 Chr.1: 151292863-151292863 on Build GRCh38
rs918529327 Chr.1: 151292615-151292615 on Build GRCh38
rs774919985 Chr.1: 151292821-151292822 on Build GRCh38
rs903864340 Chr.1: 151292710-151292710 on Build GRCh38

View Full Product Details