Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 19839469-19839973 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ID4
Entrez Gene ID
3400
Gene Name
inhibitor of DNA binding 4, HLH protein
Gene Aliases
IDB4, bHLHb27
UniGene
Hs.519601
Gene Chromosome Location
Chr.6: 19837370-19842200 on Build GRCh38
Transcripts
NM_001546.3

SNP Detail

SNP ID Chromosome Location
rs938521866 Chr.6: 19839617-19839618 on Build GRCh38
rs72547243 Chr.6: 19839830-19839831 on Build GRCh38
rs542455915 Chr.6: 19839946-19839946 on Build GRCh38
rs1028929737 Chr.6: 19839516-19839516 on Build GRCh38
rs887169829 Chr.6: 19839739-19839739 on Build GRCh38
rs115559225 Chr.6: 19839657-19839657 on Build GRCh38
rs143075248 Chr.6: 19839552-19839552 on Build GRCh38
rs867557445 Chr.6: 19839901-19839901 on Build GRCh38
rs956449370 Chr.6: 19839830-19839830 on Build GRCh38
rs987754355 Chr.6: 19839691-19839691 on Build GRCh38
rs921439625 Chr.6: 19839671-19839671 on Build GRCh38
rs911643954 Chr.6: 19839517-19839517 on Build GRCh38
rs1011270412 Chr.6: 19839610-19839610 on Build GRCh38
rs1016935722 Chr.6: 19839758-19839758 on Build GRCh38
rs946343723 Chr.6: 19839839-19839839 on Build GRCh38
rs748599180 Chr.6: 19839786-19839789 on Build GRCh38
rs779985344 Chr.6: 19839837-19839838 on Build GRCh38
rs959830953 Chr.6: 19839829-19839831 on Build GRCh38
rs1056327467 Chr.6: 19839737-19839737 on Build GRCh38
rs1004190067 Chr.6: 19839629-19839629 on Build GRCh38
rs889886105 Chr.6: 19839805-19839805 on Build GRCh38
rs1057113925 Chr.6: 19839618-19839618 on Build GRCh38
rs560995194 Chr.6: 19839825-19839827 on Build GRCh38
rs1002166679 Chr.6: 19839948-19839949 on Build GRCh38
rs72552349 Chr.6: 19839572-19839572 on Build GRCh38
rs774568104 Chr.6: 19839920-19839920 on Build GRCh38
rs72547244 Chr.6: 19839760-19839763 on Build GRCh38
rs537919881 Chr.6: 19839845-19839845 on Build GRCh38
rs151179104 Chr.6: 19839667-19839667 on Build GRCh38
rs558479253 Chr.6: 19839783-19839783 on Build GRCh38
rs542602931 Chr.6: 19839812-19839813 on Build GRCh38
rs1021268144 Chr.6: 19839710-19839710 on Build GRCh38
rs1021871831 Chr.6: 19839813-19839813 on Build GRCh38
rs572068184 Chr.6: 19839776-19839779 on Build GRCh38
rs939155575 Chr.6: 19839727-19839727 on Build GRCh38
rs935105875 Chr.6: 19839686-19839686 on Build GRCh38
rs988443767 Chr.6: 19839607-19839607 on Build GRCh38
rs1802808 Chr.6: 19839578-19839578 on Build GRCh38
rs940555036 Chr.6: 19839633-19839633 on Build GRCh38
rs973299100 Chr.6: 19839772-19839772 on Build GRCh38
rs750333423 Chr.6: 19839658-19839658 on Build GRCh38
rs996020621 Chr.6: 19839669-19839672 on Build GRCh38
rs1010418898 Chr.6: 19839700-19839700 on Build GRCh38
rs940607236 Chr.6: 19839767-19839767 on Build GRCh38
rs907681765 Chr.6: 19839753-19839753 on Build GRCh38
rs1001213342 Chr.6: 19839827-19839827 on Build GRCh38
rs78171725 Chr.6: 19839616-19839616 on Build GRCh38
rs955687839 Chr.6: 19839672-19839672 on Build GRCh38
rs987890638 Chr.6: 19839837-19839837 on Build GRCh38
rs775977936 Chr.6: 19839750-19839750 on Build GRCh38
rs761037704 Chr.6: 19839717-19839717 on Build GRCh38
rs777551026 Chr.6: 19839584-19839584 on Build GRCh38
rs905066432 Chr.6: 19839614-19839614 on Build GRCh38
rs35660770 Chr.6: 19839617-19839617 on Build GRCh38
rs956251563 Chr.6: 19839551-19839551 on Build GRCh38
rs866310717 Chr.6: 19839631-19839631 on Build GRCh38
rs1004327334 Chr.6: 19839748-19839748 on Build GRCh38
rs8214 Chr.6: 19839548-19839548 on Build GRCh38
rs926506221 Chr.6: 19839888-19839888 on Build GRCh38
rs893706110 Chr.6: 19839581-19839581 on Build GRCh38
rs559386429 Chr.6: 19839711-19839714 on Build GRCh38
rs1028957913 Chr.6: 19839801-19839801 on Build GRCh38
rs528136321 Chr.6: 19839542-19839542 on Build GRCh38
rs955663548 Chr.6: 19839809-19839809 on Build GRCh38
rs1014790431 Chr.6: 19839835-19839836 on Build GRCh38
rs779652840 Chr.6: 19839802-19839805 on Build GRCh38
rs565810148 Chr.6: 19839781-19839781 on Build GRCh38
rs886974013 Chr.6: 19839908-19839908 on Build GRCh38
rs940083270 Chr.6: 19839912-19839912 on Build GRCh38
rs1031830401 Chr.6: 19839814-19839814 on Build GRCh38
rs1038422752 Chr.6: 19839941-19839941 on Build GRCh38
rs1041213410 Chr.6: 19839807-19839810 on Build GRCh38
rs920525186 Chr.6: 19839838-19839841 on Build GRCh38
rs944146862 Chr.6: 19839886-19839886 on Build GRCh38
rs914944041 Chr.6: 19839694-19839694 on Build GRCh38
rs973355983 Chr.6: 19839838-19839838 on Build GRCh38
rs951453482 Chr.6: 19839627-19839627 on Build GRCh38
rs914208231 Chr.6: 19839614-19839615 on Build GRCh38
rs770326502 Chr.6: 19839719-19839719 on Build GRCh38
rs533933383 Chr.6: 19839735-19839735 on Build GRCh38
rs770560171 Chr.6: 19839791-19839795 on Build GRCh38
rs575438417 Chr.6: 19839900-19839900 on Build GRCh38
rs72552348 Chr.6: 19839519-19839519 on Build GRCh38
rs1034398909 Chr.6: 19839828-19839831 on Build GRCh38
rs551220382 Chr.6: 19839558-19839558 on Build GRCh38
rs1051987438 Chr.6: 19839512-19839512 on Build GRCh38
rs986995434 Chr.6: 19839673-19839673 on Build GRCh38
rs1045348509 Chr.6: 19839718-19839718 on Build GRCh38
rs1037048793 Chr.6: 19839647-19839647 on Build GRCh38
rs931326985 Chr.6: 19839796-19839797 on Build GRCh38
rs946653830 Chr.6: 19839697-19839697 on Build GRCh38
rs1010471514 Chr.6: 19839812-19839812 on Build GRCh38
rs968398240 Chr.6: 19839714-19839715 on Build GRCh38
rs12663067 Chr.6: 19839889-19839889 on Build GRCh38
rs554450345 Chr.6: 19839759-19839762 on Build GRCh38
rs1008928321 Chr.6: 19839525-19839526 on Build GRCh38
rs992554285 Chr.6: 19839830-19839833 on Build GRCh38
rs113066030 Chr.6: 19839846-19839846 on Build GRCh38
rs898528134 Chr.6: 19839764-19839764 on Build GRCh38
rs1044899044 Chr.6: 19839611-19839611 on Build GRCh38
rs746752501 Chr.6: 19839762-19839762 on Build GRCh38
rs375230011 Chr.6: 19839875-19839875 on Build GRCh38
rs919951465 Chr.6: 19839785-19839785 on Build GRCh38
rs956302192 Chr.6: 19839698-19839698 on Build GRCh38
rs1035072099 Chr.6: 19839617-19839617 on Build GRCh38
rs904783726 Chr.6: 19839931-19839931 on Build GRCh38
rs987355970 Chr.6: 19839810-19839810 on Build GRCh38
rs189127402 Chr.6: 19839680-19839680 on Build GRCh38
rs398084798 Chr.6: 19839625-19839625 on Build GRCh38
rs181539743 Chr.6: 19839838-19839838 on Build GRCh38

View Full Product Details