Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 27871755-27872248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H3I
Entrez Gene ID
8354
Gene Name
histone cluster 1, H3i
Gene Aliases
H3.f, H3/f, H3FF
UniGene
Hs.132854
Gene Chromosome Location
Chr.6: 27871845-27872321 on Build GRCh38
Transcripts
NM_003533.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
741687 c.156C>T p.I52I Chr.6: 27872160-27872160 on Build GRCh38
398313 c.411A>C p.*137Y Chr.6: 27871905-27871905 on Build GRCh38
6452899 c.104G>A p.G35D Chr.6: 27872212-27872212 on Build GRCh38
7371179 c.178G>A p.E60K Chr.6: 27872138-27872138 on Build GRCh38
3921226 c.231G>A p.Q77Q Chr.6: 27872085-27872085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148997555 Chr.6: 27871974-27871974 on Build GRCh38
rs751570565 Chr.6: 27871890-27871890 on Build GRCh38
rs559511508 Chr.6: 27872119-27872119 on Build GRCh38
rs926284215 Chr.6: 27872210-27872210 on Build GRCh38
rs747419710 Chr.6: 27871933-27871933 on Build GRCh38
rs747510120 Chr.6: 27871979-27871979 on Build GRCh38
rs755951115 Chr.6: 27871901-27871901 on Build GRCh38
rs763507170 Chr.6: 27871889-27871889 on Build GRCh38
rs145876619 Chr.6: 27871882-27871882 on Build GRCh38
rs768217798 Chr.6: 27872004-27872004 on Build GRCh38
rs930100293 Chr.6: 27871828-27871828 on Build GRCh38
rs764233454 Chr.6: 27872139-27872139 on Build GRCh38
rs770166385 Chr.6: 27872106-27872106 on Build GRCh38
rs773656938 Chr.6: 27871930-27871930 on Build GRCh38
rs755564606 Chr.6: 27872185-27872223 on Build GRCh38
rs757306085 Chr.6: 27872144-27872144 on Build GRCh38
rs368144811 Chr.6: 27871894-27871894 on Build GRCh38
rs773766489 Chr.6: 27871943-27871943 on Build GRCh38
rs368456166 Chr.6: 27871857-27871857 on Build GRCh38
rs1053913919 Chr.6: 27871922-27871922 on Build GRCh38
rs761449012 Chr.6: 27872015-27872015 on Build GRCh38
rs765608048 Chr.6: 27871901-27871902 on Build GRCh38
rs770440163 Chr.6: 27872178-27872178 on Build GRCh38
rs761867025 Chr.6: 27872222-27872222 on Build GRCh38
rs780465027 Chr.6: 27871915-27871915 on Build GRCh38
rs149868064 Chr.6: 27871788-27871788 on Build GRCh38
rs774035069 Chr.6: 27872007-27872007 on Build GRCh38
rs199797213 Chr.6: 27871917-27871917 on Build GRCh38
rs975924574 Chr.6: 27872132-27872132 on Build GRCh38
rs781590720 Chr.6: 27872146-27872146 on Build GRCh38
rs768591870 Chr.6: 27871856-27871856 on Build GRCh38
rs377227516 Chr.6: 27872135-27872135 on Build GRCh38
rs373244072 Chr.6: 27872136-27872136 on Build GRCh38
rs920106380 Chr.6: 27871821-27871821 on Build GRCh38
rs200525123 Chr.6: 27872172-27872172 on Build GRCh38
rs1027454605 Chr.6: 27872108-27872108 on Build GRCh38
rs747792724 Chr.6: 27872116-27872116 on Build GRCh38
rs757982209 Chr.6: 27872103-27872103 on Build GRCh38
rs1047321778 Chr.6: 27871839-27871839 on Build GRCh38
rs767611660 Chr.6: 27871950-27871950 on Build GRCh38
rs1012896431 Chr.6: 27871893-27871893 on Build GRCh38
rs777522650 Chr.6: 27872174-27872174 on Build GRCh38
rs369709227 Chr.6: 27872127-27872127 on Build GRCh38
rs764349796 Chr.6: 27872211-27872211 on Build GRCh38
rs748194741 Chr.6: 27871926-27871926 on Build GRCh38
rs778497947 Chr.6: 27872109-27872109 on Build GRCh38
rs968129383 Chr.6: 27872088-27872088 on Build GRCh38
rs751750041 Chr.6: 27872219-27872219 on Build GRCh38
rs147096368 Chr.6: 27872157-27872157 on Build GRCh38
rs528245366 Chr.6: 27872129-27872129 on Build GRCh38
rs779848426 Chr.6: 27872156-27872156 on Build GRCh38
rs963972613 Chr.6: 27872058-27872058 on Build GRCh38
rs201143748 Chr.6: 27871923-27871923 on Build GRCh38
rs751668453 Chr.6: 27872141-27872141 on Build GRCh38
rs760706393 Chr.6: 27872030-27872030 on Build GRCh38
rs142221765 Chr.6: 27872018-27872018 on Build GRCh38
rs766354606 Chr.6: 27872055-27872055 on Build GRCh38
rs746267984 Chr.6: 27872105-27872105 on Build GRCh38
rs376425183 Chr.6: 27871941-27871941 on Build GRCh38
rs374209842 Chr.6: 27872199-27872199 on Build GRCh38
rs772575974 Chr.6: 27871863-27871863 on Build GRCh38
rs750268450 Chr.6: 27871900-27871900 on Build GRCh38
rs747019671 Chr.6: 27872223-27872223 on Build GRCh38
rs370130075 Chr.6: 27872175-27872175 on Build GRCh38
rs555527061 Chr.6: 27871871-27871871 on Build GRCh38
rs773567261 Chr.6: 27871864-27871864 on Build GRCh38
rs749890672 Chr.6: 27871973-27871973 on Build GRCh38
rs760225714 Chr.6: 27872120-27872120 on Build GRCh38
rs984692698 Chr.6: 27872151-27872151 on Build GRCh38
rs200956 Chr.6: 27871968-27871968 on Build GRCh38
rs753613247 Chr.6: 27871957-27871957 on Build GRCh38
rs773344869 Chr.6: 27872023-27872023 on Build GRCh38
rs749599470 Chr.6: 27872171-27872171 on Build GRCh38
rs779153359 Chr.6: 27871964-27871964 on Build GRCh38
rs769559224 Chr.6: 27872196-27872196 on Build GRCh38
rs753196469 Chr.6: 27871940-27871940 on Build GRCh38
rs756043095 Chr.6: 27871956-27871956 on Build GRCh38
rs765421554 Chr.6: 27872073-27872073 on Build GRCh38
rs771253043 Chr.6: 27871937-27871937 on Build GRCh38
rs780482781 Chr.6: 27872168-27872168 on Build GRCh38
rs372722417 Chr.6: 27872229-27872229 on Build GRCh38
rs549064212 Chr.6: 27872056-27872056 on Build GRCh38
rs772596697 Chr.6: 27871928-27871928 on Build GRCh38
rs748197388 Chr.6: 27871967-27871967 on Build GRCh38
rs762695270 Chr.6: 27872200-27872200 on Build GRCh38
rs372624094 Chr.6: 27871847-27871847 on Build GRCh38
rs776497019 Chr.6: 27872179-27872179 on Build GRCh38
rs563300662 Chr.6: 27871994-27871994 on Build GRCh38
rs749942493 Chr.6: 27872136-27872137 on Build GRCh38
rs758539201 Chr.6: 27872091-27872091 on Build GRCh38
rs781236409 Chr.6: 27871896-27871896 on Build GRCh38
rs758933281 Chr.6: 27872075-27872077 on Build GRCh38
rs755163039 Chr.6: 27871963-27871963 on Build GRCh38
rs749470205 Chr.6: 27871919-27871919 on Build GRCh38
rs750356284 Chr.6: 27871952-27871952 on Build GRCh38
rs767622437 Chr.6: 27872224-27872224 on Build GRCh38
rs770793237 Chr.6: 27872121-27872121 on Build GRCh38
rs760013135 Chr.6: 27871898-27871898 on Build GRCh38
rs755265369 Chr.6: 27871785-27871785 on Build GRCh38
rs751578458 Chr.6: 27872102-27872102 on Build GRCh38
rs776604676 Chr.6: 27872125-27872125 on Build GRCh38
rs535583756 Chr.6: 27871824-27871824 on Build GRCh38
rs771342766 Chr.6: 27871983-27871983 on Build GRCh38
rs761339099 Chr.6: 27871948-27871948 on Build GRCh38
rs767003515 Chr.6: 27872078-27872079 on Build GRCh38
rs775189331 Chr.6: 27871938-27871938 on Build GRCh38
rs759503770 Chr.6: 27872194-27872194 on Build GRCh38
rs776870520 Chr.6: 27871992-27871992 on Build GRCh38
rs1000620583 Chr.6: 27871918-27871918 on Build GRCh38
rs200523036 Chr.6: 27871877-27871877 on Build GRCh38
rs771795877 Chr.6: 27872118-27872118 on Build GRCh38
rs776940405 Chr.6: 27871855-27871856 on Build GRCh38
rs576128698 Chr.6: 27871811-27871811 on Build GRCh38
rs1008216616 Chr.6: 27871986-27871986 on Build GRCh38
rs1044888012 Chr.6: 27871911-27871911 on Build GRCh38

View Full Product Details