Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 28276890-28277403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZSCAN26
Entrez Gene ID
7741
Gene Name
zinc finger and SCAN domain containing 26
Gene Aliases
SRE-ZBP, SREZBP, ZNF187
UniGene
Hs.157883
Gene Chromosome Location
Chr.6: 28267010-28278224 on Build GRCh38
Transcripts
NM_001023560.3 NM_001111039.2 NM_001287421.1 NM_001287422.1 NM_152736.5 XM_011514862.2 XM_011514864.1 XM_011514866.1 XM_011514867.1 XM_011514869.1 XM_017011264.1 XM_017011265.1

SNP Detail

SNP ID Chromosome Location
rs188497598 Chr.6: 28277232-28277232 on Build GRCh38
rs779641149 Chr.6: 28276965-28276965 on Build GRCh38
rs752744669 Chr.6: 28276980-28276981 on Build GRCh38
rs771309593 Chr.6: 28277008-28277008 on Build GRCh38
rs972112881 Chr.6: 28276974-28276974 on Build GRCh38
rs962136295 Chr.6: 28276973-28276973 on Build GRCh38
rs375309268 Chr.6: 28276954-28276954 on Build GRCh38
rs934425617 Chr.6: 28277139-28277139 on Build GRCh38
rs909249453 Chr.6: 28277010-28277010 on Build GRCh38
rs529692636 Chr.6: 28277069-28277069 on Build GRCh38
rs756093540 Chr.6: 28277130-28277130 on Build GRCh38
rs891960370 Chr.6: 28277204-28277204 on Build GRCh38
rs751403747 Chr.6: 28276943-28276943 on Build GRCh38
rs1053217880 Chr.6: 28277203-28277203 on Build GRCh38
rs888320862 Chr.6: 28276939-28276939 on Build GRCh38
rs548519395 Chr.6: 28277154-28277154 on Build GRCh38
rs556483159 Chr.6: 28277377-28277377 on Build GRCh38
rs888342740 Chr.6: 28277231-28277231 on Build GRCh38
rs367896865 Chr.6: 28277319-28277319 on Build GRCh38
rs537254525 Chr.6: 28276979-28276979 on Build GRCh38
rs376665599 Chr.6: 28277050-28277050 on Build GRCh38
rs112468414 Chr.6: 28277222-28277222 on Build GRCh38
rs1016379009 Chr.6: 28276960-28276960 on Build GRCh38
rs748499677 Chr.6: 28277031-28277031 on Build GRCh38
rs745864418 Chr.6: 28276961-28276961 on Build GRCh38
rs774057290 Chr.6: 28277019-28277019 on Build GRCh38
rs1026192003 Chr.6: 28277378-28277378 on Build GRCh38
rs960276641 Chr.6: 28276986-28276986 on Build GRCh38
rs975959439 Chr.6: 28277373-28277373 on Build GRCh38
rs1040736733 Chr.6: 28277152-28277152 on Build GRCh38
rs558740794 Chr.6: 28277358-28277358 on Build GRCh38
rs557609192 Chr.6: 28276964-28276964 on Build GRCh38
rs1017254185 Chr.6: 28277361-28277362 on Build GRCh38
rs769518702 Chr.6: 28277046-28277046 on Build GRCh38
rs916143982 Chr.6: 28276993-28276993 on Build GRCh38
rs374650003 Chr.6: 28277109-28277109 on Build GRCh38
rs568471716 Chr.6: 28277159-28277159 on Build GRCh38
rs907476339 Chr.6: 28277108-28277108 on Build GRCh38
rs750281794 Chr.6: 28277122-28277122 on Build GRCh38
rs1005447135 Chr.6: 28277287-28277287 on Build GRCh38
rs116288753 Chr.6: 28277044-28277044 on Build GRCh38
rs369045087 Chr.6: 28277015-28277015 on Build GRCh38
rs965961406 Chr.6: 28277365-28277365 on Build GRCh38
rs932300364 Chr.6: 28277206-28277206 on Build GRCh38
rs377608230 Chr.6: 28277118-28277118 on Build GRCh38
rs531656144 Chr.6: 28277113-28277113 on Build GRCh38
rs148763465 Chr.6: 28277170-28277170 on Build GRCh38
rs757181763 Chr.6: 28276953-28276953 on Build GRCh38
rs1015447272 Chr.6: 28277318-28277318 on Build GRCh38
rs765548403 Chr.6: 28277091-28277091 on Build GRCh38
rs192023318 Chr.6: 28277006-28277006 on Build GRCh38
rs763923028 Chr.6: 28276926-28276926 on Build GRCh38
rs371333289 Chr.6: 28277032-28277032 on Build GRCh38
rs753022561 Chr.6: 28277099-28277099 on Build GRCh38
rs1035776512 Chr.6: 28277363-28277363 on Build GRCh38
rs1005784837 Chr.6: 28276949-28276949 on Build GRCh38
rs779368832 Chr.6: 28277136-28277136 on Build GRCh38
rs748885770 Chr.6: 28276968-28276968 on Build GRCh38
rs776785015 Chr.6: 28277064-28277064 on Build GRCh38
rs369880461 Chr.6: 28277072-28277072 on Build GRCh38
rs991796657 Chr.6: 28276989-28276989 on Build GRCh38
rs758411154 Chr.6: 28277070-28277071 on Build GRCh38

View Full Product Details