Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39334984-39335531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF6
Entrez Gene ID
221458
Gene Name
kinesin family member 6
Gene Aliases
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
UniGene
Hs.588202
Gene Chromosome Location
Chr.6: 39335100-39725405 on Build GRCh38
Transcripts
NM_001289020.1 NM_001289021.1 NM_001289024.1 NM_145027.4

SNP Detail

SNP ID Chromosome Location
rs923741875 Chr.6: 39335142-39335142 on Build GRCh38
rs548477620 Chr.6: 39335205-39335205 on Build GRCh38
rs917850083 Chr.6: 39335220-39335220 on Build GRCh38
rs914890891 Chr.6: 39335163-39335163 on Build GRCh38
rs994602971 Chr.6: 39335024-39335024 on Build GRCh38
rs897242351 Chr.6: 39335327-39335327 on Build GRCh38
rs897348280 Chr.6: 39335054-39335054 on Build GRCh38
rs781072162 Chr.6: 39335092-39335092 on Build GRCh38
rs941427964 Chr.6: 39335295-39335295 on Build GRCh38
rs979568129 Chr.6: 39335439-39335439 on Build GRCh38
rs1017883551 Chr.6: 39335402-39335402 on Build GRCh38
rs530786420 Chr.6: 39335224-39335224 on Build GRCh38
rs574638149 Chr.6: 39335438-39335438 on Build GRCh38
rs539423221 Chr.6: 39335359-39335359 on Build GRCh38
rs1030292320 Chr.6: 39335372-39335372 on Build GRCh38
rs1038465863 Chr.6: 39335317-39335317 on Build GRCh38
rs554799040 Chr.6: 39335422-39335422 on Build GRCh38
rs532668338 Chr.6: 39335154-39335154 on Build GRCh38
rs144747535 Chr.6: 39335363-39335363 on Build GRCh38
rs888419377 Chr.6: 39335397-39335397 on Build GRCh38
rs775625832 Chr.6: 39335139-39335139 on Build GRCh38
rs568454515 Chr.6: 39335207-39335207 on Build GRCh38
rs185264696 Chr.6: 39335019-39335019 on Build GRCh38
rs557259004 Chr.6: 39335482-39335482 on Build GRCh38
rs72858469 Chr.6: 39335390-39335390 on Build GRCh38
rs546351750 Chr.6: 39335234-39335234 on Build GRCh38
rs1887716 Chr.6: 39335056-39335056 on Build GRCh38
rs570530741 Chr.6: 39335238-39335238 on Build GRCh38
rs956681517 Chr.6: 39335465-39335465 on Build GRCh38
rs956713982 Chr.6: 39335467-39335467 on Build GRCh38
rs897217216 Chr.6: 39335020-39335020 on Build GRCh38
rs16891944 Chr.6: 39335506-39335506 on Build GRCh38
rs906441816 Chr.6: 39335444-39335444 on Build GRCh38
rs766585350 Chr.6: 39335043-39335043 on Build GRCh38
rs866855069 Chr.6: 39335175-39335175 on Build GRCh38
rs1030806306 Chr.6: 39335457-39335457 on Build GRCh38
rs114582772 Chr.6: 39335162-39335162 on Build GRCh38
rs1049974047 Chr.6: 39335188-39335188 on Build GRCh38
rs989207509 Chr.6: 39335503-39335503 on Build GRCh38
rs979234180 Chr.6: 39335140-39335140 on Build GRCh38
rs917796232 Chr.6: 39335497-39335497 on Build GRCh38
rs1023432817 Chr.6: 39335074-39335074 on Build GRCh38
rs770046813 Chr.6: 39335114-39335114 on Build GRCh38
rs991999462 Chr.6: 39335472-39335472 on Build GRCh38
rs566837496 Chr.6: 39335367-39335367 on Build GRCh38
rs190137715 Chr.6: 39335451-39335451 on Build GRCh38
rs970568509 Chr.6: 39335075-39335075 on Build GRCh38
rs563274951 Chr.6: 39335072-39335072 on Build GRCh38
rs531115051 Chr.6: 39335233-39335233 on Build GRCh38
rs72858468 Chr.6: 39335100-39335100 on Build GRCh38
rs985099528 Chr.6: 39335086-39335086 on Build GRCh38
rs559153397 Chr.6: 39335113-39335113 on Build GRCh38
rs532104217 Chr.6: 39335088-39335088 on Build GRCh38
rs947988766 Chr.6: 39335226-39335226 on Build GRCh38
rs1012195091 Chr.6: 39335428-39335428 on Build GRCh38
rs530126032 Chr.6: 39335475-39335475 on Build GRCh38
rs756237670 Chr.6: 39335377-39335377 on Build GRCh38

View Full Product Details