Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152310225-152310719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG
Entrez Gene ID
2312
Gene Name
filaggrin
Gene Aliases
ATOD2
UniGene
Hs.654510
Gene Chromosome Location
Chr.1: 152302175-152325203 on Build GRCh38
Transcripts
NM_002016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7249892 c.4568C>T p.T1523I Chr.1: 152310318-152310318 on Build GRCh38
2206617 c.4577G>A p.G1526E Chr.1: 152310309-152310309 on Build GRCh38
4548517 c.4490G>A p.R1497K Chr.1: 152310396-152310396 on Build GRCh38
6572961 c.4210G>C p.D1404H Chr.1: 152310676-152310676 on Build GRCh38
462986 c.4493G>A p.G1498E Chr.1: 152310393-152310393 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145675213 Chr.1: 152310480-152310480 on Build GRCh38
rs777512372 Chr.1: 152310286-152310286 on Build GRCh38
rs112252908 Chr.1: 152310697-152310697 on Build GRCh38
rs538202195 Chr.1: 152310298-152310298 on Build GRCh38
rs370219834 Chr.1: 152310383-152310383 on Build GRCh38
rs765229810 Chr.1: 152310651-152310651 on Build GRCh38
rs781640209 Chr.1: 152310632-152310632 on Build GRCh38
rs547750140 Chr.1: 152310439-152310439 on Build GRCh38
rs202237346 Chr.1: 152310523-152310523 on Build GRCh38
rs200756104 Chr.1: 152310331-152310331 on Build GRCh38
rs750640524 Chr.1: 152310536-152310536 on Build GRCh38
rs199810477 Chr.1: 152310351-152310351 on Build GRCh38
rs749451902 Chr.1: 152310614-152310615 on Build GRCh38
rs201565622 Chr.1: 152310584-152310584 on Build GRCh38
rs371007229 Chr.1: 152310248-152310248 on Build GRCh38
rs752267105 Chr.1: 152310327-152310327 on Build GRCh38
rs141177690 Chr.1: 152310664-152310664 on Build GRCh38
rs759296038 Chr.1: 152310370-152310370 on Build GRCh38
rs779352008 Chr.1: 152310554-152310554 on Build GRCh38
rs752409908 Chr.1: 152310506-152310507 on Build GRCh38
rs367594853 Chr.1: 152310576-152310576 on Build GRCh38
rs12732920 Chr.1: 152310476-152310476 on Build GRCh38
rs147032729 Chr.1: 152310647-152310647 on Build GRCh38
rs200422428 Chr.1: 152310289-152310289 on Build GRCh38
rs554065456 Chr.1: 152310520-152310520 on Build GRCh38
rs757349097 Chr.1: 152310530-152310530 on Build GRCh38
rs373351328 Chr.1: 152310636-152310636 on Build GRCh38
rs765038850 Chr.1: 152310416-152310416 on Build GRCh38
rs769031008 Chr.1: 152310598-152310598 on Build GRCh38
rs752573463 Chr.1: 152310360-152310360 on Build GRCh38
rs375084109 Chr.1: 152310619-152310619 on Build GRCh38
rs754660081 Chr.1: 152310666-152310666 on Build GRCh38
rs780674540 Chr.1: 152310373-152310373 on Build GRCh38
rs145478188 Chr.1: 152310592-152310592 on Build GRCh38
rs150981235 Chr.1: 152310641-152310641 on Build GRCh38
rs149975479 Chr.1: 152310283-152310283 on Build GRCh38
rs772962994 Chr.1: 152310456-152310456 on Build GRCh38
rs142950612 Chr.1: 152310521-152310521 on Build GRCh38
rs753353085 Chr.1: 152310367-152310367 on Build GRCh38
rs762625752 Chr.1: 152310348-152310348 on Build GRCh38
rs776816341 Chr.1: 152310309-152310309 on Build GRCh38
rs374409476 Chr.1: 152310398-152310398 on Build GRCh38
rs771673904 Chr.1: 152310549-152310549 on Build GRCh38
rs142928947 Chr.1: 152310316-152310316 on Build GRCh38
rs148844389 Chr.1: 152310607-152310607 on Build GRCh38
rs779141143 Chr.1: 152310479-152310479 on Build GRCh38
rs761586008 Chr.1: 152310649-152310649 on Build GRCh38
rs747402421 Chr.1: 152310597-152310597 on Build GRCh38
rs113636527 Chr.1: 152310541-152310541 on Build GRCh38
rs1050899017 Chr.1: 152310440-152310440 on Build GRCh38
rs759936037 Chr.1: 152310510-152310510 on Build GRCh38
rs562855035 Chr.1: 152310401-152310401 on Build GRCh38
rs12750571 Chr.1: 152310577-152310577 on Build GRCh38
rs774411752 Chr.1: 152310675-152310675 on Build GRCh38
rs146686141 Chr.1: 152310466-152310466 on Build GRCh38
rs776694382 Chr.1: 152310450-152310450 on Build GRCh38
rs71626706 Chr.1: 152310434-152310434 on Build GRCh38
rs768713890 Chr.1: 152310344-152310344 on Build GRCh38
rs767607830 Chr.1: 152310255-152310255 on Build GRCh38
rs748238097 Chr.1: 152310430-152310430 on Build GRCh38
rs144605772 Chr.1: 152310461-152310461 on Build GRCh38
rs377074122 Chr.1: 152310684-152310684 on Build GRCh38
rs369925879 Chr.1: 152310659-152310659 on Build GRCh38
rs549641255 Chr.1: 152310284-152310284 on Build GRCh38
rs755889087 Chr.1: 152310509-152310510 on Build GRCh38
rs761473196 Chr.1: 152310452-152310452 on Build GRCh38
rs757949828 Chr.1: 152310662-152310662 on Build GRCh38
rs267598030 Chr.1: 152310589-152310589 on Build GRCh38
rs753536196 Chr.1: 152310611-152310611 on Build GRCh38
rs189279130 Chr.1: 152310604-152310604 on Build GRCh38
rs375584758 Chr.1: 152310382-152310382 on Build GRCh38
rs754487205 Chr.1: 152310280-152310280 on Build GRCh38
rs545558175 Chr.1: 152310599-152310599 on Build GRCh38
rs781315651 Chr.1: 152310486-152310486 on Build GRCh38
rs201782279 Chr.1: 152310338-152310338 on Build GRCh38
rs147929070 Chr.1: 152310313-152310313 on Build GRCh38
rs12750081 Chr.1: 152310318-152310318 on Build GRCh38
rs375923587 Chr.1: 152310608-152310608 on Build GRCh38
rs374456748 Chr.1: 152310393-152310393 on Build GRCh38
rs761352191 Chr.1: 152310407-152310407 on Build GRCh38
rs767528401 Chr.1: 152310481-152310481 on Build GRCh38
rs143101193 Chr.1: 152310417-152310417 on Build GRCh38
rs777741418 Chr.1: 152310593-152310593 on Build GRCh38
rs372630215 Chr.1: 152310562-152310562 on Build GRCh38
rs760983085 Chr.1: 152310259-152310259 on Build GRCh38
rs779724442 Chr.1: 152310624-152310624 on Build GRCh38
rs541202491 Chr.1: 152310605-152310605 on Build GRCh38
rs138166932 Chr.1: 152310656-152310656 on Build GRCh38
rs201320426 Chr.1: 152310320-152310320 on Build GRCh38
rs955282900 Chr.1: 152310691-152310691 on Build GRCh38
rs376165154 Chr.1: 152310683-152310683 on Build GRCh38
rs762000612 Chr.1: 152310346-152310346 on Build GRCh38
rs141337128 Chr.1: 152310256-152310256 on Build GRCh38
rs3120648 Chr.1: 152310423-152310423 on Build GRCh38
rs745318087 Chr.1: 152310588-152310588 on Build GRCh38
rs759757676 Chr.1: 152310251-152310251 on Build GRCh38
rs373475578 Chr.1: 152310339-152310339 on Build GRCh38
rs143805317 Chr.1: 152310573-152310573 on Build GRCh38
rs147249511 Chr.1: 152310305-152310305 on Build GRCh38
rs537358852 Chr.1: 152310473-152310473 on Build GRCh38
rs758833597 Chr.1: 152310418-152310418 on Build GRCh38
rs753011537 Chr.1: 152310519-152310519 on Build GRCh38
rs899710580 Chr.1: 152310254-152310254 on Build GRCh38
rs755509585 Chr.1: 152310428-152310428 on Build GRCh38
rs769152063 Chr.1: 152310453-152310453 on Build GRCh38
rs755820548 Chr.1: 152310285-152310285 on Build GRCh38
rs767316041 Chr.1: 152310325-152310325 on Build GRCh38
rs529959776 Chr.1: 152310630-152310630 on Build GRCh38
rs376426043 Chr.1: 152310402-152310402 on Build GRCh38
rs370908995 Chr.1: 152310504-152310504 on Build GRCh38
rs147239265 Chr.1: 152310406-152310406 on Build GRCh38
rs749729144 Chr.1: 152310288-152310288 on Build GRCh38
rs751085109 Chr.1: 152310433-152310434 on Build GRCh38
rs1024031276 Chr.1: 152310380-152310380 on Build GRCh38
rs199927704 Chr.1: 152310603-152310603 on Build GRCh38
rs764185577 Chr.1: 152310260-152310260 on Build GRCh38
rs200740047 Chr.1: 152310433-152310433 on Build GRCh38
rs775672711 Chr.1: 152310301-152310301 on Build GRCh38
rs200551704 Chr.1: 152310465-152310465 on Build GRCh38
rs755568153 Chr.1: 152310330-152310330 on Build GRCh38
rs147463305 Chr.1: 152310542-152310542 on Build GRCh38
rs747246697 Chr.1: 152310600-152310600 on Build GRCh38
rs746367318 Chr.1: 152310291-152310291 on Build GRCh38
rs369287142 Chr.1: 152310617-152310617 on Build GRCh38
rs770501167 Chr.1: 152310506-152310506 on Build GRCh38
rs145962910 Chr.1: 152310646-152310646 on Build GRCh38
rs202069386 Chr.1: 152310545-152310545 on Build GRCh38
rs766626335 Chr.1: 152310419-152310419 on Build GRCh38
rs761046630 Chr.1: 152310553-152310553 on Build GRCh38
rs749835557 Chr.1: 152310685-152310685 on Build GRCh38
rs757429282 Chr.1: 152310478-152310478 on Build GRCh38
rs771524427 Chr.1: 152310692-152310692 on Build GRCh38
rs757749982 Chr.1: 152310334-152310334 on Build GRCh38
rs559621510 Chr.1: 152310262-152310262 on Build GRCh38
rs909581264 Chr.1: 152310414-152310414 on Build GRCh38
rs751489934 Chr.1: 152310277-152310277 on Build GRCh38
rs760272161 Chr.1: 152310362-152310362 on Build GRCh38
rs534103512 Chr.1: 152310512-152310512 on Build GRCh38
rs778425884 Chr.1: 152310585-152310585 on Build GRCh38
rs762458364 Chr.1: 152310505-152310505 on Build GRCh38
rs779446237 Chr.1: 152310335-152310335 on Build GRCh38
rs368302261 Chr.1: 152310547-152310547 on Build GRCh38
rs372594855 Chr.1: 152310681-152310681 on Build GRCh38
rs755099094 Chr.1: 152310378-152310378 on Build GRCh38
rs758479567 Chr.1: 152310571-152310571 on Build GRCh38
rs538907901 Chr.1: 152310614-152310614 on Build GRCh38
rs773978032 Chr.1: 152310507-152310507 on Build GRCh38
rs748391584 Chr.1: 152310634-152310634 on Build GRCh38
rs535024448 Chr.1: 152310534-152310534 on Build GRCh38
rs772413729 Chr.1: 152310292-152310292 on Build GRCh38
rs747439047 Chr.1: 152310376-152310376 on Build GRCh38
rs149284528 Chr.1: 152310357-152310357 on Build GRCh38
rs757004590 Chr.1: 152310369-152310369 on Build GRCh38
rs746736722 Chr.1: 152310449-152310449 on Build GRCh38
rs180768115 Chr.1: 152310342-152310342 on Build GRCh38
rs11204978 Chr.1: 152310441-152310441 on Build GRCh38
rs146433981 Chr.1: 152310336-152310336 on Build GRCh38
rs536230632 Chr.1: 152310271-152310271 on Build GRCh38
rs138596526 Chr.1: 152310353-152310353 on Build GRCh38
rs766298491 Chr.1: 152310312-152310313 on Build GRCh38
rs769718883 Chr.1: 152310503-152310503 on Build GRCh38
rs551853917 Chr.1: 152310496-152310496 on Build GRCh38
rs376213213 Chr.1: 152310548-152310548 on Build GRCh38
rs758674459 Chr.1: 152310371-152310371 on Build GRCh38
rs200558860 Chr.1: 152310637-152310637 on Build GRCh38
rs757562290 Chr.1: 152310689-152310692 on Build GRCh38
rs745444808 Chr.1: 152310623-152310623 on Build GRCh38
rs769622877 Chr.1: 152310543-152310543 on Build GRCh38
rs763191143 Chr.1: 152310639-152310639 on Build GRCh38
rs765382640 Chr.1: 152310570-152310570 on Build GRCh38
rs951162985 Chr.1: 152310517-152310517 on Build GRCh38
rs775841485 Chr.1: 152310552-152310552 on Build GRCh38
rs754859409 Chr.1: 152310437-152310444 on Build GRCh38
rs753681644 Chr.1: 152310648-152310648 on Build GRCh38
rs375638840 Chr.1: 152310364-152310364 on Build GRCh38
rs766357585 Chr.1: 152310270-152310270 on Build GRCh38
rs768563961 Chr.1: 152310481-152310481 on Build GRCh38
rs200013648 Chr.1: 152310475-152310475 on Build GRCh38
rs768142420 Chr.1: 152310631-152310631 on Build GRCh38
rs775547727 Chr.1: 152310400-152310400 on Build GRCh38
rs571541022 Chr.1: 152310498-152310498 on Build GRCh38
rs758127913 Chr.1: 152310268-152310268 on Build GRCh38
rs765808900 Chr.1: 152310365-152310365 on Build GRCh38

View Full Product Details