Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42965172-42965676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX6
Entrez Gene ID
5190
Gene Name
peroxisomal biogenesis factor 6
Gene Aliases
HMLR2, PAF-2, PAF2, PBD4A, PDB4B, PXAAA1
UniGene
Hs.656425
Gene Chromosome Location
Chr.6: 42963873-42979243 on Build GRCh38
Transcripts
NM_000287.3 NM_001316313.1 NR_133009.1 XM_011514661.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6744186 c.2483A>G p.Q828R Chr.6: 42965357-42965357 on Build GRCh38
339874 c.2524G>T p.V842L Chr.6: 42965316-42965316 on Build GRCh38
360994 c.2588G>A p.R863K Chr.6: 42965252-42965252 on Build GRCh38
5345182 c.2579G>A p.R860Q Chr.6: 42965261-42965261 on Build GRCh38
4565750 c.2487_2488CC>TT p.L830F Chr.6: 42965352-42965353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61753231 Chr.6: 42965261-42965261 on Build GRCh38
rs775778545 Chr.6: 42965347-42965347 on Build GRCh38
rs986551351 Chr.6: 42965475-42965475 on Build GRCh38
rs755834884 Chr.6: 42965207-42965207 on Build GRCh38
rs267608244 Chr.6: 42965294-42965294 on Build GRCh38
rs34016292 Chr.6: 42965648-42965648 on Build GRCh38
rs763098526 Chr.6: 42965254-42965254 on Build GRCh38
rs779395697 Chr.6: 42965208-42965208 on Build GRCh38
rs149571630 Chr.6: 42965583-42965583 on Build GRCh38
rs758760645 Chr.6: 42965223-42965223 on Build GRCh38
rs892554812 Chr.6: 42965234-42965234 on Build GRCh38
rs1017258935 Chr.6: 42965552-42965552 on Build GRCh38
rs537970715 Chr.6: 42965226-42965226 on Build GRCh38
rs774457810 Chr.6: 42965402-42965402 on Build GRCh38
rs778417057 Chr.6: 42965232-42965232 on Build GRCh38
rs951235418 Chr.6: 42965469-42965469 on Build GRCh38
rs757044378 Chr.6: 42965199-42965199 on Build GRCh38
rs748840257 Chr.6: 42965214-42965214 on Build GRCh38
rs759682469 Chr.6: 42965328-42965328 on Build GRCh38
rs746036884 Chr.6: 42965246-42965246 on Build GRCh38
rs753497339 Chr.6: 42965304-42965304 on Build GRCh38
rs761084004 Chr.6: 42965206-42965206 on Build GRCh38
rs372900304 Chr.6: 42965632-42965632 on Build GRCh38
rs899386415 Chr.6: 42965407-42965407 on Build GRCh38
rs772811890 Chr.6: 42965644-42965644 on Build GRCh38
rs1027403473 Chr.6: 42965446-42965446 on Build GRCh38
rs187435179 Chr.6: 42965309-42965309 on Build GRCh38
rs61753230 Chr.6: 42965262-42965262 on Build GRCh38
rs533831463 Chr.6: 42965346-42965346 on Build GRCh38
rs139250435 Chr.6: 42965450-42965450 on Build GRCh38
rs369587901 Chr.6: 42965289-42965289 on Build GRCh38
rs776069157 Chr.6: 42965197-42965197 on Build GRCh38
rs764227040 Chr.6: 42965255-42965255 on Build GRCh38
rs372243125 Chr.6: 42965517-42965517 on Build GRCh38
rs368362933 Chr.6: 42965316-42965316 on Build GRCh38
rs1025280370 Chr.6: 42965461-42965461 on Build GRCh38
rs770076606 Chr.6: 42965247-42965247 on Build GRCh38
rs766143141 Chr.6: 42965654-42965654 on Build GRCh38
rs267608242 Chr.6: 42965370-42965370 on Build GRCh38
rs768703242 Chr.6: 42965381-42965381 on Build GRCh38
rs781648628 Chr.6: 42965330-42965330 on Build GRCh38
rs746258580 Chr.6: 42965334-42965334 on Build GRCh38
rs61753226 Chr.6: 42965306-42965306 on Build GRCh38
rs553917763 Chr.6: 42965423-42965423 on Build GRCh38
rs975984069 Chr.6: 42965566-42965566 on Build GRCh38
rs760511051 Chr.6: 42965645-42965645 on Build GRCh38
rs1007319339 Chr.6: 42965457-42965457 on Build GRCh38
rs775374576 Chr.6: 42965252-42965252 on Build GRCh38
rs1019907560 Chr.6: 42965557-42965557 on Build GRCh38
rs777175136 Chr.6: 42965240-42965240 on Build GRCh38
rs975211816 Chr.6: 42965596-42965596 on Build GRCh38
rs749533099 Chr.6: 42965352-42965352 on Build GRCh38
rs755677982 Chr.6: 42965301-42965301 on Build GRCh38
rs267608243 Chr.6: 42965358-42965358 on Build GRCh38
rs146266265 Chr.6: 42965606-42965606 on Build GRCh38
rs370514392 Chr.6: 42965196-42965196 on Build GRCh38
rs536278797 Chr.6: 42965478-42965478 on Build GRCh38
rs761702297 Chr.6: 42965419-42965419 on Build GRCh38
rs766181165 Chr.6: 42965421-42965421 on Build GRCh38
rs925207866 Chr.6: 42965605-42965605 on Build GRCh38
rs774259819 Chr.6: 42965259-42965259 on Build GRCh38
rs747574572 Chr.6: 42965239-42965239 on Build GRCh38
rs767271773 Chr.6: 42965281-42965281 on Build GRCh38

View Full Product Details