Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43182068-43182563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL9
Entrez Gene ID
23113
Gene Name
cullin 9
Gene Aliases
H7AP1, PARC
UniGene
Hs.485434
Gene Chromosome Location
Chr.6: 43182170-43224587 on Build GRCh38
Transcripts
NM_015089.3 XM_011514422.1 XM_011514423.2 XM_011514424.1 XM_011514425.1 XM_011514426.1 XM_011514428.1 XM_011514430.1 XM_017010589.1

SNP Detail

SNP ID Chromosome Location
rs958232899 Chr.6: 43182408-43182408 on Build GRCh38
rs927648750 Chr.6: 43182461-43182461 on Build GRCh38
rs372896905 Chr.6: 43182088-43182088 on Build GRCh38
rs992347363 Chr.6: 43182346-43182346 on Build GRCh38
rs966039792 Chr.6: 43182360-43182360 on Build GRCh38
rs971139904 Chr.6: 43182403-43182403 on Build GRCh38
rs941307749 Chr.6: 43182202-43182202 on Build GRCh38
rs995603667 Chr.6: 43182127-43182127 on Build GRCh38
rs1051574780 Chr.6: 43182242-43182242 on Build GRCh38
rs778520192 Chr.6: 43182516-43182516 on Build GRCh38
rs143033117 Chr.6: 43182515-43182515 on Build GRCh38
rs753764795 Chr.6: 43182402-43182402 on Build GRCh38
rs533662124 Chr.6: 43182229-43182229 on Build GRCh38
rs1036643589 Chr.6: 43182250-43182250 on Build GRCh38
rs564809860 Chr.6: 43182220-43182220 on Build GRCh38
rs955802865 Chr.6: 43182149-43182149 on Build GRCh38
rs1019615524 Chr.6: 43182359-43182359 on Build GRCh38
rs529401299 Chr.6: 43182368-43182368 on Build GRCh38
rs1038734830 Chr.6: 43182512-43182512 on Build GRCh38
rs929762356 Chr.6: 43182536-43182536 on Build GRCh38
rs562526979 Chr.6: 43182111-43182111 on Build GRCh38
rs545004756 Chr.6: 43182190-43182190 on Build GRCh38
rs947793764 Chr.6: 43182421-43182421 on Build GRCh38
rs897016783 Chr.6: 43182232-43182232 on Build GRCh38
rs906433666 Chr.6: 43182093-43182093 on Build GRCh38
rs947740071 Chr.6: 43182213-43182213 on Build GRCh38
rs917009193 Chr.6: 43182153-43182153 on Build GRCh38
rs994262425 Chr.6: 43182237-43182237 on Build GRCh38
rs576134072 Chr.6: 43182148-43182148 on Build GRCh38
rs1005137017 Chr.6: 43182099-43182099 on Build GRCh38
rs369076811 Chr.6: 43182388-43182388 on Build GRCh38
rs149448744 Chr.6: 43182471-43182471 on Build GRCh38
rs954087428 Chr.6: 43182331-43182331 on Build GRCh38
rs547172421 Chr.6: 43182297-43182297 on Build GRCh38
rs949687906 Chr.6: 43182165-43182165 on Build GRCh38
rs898991170 Chr.6: 43182270-43182270 on Build GRCh38
rs1018303863 Chr.6: 43182292-43182292 on Build GRCh38
rs978366538 Chr.6: 43182367-43182367 on Build GRCh38
rs926992682 Chr.6: 43182490-43182490 on Build GRCh38
rs371155620 Chr.6: 43182147-43182147 on Build GRCh38
rs940637039 Chr.6: 43182467-43182467 on Build GRCh38
rs924116295 Chr.6: 43182188-43182188 on Build GRCh38
rs978696789 Chr.6: 43182157-43182157 on Build GRCh38
rs1031161357 Chr.6: 43182303-43182303 on Build GRCh38
rs916290521 Chr.6: 43182212-43182212 on Build GRCh38
rs766436067 Chr.6: 43182353-43182353 on Build GRCh38
rs547155958 Chr.6: 43182110-43182110 on Build GRCh38
rs537893158 Chr.6: 43182483-43182483 on Build GRCh38
rs148364641 Chr.6: 43182538-43182538 on Build GRCh38
rs989000861 Chr.6: 43182152-43182152 on Build GRCh38
rs1004784149 Chr.6: 43182223-43182223 on Build GRCh38
rs549241001 Chr.6: 43182407-43182407 on Build GRCh38
rs1044054798 Chr.6: 43182178-43182178 on Build GRCh38
rs754142599 Chr.6: 43182274-43182274 on Build GRCh38
rs764219533 Chr.6: 43182136-43182136 on Build GRCh38
rs916957500 Chr.6: 43182352-43182352 on Build GRCh38
rs1046071430 Chr.6: 43182422-43182422 on Build GRCh38
rs80140174 Chr.6: 43182299-43182299 on Build GRCh38
rs374866441 Chr.6: 43182195-43182195 on Build GRCh38
rs186210709 Chr.6: 43182384-43182384 on Build GRCh38
rs957131536 Chr.6: 43182318-43182318 on Build GRCh38
rs764309498 Chr.6: 43182174-43182174 on Build GRCh38
rs796583016 Chr.6: 43182519-43182519 on Build GRCh38
rs989582769 Chr.6: 43182420-43182420 on Build GRCh38
rs927191437 Chr.6: 43182193-43182193 on Build GRCh38
rs924152673 Chr.6: 43182399-43182399 on Build GRCh38
rs968271074 Chr.6: 43182295-43182295 on Build GRCh38
rs961783159 Chr.6: 43182117-43182117 on Build GRCh38
rs941275185 Chr.6: 43182495-43182495 on Build GRCh38
rs1027074734 Chr.6: 43182305-43182305 on Build GRCh38
rs891216777 Chr.6: 43182269-43182269 on Build GRCh38

View Full Product Details