Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43185375-43185874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL9
Entrez Gene ID
23113
Gene Name
cullin 9
Gene Aliases
H7AP1, PARC
UniGene
Hs.485434
Gene Chromosome Location
Chr.6: 43182170-43224587 on Build GRCh38
Transcripts
NM_015089.3 XM_011514422.1 XM_011514423.2 XM_011514424.1 XM_011514425.1 XM_011514426.1 XM_011514428.1 XM_011514430.1 XM_017010589.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1079478 c.622C>T p.L208L Chr.6: 43185482-43185482 on Build GRCh38
3430686 c.739C>A p.H247N Chr.6: 43185599-43185599 on Build GRCh38
1079479 c.643G>A p.E215K Chr.6: 43185503-43185503 on Build GRCh38
4928819 c.668G>A p.R223H Chr.6: 43185528-43185528 on Build GRCh38
6252592 c.655G>T p.D219Y Chr.6: 43185515-43185515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560399100 Chr.6: 43185625-43185625 on Build GRCh38
rs45553043 Chr.6: 43185746-43185746 on Build GRCh38
rs900512535 Chr.6: 43185592-43185592 on Build GRCh38
rs111962935 Chr.6: 43185498-43185498 on Build GRCh38
rs762436503 Chr.6: 43185480-43185480 on Build GRCh38
rs748979879 Chr.6: 43185637-43185637 on Build GRCh38
rs937040305 Chr.6: 43185760-43185760 on Build GRCh38
rs916458610 Chr.6: 43185689-43185689 on Build GRCh38
rs780429384 Chr.6: 43185552-43185552 on Build GRCh38
rs777956158 Chr.6: 43185561-43185561 on Build GRCh38
rs978434790 Chr.6: 43185462-43185462 on Build GRCh38
rs1040444953 Chr.6: 43185776-43185776 on Build GRCh38
rs554818885 Chr.6: 43185491-43185491 on Build GRCh38
rs146200421 Chr.6: 43185502-43185502 on Build GRCh38
rs372291127 Chr.6: 43185445-43185445 on Build GRCh38
rs759810417 Chr.6: 43185655-43185655 on Build GRCh38
rs985015157 Chr.6: 43185728-43185728 on Build GRCh38
rs755109855 Chr.6: 43185435-43185435 on Build GRCh38
rs1047323061 Chr.6: 43185560-43185560 on Build GRCh38
rs746979615 Chr.6: 43185461-43185461 on Build GRCh38
rs369512270 Chr.6: 43185531-43185531 on Build GRCh38
rs992873807 Chr.6: 43185814-43185814 on Build GRCh38
rs201989785 Chr.6: 43185482-43185482 on Build GRCh38
rs766695841 Chr.6: 43185419-43185419 on Build GRCh38
rs745645210 Chr.6: 43185596-43185596 on Build GRCh38
rs907733318 Chr.6: 43185739-43185739 on Build GRCh38
rs761402345 Chr.6: 43185643-43185643 on Build GRCh38
rs753030432 Chr.6: 43185545-43185545 on Build GRCh38
rs372176723 Chr.6: 43185493-43185493 on Build GRCh38
rs779705398 Chr.6: 43185707-43185707 on Build GRCh38
rs775103009 Chr.6: 43185619-43185619 on Build GRCh38
rs960329050 Chr.6: 43185670-43185670 on Build GRCh38
rs886547561 Chr.6: 43185562-43185562 on Build GRCh38
rs754262116 Chr.6: 43185426-43185426 on Build GRCh38
rs901938550 Chr.6: 43185823-43185823 on Build GRCh38
rs757082579 Chr.6: 43185581-43185581 on Build GRCh38
rs552861687 Chr.6: 43185409-43185409 on Build GRCh38
rs139036736 Chr.6: 43185512-43185512 on Build GRCh38
rs537509494 Chr.6: 43185527-43185527 on Build GRCh38
rs769634394 Chr.6: 43185614-43185614 on Build GRCh38
rs768330889 Chr.6: 43185640-43185640 on Build GRCh38
rs770929121 Chr.6: 43185467-43185467 on Build GRCh38
rs775319446 Chr.6: 43185474-43185474 on Build GRCh38
rs149876731 Chr.6: 43185574-43185574 on Build GRCh38
rs781192438 Chr.6: 43185582-43185582 on Build GRCh38
rs376537222 Chr.6: 43185495-43185495 on Build GRCh38
rs745688307 Chr.6: 43185470-43185470 on Build GRCh38
rs771277980 Chr.6: 43185649-43185649 on Build GRCh38
rs777265823 Chr.6: 43185652-43185652 on Build GRCh38
rs758622233 Chr.6: 43185448-43185448 on Build GRCh38
rs200174485 Chr.6: 43185436-43185436 on Build GRCh38
rs1034734470 Chr.6: 43185536-43185536 on Build GRCh38
rs989025063 Chr.6: 43185727-43185727 on Build GRCh38
rs929839079 Chr.6: 43185539-43185539 on Build GRCh38
rs576284245 Chr.6: 43185845-43185845 on Build GRCh38
rs557413964 Chr.6: 43185642-43185642 on Build GRCh38
rs968471606 Chr.6: 43185731-43185731 on Build GRCh38
rs748380483 Chr.6: 43185651-43185656 on Build GRCh38
rs777827349 Chr.6: 43185459-43185459 on Build GRCh38
rs545864146 Chr.6: 43185702-43185702 on Build GRCh38
rs535261094 Chr.6: 43185469-43185469 on Build GRCh38
rs16896326 Chr.6: 43185671-43185671 on Build GRCh38
rs769262435 Chr.6: 43185472-43185472 on Build GRCh38
rs775190952 Chr.6: 43185478-43185478 on Build GRCh38
rs755424124 Chr.6: 43185697-43185697 on Build GRCh38
rs765733524 Chr.6: 43185659-43185659 on Build GRCh38
rs573053136 Chr.6: 43185826-43185826 on Build GRCh38
rs758517410 Chr.6: 43185551-43185551 on Build GRCh38

View Full Product Details