Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70481718-70482235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM135A
Entrez Gene ID
57579
Gene Name
family with sequence similarity 135 member A
Gene Aliases
KIAA1411
UniGene
Hs.211700
Gene Chromosome Location
Chr.6: 70413404-70561174 on Build GRCh38
Transcripts
NM_001105531.2 NM_001162529.1 NM_020819.4 XM_005248743.1 XM_005248744.4 XM_005248745.2 XM_005248748.2 XM_005248749.1 XM_005248751.2 XM_011535993.2 XM_011535994.2 XM_011535995.2 XM_011535996.2 XM_011535997.2 XM_017011124.1 XM_017011125.1 XM_017011126.1 XM_017011127.1 XM_017011128.1 XM_017011129.1 XM_017011130.1 XM_017011131.1 XM_017011132.1 XM_017011133.1 XM_017011134.1 XM_017011135.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5405765 c.799C>T p.P267S Chr.6: 70482130-70482130 on Build GRCh38
1697525 c.721C>T p.H241Y Chr.6: 70482052-70482052 on Build GRCh38
6213646 c.618G>T p.L206L Chr.6: 70482078-70482078 on Build GRCh38
3629410 c.715C>T p.R239C Chr.6: 70482046-70482046 on Build GRCh38
4586893 c.591T>C p.F197F Chr.6: 70482051-70482051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891403415 Chr.6: 70482089-70482089 on Build GRCh38
rs531433480 Chr.6: 70482167-70482167 on Build GRCh38
rs780442601 Chr.6: 70482012-70482012 on Build GRCh38
rs964035472 Chr.6: 70482181-70482181 on Build GRCh38
rs369148670 Chr.6: 70482185-70482185 on Build GRCh38
rs991297308 Chr.6: 70481904-70481904 on Build GRCh38
rs202076605 Chr.6: 70482042-70482042 on Build GRCh38
rs923892012 Chr.6: 70481831-70481831 on Build GRCh38
rs781198008 Chr.6: 70482079-70482079 on Build GRCh38
rs745662509 Chr.6: 70481999-70481999 on Build GRCh38
rs947839717 Chr.6: 70481936-70481936 on Build GRCh38
rs990227131 Chr.6: 70481758-70481758 on Build GRCh38
rs916678245 Chr.6: 70481947-70481947 on Build GRCh38
rs770283959 Chr.6: 70482034-70482034 on Build GRCh38
rs946572055 Chr.6: 70481792-70481792 on Build GRCh38
rs749755255 Chr.6: 70482129-70482129 on Build GRCh38
rs376512666 Chr.6: 70482046-70482046 on Build GRCh38
rs925104365 Chr.6: 70481869-70481869 on Build GRCh38
rs766143123 Chr.6: 70482197-70482197 on Build GRCh38
rs764796031 Chr.6: 70482179-70482179 on Build GRCh38
rs761430248 Chr.6: 70482171-70482171 on Build GRCh38
rs371537662 Chr.6: 70482015-70482015 on Build GRCh38
rs1056080616 Chr.6: 70481852-70481852 on Build GRCh38
rs773646714 Chr.6: 70482035-70482035 on Build GRCh38
rs746441837 Chr.6: 70482176-70482177 on Build GRCh38
rs758209842 Chr.6: 70482010-70482010 on Build GRCh38
rs1017382730 Chr.6: 70481793-70481793 on Build GRCh38
rs770918600 Chr.6: 70482201-70482202 on Build GRCh38
rs773549315 Chr.6: 70482047-70482047 on Build GRCh38
rs780390982 Chr.6: 70481982-70481982 on Build GRCh38
rs771326666 Chr.6: 70482130-70482130 on Build GRCh38
rs535965480 Chr.6: 70481790-70481790 on Build GRCh38
rs778915980 Chr.6: 70481998-70481998 on Build GRCh38
rs1049033239 Chr.6: 70481756-70481756 on Build GRCh38
rs751775449 Chr.6: 70482069-70482069 on Build GRCh38
rs144846534 Chr.6: 70482135-70482135 on Build GRCh38
rs748268144 Chr.6: 70482114-70482114 on Build GRCh38
rs769811664 Chr.6: 70482116-70482116 on Build GRCh38
rs982119234 Chr.6: 70481963-70481963 on Build GRCh38
rs755601168 Chr.6: 70481967-70481967 on Build GRCh38
rs149338749 Chr.6: 70482075-70482075 on Build GRCh38
rs374597098 Chr.6: 70481959-70481959 on Build GRCh38
rs187733653 Chr.6: 70482086-70482086 on Build GRCh38
rs564135475 Chr.6: 70481979-70481979 on Build GRCh38
rs750421968 Chr.6: 70482191-70482191 on Build GRCh38
rs747209939 Chr.6: 70482013-70482013 on Build GRCh38
rs147627157 Chr.6: 70481778-70481778 on Build GRCh38
rs748663798 Chr.6: 70482026-70482026 on Build GRCh38
rs555407216 Chr.6: 70481821-70481821 on Build GRCh38
rs758295772 Chr.6: 70482059-70482059 on Build GRCh38
rs763322121 Chr.6: 70482038-70482038 on Build GRCh38
rs760899836 Chr.6: 70482175-70482175 on Build GRCh38
rs756492484 Chr.6: 70481971-70481972 on Build GRCh38
rs747836556 Chr.6: 70481989-70481989 on Build GRCh38
rs763621317 Chr.6: 70481971-70481971 on Build GRCh38
rs141045723 Chr.6: 70481874-70481874 on Build GRCh38
rs781307585 Chr.6: 70482025-70482025 on Build GRCh38
rs565789540 Chr.6: 70481902-70481902 on Build GRCh38
rs779961700 Chr.6: 70482061-70482061 on Build GRCh38
rs763568587 Chr.6: 70482043-70482043 on Build GRCh38
rs575318205 Chr.6: 70481824-70481824 on Build GRCh38
rs1011543244 Chr.6: 70482194-70482194 on Build GRCh38
rs866347079 Chr.6: 70482212-70482212 on Build GRCh38
rs367701011 Chr.6: 70481982-70481982 on Build GRCh38
rs145224563 Chr.6: 70481745-70481745 on Build GRCh38
rs777282835 Chr.6: 70482141-70482142 on Build GRCh38
rs375876344 Chr.6: 70482056-70482056 on Build GRCh38
rs148577442 Chr.6: 70482139-70482139 on Build GRCh38
rs142170054 Chr.6: 70481939-70481939 on Build GRCh38
rs768358352 Chr.6: 70482145-70482145 on Build GRCh38
rs916233436 Chr.6: 70481857-70481857 on Build GRCh38
rs778276453 Chr.6: 70482117-70482117 on Build GRCh38
rs529045946 Chr.6: 70482052-70482052 on Build GRCh38
rs771688071 Chr.6: 70482105-70482106 on Build GRCh38
rs964318570 Chr.6: 70481807-70481807 on Build GRCh38
rs372130285 Chr.6: 70482195-70482195 on Build GRCh38
rs939240913 Chr.6: 70481848-70481848 on Build GRCh38

View Full Product Details