Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127708343-127708854 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THEMIS
Entrez Gene ID
387357
Gene Name
thymocyte selection associated
Gene Aliases
C6orf190, C6orf207, GASP, SPOT, TSEPA
UniGene
Hs.661756
Gene Chromosome Location
Chr.6: 127697191-127918631 on Build GRCh38
Transcripts
NM_001010923.2 NM_001164685.1 NM_001164687.1 NM_001318531.1 XM_011535813.2 XM_011535814.2 XM_011535816.1 XM_017010849.1

SNP Detail

SNP ID Chromosome Location
rs991447200 Chr.6: 127708502-127708502 on Build GRCh38
rs1037202541 Chr.6: 127708688-127708688 on Build GRCh38
rs943636138 Chr.6: 127708684-127708684 on Build GRCh38
rs944102579 Chr.6: 127708401-127708401 on Build GRCh38
rs546084206 Chr.6: 127708379-127708379 on Build GRCh38
rs991783003 Chr.6: 127708464-127708464 on Build GRCh38
rs1050854532 Chr.6: 127708749-127708749 on Build GRCh38
rs909336036 Chr.6: 127708645-127708645 on Build GRCh38
rs555156757 Chr.6: 127708715-127708715 on Build GRCh38
rs977246125 Chr.6: 127708541-127708541 on Build GRCh38
rs967232477 Chr.6: 127708426-127708426 on Build GRCh38
rs1049218863 Chr.6: 127708567-127708567 on Build GRCh38
rs915918055 Chr.6: 127708488-127708488 on Build GRCh38
rs1021269425 Chr.6: 127708798-127708798 on Build GRCh38
rs968609352 Chr.6: 127708811-127708811 on Build GRCh38
rs183635050 Chr.6: 127708551-127708551 on Build GRCh38
rs563591548 Chr.6: 127708796-127708796 on Build GRCh38
rs1041815785 Chr.6: 127708386-127708386 on Build GRCh38
rs899798479 Chr.6: 127708409-127708409 on Build GRCh38
rs1040830598 Chr.6: 127708781-127708781 on Build GRCh38
rs977264874 Chr.6: 127708440-127708440 on Build GRCh38
rs977275656 Chr.6: 127708814-127708814 on Build GRCh38
rs1015741678 Chr.6: 127708467-127708467 on Build GRCh38
rs1033383073 Chr.6: 127708442-127708442 on Build GRCh38
rs145186546 Chr.6: 127708779-127708779 on Build GRCh38
rs139684148 Chr.6: 127708612-127708612 on Build GRCh38
rs932623706 Chr.6: 127708615-127708615 on Build GRCh38
rs958606390 Chr.6: 127708497-127708497 on Build GRCh38
rs770528829 Chr.6: 127708434-127708434 on Build GRCh38
rs957395652 Chr.6: 127708805-127708805 on Build GRCh38
rs147421458 Chr.6: 127708542-127708542 on Build GRCh38
rs888708335 Chr.6: 127708669-127708669 on Build GRCh38
rs545194223 Chr.6: 127708792-127708792 on Build GRCh38
rs1048669464 Chr.6: 127708652-127708652 on Build GRCh38
rs903292340 Chr.6: 127708393-127708393 on Build GRCh38
rs1041027579 Chr.6: 127708406-127708406 on Build GRCh38
rs761453529 Chr.6: 127708591-127708591 on Build GRCh38
rs1029389483 Chr.6: 127708391-127708391 on Build GRCh38
rs888644169 Chr.6: 127708430-127708430 on Build GRCh38
rs944135209 Chr.6: 127708565-127708565 on Build GRCh38
rs557211832 Chr.6: 127708527-127708527 on Build GRCh38
rs1039670555 Chr.6: 127708706-127708706 on Build GRCh38
rs957304030 Chr.6: 127708461-127708461 on Build GRCh38
rs906665339 Chr.6: 127708695-127708695 on Build GRCh38
rs113549810 Chr.6: 127708528-127708528 on Build GRCh38
rs540711676 Chr.6: 127708741-127708741 on Build GRCh38
rs981507834 Chr.6: 127708501-127708501 on Build GRCh38
rs17054936 Chr.6: 127708776-127708776 on Build GRCh38
rs776324081 Chr.6: 127708492-127708492 on Build GRCh38
rs961806609 Chr.6: 127708485-127708485 on Build GRCh38

View Full Product Details