Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159735877-159736401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOD2
Entrez Gene ID
6648
Gene Name
superoxide dismutase 2, mitochondrial
Gene Aliases
IPO-B, IPOB, MNSOD, MVCD6, Mn-SOD
UniGene
Hs.487046
Gene Chromosome Location
Chr.6: 159679064-159762529 on Build GRCh38
Transcripts
NM_001322817.1 NM_001270531.1 NM_001270532.1 NM_001270533.1 NM_004906.4 NM_152857.2 NM_152858.2 XM_017011514.1 XM_017011516.1

Gene Symbol
WTAP
Entrez Gene ID
9589
Gene Name
Wilms tumor 1 associated protein
Gene Aliases
Mum2
UniGene
Hs.446091
Gene Chromosome Location
Chr.6: 159726680-159756319 on Build GRCh38
Transcripts
NM_001322817.1 NM_001270531.1 NM_001270532.1 NM_001270533.1 NM_004906.4 NM_152857.2 NM_152858.2 XM_017011514.1 XM_017011516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6285094 c.9C>T p.N3N Chr.6: 159736274-159736274 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs974193935 Chr.6: 159736323-159736323 on Build GRCh38
rs116271519 Chr.6: 159736108-159736108 on Build GRCh38
rs560612985 Chr.6: 159735936-159735936 on Build GRCh38
rs780539288 Chr.6: 159735926-159735926 on Build GRCh38
rs367793783 Chr.6: 159736244-159736244 on Build GRCh38
rs372783310 Chr.6: 159736274-159736274 on Build GRCh38
rs753297041 Chr.6: 159736335-159736336 on Build GRCh38
rs927888393 Chr.6: 159736352-159736352 on Build GRCh38
rs895269526 Chr.6: 159735959-159735959 on Build GRCh38
rs895196173 Chr.6: 159736363-159736363 on Build GRCh38
rs749585936 Chr.6: 159736341-159736341 on Build GRCh38
rs1056330952 Chr.6: 159736362-159736362 on Build GRCh38
rs572844538 Chr.6: 159736241-159736241 on Build GRCh38
rs948119360 Chr.6: 159736367-159736367 on Build GRCh38
rs368872375 Chr.6: 159736301-159736301 on Build GRCh38
rs752372925 Chr.6: 159736359-159736360 on Build GRCh38
rs955715352 Chr.6: 159736043-159736043 on Build GRCh38
rs554606642 Chr.6: 159736358-159736358 on Build GRCh38
rs117466922 Chr.6: 159735912-159735912 on Build GRCh38
rs755980403 Chr.6: 159736326-159736326 on Build GRCh38
rs762498568 Chr.6: 159736306-159736306 on Build GRCh38
rs180929656 Chr.6: 159736032-159736032 on Build GRCh38
rs761146958 Chr.6: 159736284-159736284 on Build GRCh38
rs1052165700 Chr.6: 159736169-159736169 on Build GRCh38
rs773549516 Chr.6: 159735952-159735952 on Build GRCh38
rs988385388 Chr.6: 159736363-159736363 on Build GRCh38
rs768851271 Chr.6: 159736231-159736231 on Build GRCh38
rs756050529 Chr.6: 159736245-159736246 on Build GRCh38
rs199792178 Chr.6: 159736166-159736169 on Build GRCh38
rs1043373857 Chr.6: 159736213-159736213 on Build GRCh38
rs974146390 Chr.6: 159736048-159736048 on Build GRCh38
rs987009896 Chr.6: 159736080-159736080 on Build GRCh38
rs927805941 Chr.6: 159736139-159736139 on Build GRCh38
rs939168972 Chr.6: 159736353-159736353 on Build GRCh38
rs920264695 Chr.6: 159736192-159736192 on Build GRCh38
rs534171230 Chr.6: 159736131-159736131 on Build GRCh38
rs769932482 Chr.6: 159736221-159736221 on Build GRCh38
rs73601308 Chr.6: 159736025-159736025 on Build GRCh38
rs751047760 Chr.6: 159736313-159736313 on Build GRCh38
rs886291300 Chr.6: 159735921-159735921 on Build GRCh38
rs558500823 Chr.6: 159736202-159736202 on Build GRCh38
rs920471064 Chr.6: 159736107-159736107 on Build GRCh38
rs185999806 Chr.6: 159735943-159735943 on Build GRCh38
rs747746284 Chr.6: 159736023-159736023 on Build GRCh38
rs566266322 Chr.6: 159736188-159736188 on Build GRCh38
rs750308316 Chr.6: 159736322-159736322 on Build GRCh38
rs757292322 Chr.6: 159736316-159736316 on Build GRCh38
rs369148848 Chr.6: 159736336-159736336 on Build GRCh38
rs538365464 Chr.6: 159736201-159736201 on Build GRCh38
rs1010492661 Chr.6: 159736209-159736209 on Build GRCh38
rs767967294 Chr.6: 159736251-159736251 on Build GRCh38
rs946463755 Chr.6: 159736372-159736373 on Build GRCh38
rs201519653 Chr.6: 159736165-159736167 on Build GRCh38
rs189990304 Chr.6: 159735990-159735990 on Build GRCh38
rs185595587 Chr.6: 159736113-159736113 on Build GRCh38
rs983788210 Chr.6: 159736238-159736238 on Build GRCh38
rs930571032 Chr.6: 159735905-159735905 on Build GRCh38
rs774404692 Chr.6: 159736240-159736240 on Build GRCh38
rs374736441 Chr.6: 159736299-159736299 on Build GRCh38
rs777415373 Chr.6: 159736029-159736029 on Build GRCh38
rs1056941331 Chr.6: 159735928-159735928 on Build GRCh38
rs767517204 Chr.6: 159736220-159736222 on Build GRCh38
rs552628728 Chr.6: 159736186-159736186 on Build GRCh38
rs765895545 Chr.6: 159736315-159736316 on Build GRCh38
rs770696642 Chr.6: 159736149-159736149 on Build GRCh38
rs750355245 Chr.6: 159736246-159736246 on Build GRCh38
rs915905760 Chr.6: 159736328-159736329 on Build GRCh38
rs763710119 Chr.6: 159736308-159736308 on Build GRCh38
rs980695211 Chr.6: 159736348-159736348 on Build GRCh38
rs1022449839 Chr.6: 159735920-159735920 on Build GRCh38
rs920504750 Chr.6: 159735904-159735904 on Build GRCh38
rs775833826 Chr.6: 159736225-159736225 on Build GRCh38
rs781228165 Chr.6: 159736321-159736321 on Build GRCh38
rs779711644 Chr.6: 159736333-159736333 on Build GRCh38
rs1043721638 Chr.6: 159736370-159736370 on Build GRCh38

View Full Product Details