Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44881452-44881959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIR4657
Entrez Gene ID
100616393
Gene Name
microRNA 4657
Gene Aliases
mir-4657
UniGene
Gene Chromosome Location
Chr.7: 44881748-44881800 on Build GRCh38
Transcripts
NR_039801.1 NM_033224.4

Gene Symbol
PURB
Entrez Gene ID
5814
Gene Name
purine rich element binding protein B
Gene Aliases
PURBETA
UniGene
Hs.349150
Gene Chromosome Location
Chr.7: 44876293-44885385 on Build GRCh38
Transcripts
NR_039801.1 NM_033224.4

SNP Detail

SNP ID Chromosome Location
rs532504448 Chr.7: 44881794-44881794 on Build GRCh38
rs932509812 Chr.7: 44881673-44881673 on Build GRCh38
rs913223868 Chr.7: 44881611-44881611 on Build GRCh38
rs747717006 Chr.7: 44881515-44881515 on Build GRCh38
rs943974199 Chr.7: 44881916-44881916 on Build GRCh38
rs529949509 Chr.7: 44881582-44881582 on Build GRCh38
rs944010855 Chr.7: 44881595-44881597 on Build GRCh38
rs1010486931 Chr.7: 44881650-44881650 on Build GRCh38
rs771843278 Chr.7: 44881528-44881528 on Build GRCh38
rs1043146690 Chr.7: 44881617-44881617 on Build GRCh38
rs904658117 Chr.7: 44881667-44881667 on Build GRCh38
rs973002552 Chr.7: 44881494-44881494 on Build GRCh38
rs760485115 Chr.7: 44881631-44881631 on Build GRCh38
rs932035823 Chr.7: 44881842-44881842 on Build GRCh38
rs896814533 Chr.7: 44881728-44881728 on Build GRCh38
rs1043653822 Chr.7: 44881666-44881666 on Build GRCh38
rs1008120388 Chr.7: 44881694-44881694 on Build GRCh38
rs946301610 Chr.7: 44881828-44881828 on Build GRCh38
rs540691497 Chr.7: 44881526-44881526 on Build GRCh38
rs892069121 Chr.7: 44881644-44881644 on Build GRCh38
rs980641572 Chr.7: 44881560-44881560 on Build GRCh38
rs1013288004 Chr.7: 44881711-44881711 on Build GRCh38
rs961624460 Chr.7: 44881730-44881730 on Build GRCh38
rs565614452 Chr.7: 44881506-44881506 on Build GRCh38
rs761886481 Chr.7: 44881830-44881830 on Build GRCh38
rs913228803 Chr.7: 44881820-44881820 on Build GRCh38
rs1026623973 Chr.7: 44881663-44881663 on Build GRCh38
rs922042793 Chr.7: 44881833-44881833 on Build GRCh38
rs1049796549 Chr.7: 44881562-44881562 on Build GRCh38
rs542193223 Chr.7: 44881661-44881661 on Build GRCh38
rs530763385 Chr.7: 44881695-44881695 on Build GRCh38
rs944105820 Chr.7: 44881792-44881792 on Build GRCh38
rs952552977 Chr.7: 44881778-44881778 on Build GRCh38
rs775583663 Chr.7: 44881846-44881846 on Build GRCh38
rs1035790734 Chr.7: 44881765-44881765 on Build GRCh38
rs770777765 Chr.7: 44881692-44881692 on Build GRCh38
rs112908972 Chr.7: 44881788-44881788 on Build GRCh38
rs776335650 Chr.7: 44881763-44881763 on Build GRCh38
rs976758844 Chr.7: 44881810-44881810 on Build GRCh38
rs371906550 Chr.7: 44881627-44881627 on Build GRCh38
rs3832522 Chr.7: 44881639-44881639 on Build GRCh38
rs988784369 Chr.7: 44881590-44881590 on Build GRCh38
rs1055960034 Chr.7: 44881910-44881910 on Build GRCh38
rs184188518 Chr.7: 44881691-44881691 on Build GRCh38
rs765148559 Chr.7: 44881779-44881779 on Build GRCh38
rs397840742 Chr.7: 44881645-44881645 on Build GRCh38
rs181165273 Chr.7: 44881547-44881547 on Build GRCh38
rs968743342 Chr.7: 44881798-44881798 on Build GRCh38
rs759499724 Chr.7: 44881764-44881764 on Build GRCh38
rs559559518 Chr.7: 44881550-44881550 on Build GRCh38
rs777483461 Chr.7: 44881826-44881826 on Build GRCh38
rs772070107 Chr.7: 44881843-44881843 on Build GRCh38
rs189834973 Chr.7: 44881749-44881749 on Build GRCh38
rs947483710 Chr.7: 44881615-44881615 on Build GRCh38
rs972641395 Chr.7: 44881739-44881739 on Build GRCh38
rs1026742965 Chr.7: 44881752-44881752 on Build GRCh38
rs925143923 Chr.7: 44881643-44881643 on Build GRCh38
rs547482196 Chr.7: 44881860-44881860 on Build GRCh38
rs913276144 Chr.7: 44881796-44881796 on Build GRCh38
rs1033405354 Chr.7: 44881685-44881685 on Build GRCh38
rs928890734 Chr.7: 44881564-44881564 on Build GRCh38
rs973830976 Chr.7: 44881480-44881480 on Build GRCh38
rs1036721399 Chr.7: 44881701-44881701 on Build GRCh38

View Full Product Details