Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 156962916-156963411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOM1
Entrez Gene ID
64434
Gene Name
nucleolar protein with MIF4G domain 1
Gene Aliases
C7orf3, PPP1R113, SGD1
UniGene
Hs.15825
Gene Chromosome Location
Chr.7: 156949723-156973182 on Build GRCh38
Transcripts
NM_138400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5601929 c.1858C>T p.P620S Chr.7: 156963122-156963122 on Build GRCh38
7379980 c.1823G>A p.R608H Chr.7: 156963087-156963087 on Build GRCh38
4834416 c.1815G>C p.Q605H Chr.7: 156963079-156963079 on Build GRCh38
4434907 c.1854G>A p.G618G Chr.7: 156963118-156963118 on Build GRCh38
7379981 c.1904T>C p.V635A Chr.7: 156963168-156963168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61741545 Chr.7: 156963037-156963037 on Build GRCh38
rs758419482 Chr.7: 156963185-156963185 on Build GRCh38
rs1021368811 Chr.7: 156963112-156963112 on Build GRCh38
rs369516563 Chr.7: 156963197-156963197 on Build GRCh38
rs759006741 Chr.7: 156963262-156963262 on Build GRCh38
rs560499011 Chr.7: 156963219-156963220 on Build GRCh38
rs923936540 Chr.7: 156963214-156963214 on Build GRCh38
rs772236340 Chr.7: 156963050-156963050 on Build GRCh38
rs900246274 Chr.7: 156963114-156963114 on Build GRCh38
rs781422578 Chr.7: 156963281-156963281 on Build GRCh38
rs564466837 Chr.7: 156963196-156963196 on Build GRCh38
rs527932395 Chr.7: 156963321-156963321 on Build GRCh38
rs745608759 Chr.7: 156963288-156963288 on Build GRCh38
rs201678028 Chr.7: 156963139-156963139 on Build GRCh38
rs368526716 Chr.7: 156962958-156962958 on Build GRCh38
rs764514798 Chr.7: 156963322-156963327 on Build GRCh38
rs374092429 Chr.7: 156963021-156963021 on Build GRCh38
rs925887909 Chr.7: 156963066-156963066 on Build GRCh38
rs200453645 Chr.7: 156962975-156962975 on Build GRCh38
rs1038823702 Chr.7: 156963299-156963299 on Build GRCh38
rs1049221063 Chr.7: 156963336-156963336 on Build GRCh38
rs62492704 Chr.7: 156963211-156963211 on Build GRCh38
rs774647900 Chr.7: 156963012-156963012 on Build GRCh38
rs778368958 Chr.7: 156963024-156963024 on Build GRCh38
rs759285038 Chr.7: 156963381-156963381 on Build GRCh38
rs1035186898 Chr.7: 156963154-156963154 on Build GRCh38
rs747223828 Chr.7: 156963192-156963192 on Build GRCh38
rs1023766419 Chr.7: 156963199-156963199 on Build GRCh38
rs149569042 Chr.7: 156963047-156963047 on Build GRCh38
rs765715470 Chr.7: 156963244-156963244 on Build GRCh38
rs764346285 Chr.7: 156963002-156963002 on Build GRCh38
rs867419065 Chr.7: 156963135-156963135 on Build GRCh38
rs758053177 Chr.7: 156963029-156963029 on Build GRCh38
rs775305048 Chr.7: 156963215-156963215 on Build GRCh38
rs758403704 Chr.7: 156963323-156963323 on Build GRCh38
rs1031421261 Chr.7: 156963345-156963345 on Build GRCh38
rs900830913 Chr.7: 156963350-156963350 on Build GRCh38
rs201968252 Chr.7: 156963123-156963123 on Build GRCh38
rs1028363306 Chr.7: 156962954-156962954 on Build GRCh38
rs749324849 Chr.7: 156963045-156963045 on Build GRCh38
rs547021901 Chr.7: 156963351-156963351 on Build GRCh38
rs779031788 Chr.7: 156963107-156963107 on Build GRCh38
rs138999947 Chr.7: 156963174-156963174 on Build GRCh38
rs776177762 Chr.7: 156963217-156963217 on Build GRCh38
rs200947524 Chr.7: 156963110-156963110 on Build GRCh38
rs201457653 Chr.7: 156963092-156963092 on Build GRCh38
rs979871616 Chr.7: 156963205-156963205 on Build GRCh38
rs527599221 Chr.7: 156963373-156963373 on Build GRCh38
rs757422016 Chr.7: 156963278-156963278 on Build GRCh38
rs115075018 Chr.7: 156963131-156963131 on Build GRCh38
rs777795377 Chr.7: 156963265-156963265 on Build GRCh38
rs764798949 Chr.7: 156963073-156963073 on Build GRCh38
rs561047627 Chr.7: 156963082-156963082 on Build GRCh38
rs199664861 Chr.7: 156963067-156963067 on Build GRCh38
rs773014326 Chr.7: 156963059-156963059 on Build GRCh38
rs552120489 Chr.7: 156962970-156962970 on Build GRCh38
rs766729357 Chr.7: 156963220-156963220 on Build GRCh38
rs769464702 Chr.7: 156963303-156963303 on Build GRCh38
rs186610955 Chr.7: 156963019-156963019 on Build GRCh38
rs142522087 Chr.7: 156963017-156963017 on Build GRCh38
rs141585452 Chr.7: 156963147-156963147 on Build GRCh38
rs757037589 Chr.7: 156963140-156963140 on Build GRCh38
rs930380426 Chr.7: 156963333-156963333 on Build GRCh38
rs141690153 Chr.7: 156963036-156963036 on Build GRCh38
rs1035155875 Chr.7: 156962995-156962995 on Build GRCh38
rs754218086 Chr.7: 156963232-156963232 on Build GRCh38
rs753784681 Chr.7: 156963071-156963071 on Build GRCh38
rs770695085 Chr.7: 156963124-156963124 on Build GRCh38
rs775424938 Chr.7: 156963306-156963306 on Build GRCh38
rs11549567 Chr.7: 156963172-156963172 on Build GRCh38
rs776386954 Chr.7: 156963198-156963198 on Build GRCh38
rs921705918 Chr.7: 156963307-156963307 on Build GRCh38
rs780534842 Chr.7: 156963148-156963148 on Build GRCh38
rs755306008 Chr.7: 156963238-156963238 on Build GRCh38
rs998975611 Chr.7: 156963334-156963334 on Build GRCh38
rs761538430 Chr.7: 156963218-156963218 on Build GRCh38
rs777311659 Chr.7: 156963086-156963086 on Build GRCh38
rs752201654 Chr.7: 156963132-156963132 on Build GRCh38
rs749808765 Chr.7: 156963094-156963094 on Build GRCh38
rs375176749 Chr.7: 156963221-156963221 on Build GRCh38
rs139894978 Chr.7: 156963041-156963041 on Build GRCh38
rs749149226 Chr.7: 156963330-156963330 on Build GRCh38
rs751244384 Chr.7: 156963138-156963138 on Build GRCh38
rs778275600 Chr.7: 156963122-156963122 on Build GRCh38
rs367915170 Chr.7: 156963133-156963133 on Build GRCh38
rs138677884 Chr.7: 156963137-156963137 on Build GRCh38
rs552456263 Chr.7: 156963175-156963175 on Build GRCh38
rs759523570 Chr.7: 156963072-156963072 on Build GRCh38
rs528469274 Chr.7: 156963268-156963268 on Build GRCh38
rs762400061 Chr.7: 156963134-156963134 on Build GRCh38
rs1042886268 Chr.7: 156963324-156963324 on Build GRCh38
rs563618308 Chr.7: 156962944-156962944 on Build GRCh38
rs927916370 Chr.7: 156963186-156963186 on Build GRCh38
rs968142849 Chr.7: 156963202-156963202 on Build GRCh38
rs777885380 Chr.7: 156963188-156963188 on Build GRCh38
rs754308066 Chr.7: 156963152-156963152 on Build GRCh38
rs758466367 Chr.7: 156962978-156962978 on Build GRCh38
rs548419711 Chr.7: 156962943-156962943 on Build GRCh38
rs886064191 Chr.7: 156963363-156963363 on Build GRCh38
rs147902598 Chr.7: 156963027-156963027 on Build GRCh38
rs1012415413 Chr.7: 156963193-156963193 on Build GRCh38
rs761999667 Chr.7: 156963015-156963015 on Build GRCh38
rs147451571 Chr.7: 156962945-156962945 on Build GRCh38
rs138117452 Chr.7: 156963044-156963044 on Build GRCh38
rs764951294 Chr.7: 156963214-156963214 on Build GRCh38
rs61738590 Chr.7: 156963168-156963168 on Build GRCh38
rs146828480 Chr.7: 156963020-156963020 on Build GRCh38
rs753129113 Chr.7: 156963249-156963249 on Build GRCh38
rs763158603 Chr.7: 156962998-156962998 on Build GRCh38
rs61740219 Chr.7: 156963087-156963087 on Build GRCh38
rs938564839 Chr.7: 156963061-156963061 on Build GRCh38
rs563556242 Chr.7: 156963165-156963165 on Build GRCh38
rs557224611 Chr.7: 156963046-156963046 on Build GRCh38
rs761930230 Chr.7: 156963068-156963068 on Build GRCh38
rs535072199 Chr.7: 156963018-156963018 on Build GRCh38
rs950977728 Chr.7: 156962977-156962977 on Build GRCh38
rs374913089 Chr.7: 156963106-156963106 on Build GRCh38
rs114948943 Chr.7: 156963130-156963130 on Build GRCh38
rs776917462 Chr.7: 156962957-156962957 on Build GRCh38
rs766234962 Chr.7: 156963069-156963069 on Build GRCh38
rs775687759 Chr.7: 156962997-156962997 on Build GRCh38
rs759309482 Chr.7: 156963211-156963212 on Build GRCh38
rs745671452 Chr.7: 156963206-156963206 on Build GRCh38
rs143586682 Chr.7: 156963081-156963081 on Build GRCh38

View Full Product Details