Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143575213-143575705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAPRT
Entrez Gene ID
93100
Gene Name
nicotinate phosphoribosyltransferase
Gene Aliases
NAPRT1, PP3856
UniGene
Hs.493164
Gene Chromosome Location
Chr.8: 143574661-143579694 on Build GRCh38
Transcripts
NM_001286829.1 NM_145201.5 XM_017013975.1 XM_017013976.1 XM_017013977.1 XM_017013978.1 XM_017013979.1 XM_017013980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3834185 c.1174G>T p.G392C Chr.8: 143575636-143575636 on Build GRCh38
4162740 c.1284C>T p.G428G Chr.8: 143575430-143575430 on Build GRCh38
3698692 c.1282C>T p.L428L Chr.8: 143575283-143575283 on Build GRCh38
3646057 c.1211C>T p.P404L Chr.8: 143575503-143575503 on Build GRCh38
6996685 c.1351C>T p.R451W Chr.8: 143575214-143575214 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147154210 Chr.8: 143575467-143575467 on Build GRCh38
rs539726091 Chr.8: 143575423-143575423 on Build GRCh38
rs1010885698 Chr.8: 143575384-143575384 on Build GRCh38
rs748986138 Chr.8: 143575429-143575429 on Build GRCh38
rs372850635 Chr.8: 143575611-143575611 on Build GRCh38
rs781005385 Chr.8: 143575418-143575418 on Build GRCh38
rs773613350 Chr.8: 143575441-143575441 on Build GRCh38
rs201394698 Chr.8: 143575392-143575392 on Build GRCh38
rs778354031 Chr.8: 143575539-143575539 on Build GRCh38
rs751269568 Chr.8: 143575654-143575654 on Build GRCh38
rs962746019 Chr.8: 143575513-143575513 on Build GRCh38
rs769294649 Chr.8: 143575376-143575376 on Build GRCh38
rs746430119 Chr.8: 143575610-143575610 on Build GRCh38
rs781211163 Chr.8: 143575253-143575253 on Build GRCh38
rs772353206 Chr.8: 143575500-143575500 on Build GRCh38
rs879023271 Chr.8: 143575251-143575251 on Build GRCh38
rs369309540 Chr.8: 143575608-143575608 on Build GRCh38
rs75643271 Chr.8: 143575242-143575242 on Build GRCh38
rs759293166 Chr.8: 143575322-143575322 on Build GRCh38
rs375754313 Chr.8: 143575338-143575338 on Build GRCh38
rs369842187 Chr.8: 143575509-143575509 on Build GRCh38
rs968345348 Chr.8: 143575399-143575399 on Build GRCh38
rs774367382 Chr.8: 143575570-143575570 on Build GRCh38
rs374669823 Chr.8: 143575394-143575394 on Build GRCh38
rs760084393 Chr.8: 143575556-143575556 on Build GRCh38
rs752094147 Chr.8: 143575344-143575344 on Build GRCh38
rs141565399 Chr.8: 143575310-143575310 on Build GRCh38
rs764371052 Chr.8: 143575569-143575569 on Build GRCh38
rs757781315 Chr.8: 143575603-143575603 on Build GRCh38
rs563969288 Chr.8: 143575674-143575674 on Build GRCh38
rs147018637 Chr.8: 143575375-143575375 on Build GRCh38
rs557170563 Chr.8: 143575446-143575446 on Build GRCh38
rs746797791 Chr.8: 143575414-143575414 on Build GRCh38
rs777770319 Chr.8: 143575412-143575412 on Build GRCh38
rs768557276 Chr.8: 143575619-143575619 on Build GRCh38
rs754505066 Chr.8: 143575538-143575538 on Build GRCh38
rs761203852 Chr.8: 143575444-143575444 on Build GRCh38
rs749389473 Chr.8: 143575618-143575618 on Build GRCh38
rs777300069 Chr.8: 143575554-143575554 on Build GRCh38
rs760638287 Chr.8: 143575294-143575294 on Build GRCh38
rs939145799 Chr.8: 143575542-143575542 on Build GRCh38
rs750695593 Chr.8: 143575628-143575628 on Build GRCh38
rs754142661 Chr.8: 143575404-143575404 on Build GRCh38
rs144545750 Chr.8: 143575488-143575488 on Build GRCh38
rs139521261 Chr.8: 143575483-143575483 on Build GRCh38
rs373120622 Chr.8: 143575562-143575562 on Build GRCh38
rs757727442 Chr.8: 143575349-143575349 on Build GRCh38
rs568720796 Chr.8: 143575450-143575450 on Build GRCh38
rs778068428 Chr.8: 143575546-143575546 on Build GRCh38
rs763144993 Chr.8: 143575566-143575566 on Build GRCh38
rs760529384 Chr.8: 143575579-143575579 on Build GRCh38
rs375661749 Chr.8: 143575370-143575370 on Build GRCh38
rs756422393 Chr.8: 143575357-143575357 on Build GRCh38
rs373124776 Chr.8: 143575501-143575501 on Build GRCh38
rs765359890 Chr.8: 143575510-143575510 on Build GRCh38
rs929856956 Chr.8: 143575436-143575436 on Build GRCh38
rs757351270 Chr.8: 143575248-143575248 on Build GRCh38
rs111956857 Chr.8: 143575371-143575371 on Build GRCh38
rs745774193 Chr.8: 143575422-143575422 on Build GRCh38
rs574749028 Chr.8: 143575607-143575607 on Build GRCh38
rs899043890 Chr.8: 143575380-143575380 on Build GRCh38
rs746140281 Chr.8: 143575258-143575258 on Build GRCh38
rs779967488 Chr.8: 143575532-143575532 on Build GRCh38
rs768904692 Chr.8: 143575478-143575478 on Build GRCh38
rs201445971 Chr.8: 143575540-143575540 on Build GRCh38
rs774760060 Chr.8: 143575434-143575434 on Build GRCh38
rs757949301 Chr.8: 143575651-143575651 on Build GRCh38
rs750216204 Chr.8: 143575517-143575517 on Build GRCh38
rs140376010 Chr.8: 143575278-143575278 on Build GRCh38
rs770980537 Chr.8: 143575373-143575373 on Build GRCh38
rs775692079 Chr.8: 143575565-143575565 on Build GRCh38
rs771364854 Chr.8: 143575547-143575547 on Build GRCh38
rs1010487789 Chr.8: 143575477-143575477 on Build GRCh38
rs918585463 Chr.8: 143575525-143575525 on Build GRCh38
rs1021828296 Chr.8: 143575567-143575567 on Build GRCh38
rs779119678 Chr.8: 143575561-143575561 on Build GRCh38
rs752236565 Chr.8: 143575602-143575602 on Build GRCh38
rs371481966 Chr.8: 143575424-143575424 on Build GRCh38
rs756070810 Chr.8: 143575526-143575526 on Build GRCh38
rs774652111 Chr.8: 143575623-143575623 on Build GRCh38
rs771922687 Chr.8: 143575270-143575270 on Build GRCh38
rs369189989 Chr.8: 143575633-143575633 on Build GRCh38
rs901172906 Chr.8: 143575555-143575555 on Build GRCh38
rs369901658 Chr.8: 143575481-143575481 on Build GRCh38
rs371127967 Chr.8: 143575356-143575356 on Build GRCh38
rs753804623 Chr.8: 143575590-143575590 on Build GRCh38
rs375185490 Chr.8: 143575463-143575463 on Build GRCh38
rs765166125 Chr.8: 143575645-143575645 on Build GRCh38
rs143442304 Chr.8: 143575466-143575466 on Build GRCh38
rs777495975 Chr.8: 143575604-143575604 on Build GRCh38
rs541112174 Chr.8: 143575535-143575535 on Build GRCh38
rs752007321 Chr.8: 143575244-143575244 on Build GRCh38
rs756035328 Chr.8: 143575458-143575465 on Build GRCh38
rs368093504 Chr.8: 143575417-143575417 on Build GRCh38
rs761133109 Chr.8: 143575502-143575502 on Build GRCh38
rs149282363 Chr.8: 143575487-143575487 on Build GRCh38
rs754877551 Chr.8: 143575595-143575595 on Build GRCh38
rs761982396 Chr.8: 143575630-143575630 on Build GRCh38
rs746233783 Chr.8: 143575679-143575679 on Build GRCh38
rs148462517 Chr.8: 143575516-143575516 on Build GRCh38
rs371882017 Chr.8: 143575391-143575391 on Build GRCh38
rs929903877 Chr.8: 143575306-143575306 on Build GRCh38
rs75601593 Chr.8: 143575355-143575355 on Build GRCh38
rs770037335 Chr.8: 143575561-143575561 on Build GRCh38
rs770594113 Chr.8: 143575300-143575300 on Build GRCh38
rs552313427 Chr.8: 143575341-143575341 on Build GRCh38
rs764774540 Chr.8: 143575597-143575597 on Build GRCh38
rs1006475819 Chr.8: 143575587-143575587 on Build GRCh38
rs149178575 Chr.8: 143575235-143575235 on Build GRCh38
rs758289653 Chr.8: 143575367-143575367 on Build GRCh38
rs769884687 Chr.8: 143575262-143575262 on Build GRCh38
rs147123886 Chr.8: 143575330-143575330 on Build GRCh38
rs759527233 Chr.8: 143575640-143575640 on Build GRCh38
rs542475839 Chr.8: 143575266-143575266 on Build GRCh38
rs752520361 Chr.8: 143575649-143575649 on Build GRCh38
rs376795388 Chr.8: 143575508-143575508 on Build GRCh38
rs201086959 Chr.8: 143575237-143575237 on Build GRCh38
rs73715689 Chr.8: 143575580-143575580 on Build GRCh38
rs1017937432 Chr.8: 143575406-143575406 on Build GRCh38
rs780143799 Chr.8: 143575358-143575358 on Build GRCh38
rs781729373 Chr.8: 143575675-143575675 on Build GRCh38
rs762362757 Chr.8: 143575398-143575398 on Build GRCh38
rs771732028 Chr.8: 143575541-143575541 on Build GRCh38
rs748150406 Chr.8: 143575575-143575579 on Build GRCh38
rs775497093 Chr.8: 143575614-143575614 on Build GRCh38
rs768085460 Chr.8: 143575486-143575486 on Build GRCh38
rs763472448 Chr.8: 143575653-143575653 on Build GRCh38
rs2290416 Chr.8: 143575430-143575430 on Build GRCh38
rs568011940 Chr.8: 143575403-143575403 on Build GRCh38
rs770012110 Chr.8: 143575683-143575683 on Build GRCh38
rs894926770 Chr.8: 143575557-143575557 on Build GRCh38
rs9771743 Chr.8: 143575631-143575631 on Build GRCh38
rs199855628 Chr.8: 143575514-143575514 on Build GRCh38
rs750305065 Chr.8: 143575427-143575427 on Build GRCh38
rs767903032 Chr.8: 143575402-143575402 on Build GRCh38
rs762166562 Chr.8: 143575440-143575440 on Build GRCh38

View Full Product Details