Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 34557334-34557864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTFR
Entrez Gene ID
1271
Gene Name
ciliary neurotrophic factor receptor
Gene Aliases
-
UniGene
Hs.129966
Gene Chromosome Location
Chr.9: 34551432-34590386 on Build GRCh38
Transcripts
NM_001207011.1 NM_001842.4 NM_147164.2 XM_017014260.1 XM_017014261.1 XM_017014262.1 XM_017014263.1 XM_017014264.1 XM_017014265.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4373939 c.510C>T p.H170H Chr.9: 34557620-34557620 on Build GRCh38
5814016 c.539C>G p.S180C Chr.9: 34557591-34557591 on Build GRCh38
3926730 c.487C>T p.R163C Chr.9: 34557643-34557643 on Build GRCh38
5033401 c.438-8C>G p.? Chr.9: 34557700-34557700 on Build GRCh38
2773342 c.510C>T p.H170H Chr.9: 34557620-34557620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377365010 Chr.9: 34557717-34557717 on Build GRCh38
rs1034648497 Chr.9: 34557511-34557511 on Build GRCh38
rs143854114 Chr.9: 34557685-34557685 on Build GRCh38
rs1049330885 Chr.9: 34557381-34557382 on Build GRCh38
rs111317113 Chr.9: 34557514-34557514 on Build GRCh38
rs1002312110 Chr.9: 34557643-34557643 on Build GRCh38
rs763191215 Chr.9: 34557630-34557630 on Build GRCh38
rs933795511 Chr.9: 34557402-34557402 on Build GRCh38
rs775819617 Chr.9: 34557720-34557720 on Build GRCh38
rs761863333 Chr.9: 34557645-34557645 on Build GRCh38
rs771336868 Chr.9: 34557573-34557573 on Build GRCh38
rs754474196 Chr.9: 34557561-34557561 on Build GRCh38
rs747612441 Chr.9: 34557695-34557695 on Build GRCh38
rs976089434 Chr.9: 34557366-34557366 on Build GRCh38
rs781490668 Chr.9: 34557713-34557713 on Build GRCh38
rs778275446 Chr.9: 34557374-34557374 on Build GRCh38
rs757157223 Chr.9: 34557538-34557538 on Build GRCh38
rs781380441 Chr.9: 34557445-34557445 on Build GRCh38
rs779508397 Chr.9: 34557557-34557557 on Build GRCh38
rs574206625 Chr.9: 34557845-34557845 on Build GRCh38
rs770047753 Chr.9: 34557718-34557718 on Build GRCh38
rs757841005 Chr.9: 34557415-34557415 on Build GRCh38
rs764235933 Chr.9: 34557634-34557634 on Build GRCh38
rs770093490 Chr.9: 34557647-34557647 on Build GRCh38
rs910661316 Chr.9: 34557356-34557356 on Build GRCh38
rs966138204 Chr.9: 34557675-34557675 on Build GRCh38
rs770066263 Chr.9: 34557622-34557622 on Build GRCh38
rs541819919 Chr.9: 34557800-34557800 on Build GRCh38
rs563543000 Chr.9: 34557378-34557378 on Build GRCh38
rs976514718 Chr.9: 34557777-34557777 on Build GRCh38
rs565651839 Chr.9: 34557442-34557442 on Build GRCh38
rs746150154 Chr.9: 34557529-34557529 on Build GRCh38
rs767505611 Chr.9: 34557828-34557828 on Build GRCh38
rs771283513 Chr.9: 34557483-34557483 on Build GRCh38
rs199523536 Chr.9: 34557703-34557703 on Build GRCh38
rs370833574 Chr.9: 34557817-34557817 on Build GRCh38
rs919261934 Chr.9: 34557369-34557369 on Build GRCh38
rs796507966 Chr.9: 34557612-34557612 on Build GRCh38
rs929341175 Chr.9: 34557721-34557722 on Build GRCh38
rs1006471401 Chr.9: 34557704-34557704 on Build GRCh38
rs753541676 Chr.9: 34557844-34557844 on Build GRCh38
rs187199910 Chr.9: 34557489-34557489 on Build GRCh38
rs374204789 Chr.9: 34557652-34557652 on Build GRCh38
rs373812906 Chr.9: 34557392-34557392 on Build GRCh38
rs759785573 Chr.9: 34557503-34557503 on Build GRCh38
rs375484793 Chr.9: 34557551-34557551 on Build GRCh38
rs761089076 Chr.9: 34557477-34557477 on Build GRCh38
rs768874497 Chr.9: 34557715-34557715 on Build GRCh38
rs574117535 Chr.9: 34557793-34557793 on Build GRCh38
rs986011555 Chr.9: 34557589-34557589 on Build GRCh38
rs775558575 Chr.9: 34557625-34557625 on Build GRCh38
rs1052287850 Chr.9: 34557411-34557411 on Build GRCh38
rs774479233 Chr.9: 34557642-34557642 on Build GRCh38
rs764105643 Chr.9: 34557458-34557458 on Build GRCh38
rs746284896 Chr.9: 34557515-34557515 on Build GRCh38
rs1056241226 Chr.9: 34557831-34557831 on Build GRCh38
rs139835086 Chr.9: 34557653-34557653 on Build GRCh38
rs765334248 Chr.9: 34557509-34557509 on Build GRCh38
rs747487719 Chr.9: 34557572-34557572 on Build GRCh38
rs768950184 Chr.9: 34557727-34557727 on Build GRCh38
rs777081784 Chr.9: 34557576-34557576 on Build GRCh38
rs778321460 Chr.9: 34557571-34557571 on Build GRCh38
rs760588639 Chr.9: 34557841-34557841 on Build GRCh38
rs749497180 Chr.9: 34557724-34557724 on Build GRCh38
rs764182539 Chr.9: 34557519-34557519 on Build GRCh38
rs985427203 Chr.9: 34557730-34557730 on Build GRCh38
rs766059728 Chr.9: 34557843-34557843 on Build GRCh38
rs746147982 Chr.9: 34557609-34557609 on Build GRCh38
rs369998155 Chr.9: 34557361-34557361 on Build GRCh38
rs150628275 Chr.9: 34557686-34557686 on Build GRCh38
rs781018193 Chr.9: 34557539-34557539 on Build GRCh38
rs956123138 Chr.9: 34557802-34557802 on Build GRCh38
rs909495208 Chr.9: 34557742-34557742 on Build GRCh38
rs549094484 Chr.9: 34557423-34557423 on Build GRCh38
rs753488328 Chr.9: 34557680-34557680 on Build GRCh38
rs910556363 Chr.9: 34557358-34557358 on Build GRCh38
rs371519060 Chr.9: 34557522-34557522 on Build GRCh38
rs373944890 Chr.9: 34557833-34557833 on Build GRCh38
rs752019553 Chr.9: 34557389-34557389 on Build GRCh38
rs748812068 Chr.9: 34557558-34557558 on Build GRCh38
rs766115164 Chr.9: 34557658-34557658 on Build GRCh38
rs116401434 Chr.9: 34557689-34557689 on Build GRCh38
rs377664646 Chr.9: 34557750-34557750 on Build GRCh38
rs913793164 Chr.9: 34557479-34557479 on Build GRCh38
rs923328999 Chr.9: 34557796-34557796 on Build GRCh38
rs761918479 Chr.9: 34557819-34557819 on Build GRCh38

View Full Product Details