Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120603806-120604303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MEGF9
Entrez Gene ID
1955
Gene Name
multiple EGF like domains 9
Gene Aliases
EGFL5
UniGene
Hs.744903
Gene Chromosome Location
Chr.9: 120600918-120714487 on Build GRCh38
Transcripts
NM_001080497.2

SNP Detail

SNP ID Chromosome Location
rs539783414 Chr.9: 120604220-120604220 on Build GRCh38
rs912564467 Chr.9: 120604226-120604226 on Build GRCh38
rs942743700 Chr.9: 120604239-120604239 on Build GRCh38
rs1009718311 Chr.9: 120604008-120604008 on Build GRCh38
rs746123250 Chr.9: 120604080-120604080 on Build GRCh38
rs909188819 Chr.9: 120604230-120604230 on Build GRCh38
rs575594549 Chr.9: 120603897-120603897 on Build GRCh38
rs546753485 Chr.9: 120603959-120603959 on Build GRCh38
rs933250488 Chr.9: 120604173-120604173 on Build GRCh38
rs1013229431 Chr.9: 120603841-120603841 on Build GRCh38
rs369743039 Chr.9: 120604092-120604092 on Build GRCh38
rs563165990 Chr.9: 120603854-120603854 on Build GRCh38
rs751239959 Chr.9: 120603853-120603854 on Build GRCh38
rs115584562 Chr.9: 120603852-120603852 on Build GRCh38
rs940817727 Chr.9: 120604268-120604268 on Build GRCh38
rs188901083 Chr.9: 120603948-120603948 on Build GRCh38
rs879442409 Chr.9: 120604281-120604281 on Build GRCh38
rs577161670 Chr.9: 120604269-120604269 on Build GRCh38
rs552466769 Chr.9: 120603916-120603916 on Build GRCh38
rs972532115 Chr.9: 120604271-120604271 on Build GRCh38
rs765228669 Chr.9: 120604110-120604110 on Build GRCh38
rs187100079 Chr.9: 120604221-120604221 on Build GRCh38
rs980554788 Chr.9: 120604061-120604061 on Build GRCh38
rs778775937 Chr.9: 120604250-120604252 on Build GRCh38
rs75697542 Chr.9: 120603892-120603892 on Build GRCh38
rs182587234 Chr.9: 120604045-120604045 on Build GRCh38
rs779214807 Chr.9: 120603946-120603946 on Build GRCh38
rs760865404 Chr.9: 120603870-120603870 on Build GRCh38
rs551029987 Chr.9: 120604155-120604155 on Build GRCh38
rs956505184 Chr.9: 120603966-120603966 on Build GRCh38
rs564412546 Chr.9: 120603922-120603922 on Build GRCh38
rs965184873 Chr.9: 120604140-120604140 on Build GRCh38
rs1013806947 Chr.9: 120603865-120603865 on Build GRCh38
rs117272780 Chr.9: 120604126-120604126 on Build GRCh38
rs78550264 Chr.9: 120604195-120604195 on Build GRCh38
rs919204582 Chr.9: 120604160-120604160 on Build GRCh38
rs936885968 Chr.9: 120604276-120604276 on Build GRCh38
rs1032420643 Chr.9: 120603954-120603954 on Build GRCh38
rs963281982 Chr.9: 120604131-120604131 on Build GRCh38

View Full Product Details