Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124312103-124312603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK6
Entrez Gene ID
10783
Gene Name
NIMA related kinase 6
Gene Aliases
SID6-1512
UniGene
Hs.197071
Gene Chromosome Location
Chr.9: 124257606-124352442 on Build GRCh38
Transcripts
NM_001145001.2 NM_001166167.1 NM_001166168.1 NM_001166169.1 NM_001166170.1 NM_001166171.1 NM_014397.5 XM_005251664.1 XM_006716936.2 XM_017014217.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6205198 c.193-9C>T p.? Chr.9: 124312500-124312500 on Build GRCh38
7100616 c.191G>T p.R64L Chr.9: 124312534-124312534 on Build GRCh38
5738729 c.134G>A p.G45D Chr.9: 124312573-124312573 on Build GRCh38
3699267 c.102G>A p.S34S Chr.9: 124312541-124312541 on Build GRCh38
5738728 c.230G>A p.G77D Chr.9: 124312573-124312573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376555431 Chr.9: 124312349-124312349 on Build GRCh38
rs569137318 Chr.9: 124312185-124312185 on Build GRCh38
rs970901819 Chr.9: 124312356-124312356 on Build GRCh38
rs781287662 Chr.9: 124312559-124312559 on Build GRCh38
rs947172482 Chr.9: 124312420-124312420 on Build GRCh38
rs79935846 Chr.9: 124312225-124312225 on Build GRCh38
rs754993371 Chr.9: 124312547-124312547 on Build GRCh38
rs981870873 Chr.9: 124312172-124312172 on Build GRCh38
rs923490291 Chr.9: 124312358-124312358 on Build GRCh38
rs774694008 Chr.9: 124312522-124312522 on Build GRCh38
rs750746819 Chr.9: 124312533-124312533 on Build GRCh38
rs374325246 Chr.9: 124312447-124312447 on Build GRCh38
rs773285063 Chr.9: 124312348-124312348 on Build GRCh38
rs575505766 Chr.9: 124312491-124312491 on Build GRCh38
rs370891224 Chr.9: 124312247-124312247 on Build GRCh38
rs1039662090 Chr.9: 124312227-124312228 on Build GRCh38
rs955752680 Chr.9: 124312139-124312139 on Build GRCh38
rs1042237008 Chr.9: 124312237-124312237 on Build GRCh38
rs148262318 Chr.9: 124312560-124312560 on Build GRCh38
rs199605725 Chr.9: 124312502-124312502 on Build GRCh38
rs191128530 Chr.9: 124312261-124312261 on Build GRCh38
rs766708819 Chr.9: 124312541-124312541 on Build GRCh38
rs909191659 Chr.9: 124312206-124312206 on Build GRCh38
rs919420913 Chr.9: 124312142-124312142 on Build GRCh38
rs1011169725 Chr.9: 124312159-124312159 on Build GRCh38
rs748026924 Chr.9: 124312495-124312495 on Build GRCh38
rs768983613 Chr.9: 124312173-124312173 on Build GRCh38
rs905538342 Chr.9: 124312450-124312450 on Build GRCh38
rs758964365 Chr.9: 124312518-124312518 on Build GRCh38
rs763278157 Chr.9: 124312540-124312540 on Build GRCh38
rs745556761 Chr.9: 124312131-124312131 on Build GRCh38
rs902014370 Chr.9: 124312228-124312231 on Build GRCh38
rs374676101 Chr.9: 124312289-124312289 on Build GRCh38
rs755980284 Chr.9: 124312565-124312565 on Build GRCh38
rs182987834 Chr.9: 124312179-124312179 on Build GRCh38
rs998034936 Chr.9: 124312253-124312253 on Build GRCh38
rs377113006 Chr.9: 124312490-124312490 on Build GRCh38
rs889173236 Chr.9: 124312281-124312281 on Build GRCh38
rs891576245 Chr.9: 124312263-124312263 on Build GRCh38
rs539664097 Chr.9: 124312193-124312193 on Build GRCh38
rs780281219 Chr.9: 124312461-124312461 on Build GRCh38
rs187028612 Chr.9: 124312249-124312249 on Build GRCh38
rs769243779 Chr.9: 124312521-124312521 on Build GRCh38
rs576371804 Chr.9: 124312323-124312323 on Build GRCh38
rs893727415 Chr.9: 124312194-124312194 on Build GRCh38
rs113097292 Chr.9: 124312313-124312313 on Build GRCh38
rs749069904 Chr.9: 124312575-124312575 on Build GRCh38
rs984867725 Chr.9: 124312354-124312354 on Build GRCh38
rs960109013 Chr.9: 124312183-124312183 on Build GRCh38
rs183673274 Chr.9: 124312546-124312546 on Build GRCh38
rs1011263113 Chr.9: 124312276-124312276 on Build GRCh38
rs769601929 Chr.9: 124312497-124312497 on Build GRCh38
rs145621903 Chr.9: 124312130-124312130 on Build GRCh38
rs992257310 Chr.9: 124312184-124312184 on Build GRCh38
rs926404388 Chr.9: 124312402-124312402 on Build GRCh38
rs564864682 Chr.9: 124312501-124312501 on Build GRCh38
rs139169292 Chr.9: 124312505-124312505 on Build GRCh38
rs969687723 Chr.9: 124312301-124312301 on Build GRCh38
rs984204171 Chr.9: 124312315-124312335 on Build GRCh38
rs762937936 Chr.9: 124312449-124312449 on Build GRCh38
rs909222541 Chr.9: 124312355-124312355 on Build GRCh38
rs959344344 Chr.9: 124312429-124312429 on Build GRCh38
rs528302111 Chr.9: 124312354-124312354 on Build GRCh38
rs116276253 Chr.9: 124312523-124312523 on Build GRCh38
rs751531965 Chr.9: 124312462-124312462 on Build GRCh38
rs777558111 Chr.9: 124312572-124312572 on Build GRCh38
rs770850895 Chr.9: 124312355-124312356 on Build GRCh38
rs963661667 Chr.9: 124312336-124312336 on Build GRCh38
rs550629652 Chr.9: 124312505-124312505 on Build GRCh38
rs926304894 Chr.9: 124312175-124312175 on Build GRCh38
rs933576085 Chr.9: 124312383-124312383 on Build GRCh38
rs989145830 Chr.9: 124312471-124312471 on Build GRCh38
rs982664529 Chr.9: 124312386-124312386 on Build GRCh38
rs972948296 Chr.9: 124312327-124312327 on Build GRCh38
rs778485981 Chr.9: 124312500-124312500 on Build GRCh38
rs563656278 Chr.9: 124312484-124312484 on Build GRCh38
rs949189967 Chr.9: 124312163-124312163 on Build GRCh38
rs201673227 Chr.9: 124312483-124312483 on Build GRCh38
rs891962050 Chr.9: 124312419-124312419 on Build GRCh38

View Full Product Details