Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 18999886-19000354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRG2
Entrez Gene ID
10149
Gene Name
adhesion G protein-coupled receptor G2
Gene Aliases
EDDM6, GPR64, HE6, TM7LN2
UniGene
Hs.146978
Gene Chromosome Location
Chr.X: 18989307-19122956 on Build GRCh38
Transcripts
NM_001079858.2 NM_001079859.2 NM_001079860.2 NM_001184833.1 NM_001184834.1 NM_001184835.1 NM_001184836.1 NM_001184837.1 NM_005756.3 XM_006724455.3 XM_011545434.1 XM_011545435.2

SNP Detail

SNP ID Chromosome Location
rs993854171 Chr.X: 19000187-19000187 on Build GRCh38
rs774351011 Chr.X: 18999992-18999995 on Build GRCh38
rs200918266 Chr.X: 18999924-18999924 on Build GRCh38
rs934682223 Chr.X: 19000063-19000063 on Build GRCh38
rs748301650 Chr.X: 18999920-18999920 on Build GRCh38
rs769595498 Chr.X: 18999991-18999992 on Build GRCh38
rs180791825 Chr.X: 19000151-19000151 on Build GRCh38
rs961376383 Chr.X: 19000180-19000180 on Build GRCh38
rs763124232 Chr.X: 18999992-18999992 on Build GRCh38
rs1053495378 Chr.X: 19000079-19000079 on Build GRCh38
rs767640074 Chr.X: 18999992-19000000 on Build GRCh38
rs989087852 Chr.X: 19000241-19000241 on Build GRCh38
rs993518243 Chr.X: 19000246-19000246 on Build GRCh38
rs950935333 Chr.X: 19000022-19000022 on Build GRCh38
rs1023195655 Chr.X: 19000248-19000248 on Build GRCh38
rs368617850 Chr.X: 19000140-19000140 on Build GRCh38
rs1026936916 Chr.X: 19000012-19000012 on Build GRCh38
rs753578113 Chr.X: 18999994-18999994 on Build GRCh38
rs760092145 Chr.X: 18999984-18999984 on Build GRCh38
rs889644537 Chr.X: 19000128-19000128 on Build GRCh38
rs901166728 Chr.X: 19000085-19000085 on Build GRCh38
rs934776077 Chr.X: 19000286-19000286 on Build GRCh38
rs762979628 Chr.X: 18999951-18999951 on Build GRCh38
rs151329689 Chr.X: 19000293-19000293 on Build GRCh38
rs1049614136 Chr.X: 19000090-19000090 on Build GRCh38
rs754512355 Chr.X: 19000292-19000292 on Build GRCh38
rs1043139379 Chr.X: 19000276-19000276 on Build GRCh38
rs775240044 Chr.X: 19000107-19000107 on Build GRCh38
rs753288318 Chr.X: 19000139-19000139 on Build GRCh38
rs760224116 Chr.X: 18999987-18999987 on Build GRCh38
rs754203010 Chr.X: 19000218-19000218 on Build GRCh38
rs1005312586 Chr.X: 19000129-19000129 on Build GRCh38
rs774782254 Chr.X: 18999978-18999978 on Build GRCh38
rs771019669 Chr.X: 18999953-18999953 on Build GRCh38
rs759703894 Chr.X: 18999992-18999996 on Build GRCh38
rs933993201 Chr.X: 19000326-19000326 on Build GRCh38
rs948854977 Chr.X: 18999919-18999919 on Build GRCh38
rs926035621 Chr.X: 19000279-19000279 on Build GRCh38
rs754958892 Chr.X: 18999992-18999993 on Build GRCh38
rs986042831 Chr.X: 18999915-18999915 on Build GRCh38
rs766618512 Chr.X: 18999992-18999994 on Build GRCh38
rs960386908 Chr.X: 19000243-19000243 on Build GRCh38
rs746596366 Chr.X: 19000293-19000293 on Build GRCh38
rs756783657 Chr.X: 19000001-19000001 on Build GRCh38
rs143754029 Chr.X: 19000277-19000277 on Build GRCh38
rs758057101 Chr.X: 19000264-19000264 on Build GRCh38
rs764420562 Chr.X: 19000200-19000200 on Build GRCh38
rs779962322 Chr.X: 18999914-18999914 on Build GRCh38
rs998550113 Chr.X: 19000086-19000086 on Build GRCh38
rs926004663 Chr.X: 18999952-18999952 on Build GRCh38
rs767769852 Chr.X: 18999986-18999986 on Build GRCh38

View Full Product Details