Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 26138216-26138741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEB18
Entrez Gene ID
286514
Gene Name
MAGE family member B18
Gene Aliases
-
UniGene
Hs.350683
Gene Chromosome Location
Chr.X: 26138343-26140736 on Build GRCh38
Transcripts
NM_173699.3

SNP Detail

SNP ID Chromosome Location
rs968151369 Chr.X: 26138492-26138492 on Build GRCh38
rs1002596911 Chr.X: 26138427-26138427 on Build GRCh38
rs183028178 Chr.X: 26138526-26138526 on Build GRCh38
rs922935491 Chr.X: 26138533-26138533 on Build GRCh38
rs6526549 Chr.X: 26138331-26138331 on Build GRCh38
rs1006062291 Chr.X: 26138365-26138365 on Build GRCh38
rs910813747 Chr.X: 26138277-26138277 on Build GRCh38
rs962838866 Chr.X: 26138258-26138258 on Build GRCh38
rs41305165 Chr.X: 26138493-26138493 on Build GRCh38
rs886090219 Chr.X: 26138682-26138682 on Build GRCh38
rs961002885 Chr.X: 26138456-26138456 on Build GRCh38
rs986827004 Chr.X: 26138407-26138407 on Build GRCh38
rs758741069 Chr.X: 26138679-26138679 on Build GRCh38
rs1047526497 Chr.X: 26138326-26138326 on Build GRCh38
rs41305167 Chr.X: 26138243-26138243 on Build GRCh38
rs763174961 Chr.X: 26138320-26138320 on Build GRCh38
rs1040076691 Chr.X: 26138306-26138306 on Build GRCh38
rs928975397 Chr.X: 26138574-26138574 on Build GRCh38
rs575361870 Chr.X: 26138587-26138587 on Build GRCh38
rs1022855350 Chr.X: 26138369-26138369 on Build GRCh38
rs750754088 Chr.X: 26138588-26138588 on Build GRCh38
rs774795504 Chr.X: 26138614-26138614 on Build GRCh38
rs28697185 Chr.X: 26138683-26138683 on Build GRCh38
rs767788497 Chr.X: 26138527-26138527 on Build GRCh38
rs955979418 Chr.X: 26138523-26138523 on Build GRCh38
rs1020996498 Chr.X: 26138477-26138477 on Build GRCh38
rs1044291998 Chr.X: 26138656-26138656 on Build GRCh38
rs1028426763 Chr.X: 26138393-26138393 on Build GRCh38
rs1035055639 Chr.X: 26138428-26138428 on Build GRCh38
rs988160424 Chr.X: 26138462-26138462 on Build GRCh38
rs761205692 Chr.X: 26138626-26138626 on Build GRCh38
rs764541389 Chr.X: 26138319-26138319 on Build GRCh38
rs943023361 Chr.X: 26138300-26138300 on Build GRCh38
rs947251586 Chr.X: 26138531-26138531 on Build GRCh38
rs954231052 Chr.X: 26138406-26138406 on Build GRCh38
rs867845097 Chr.X: 26138402-26138402 on Build GRCh38
rs908970988 Chr.X: 26138623-26138623 on Build GRCh38
rs868252026 Chr.X: 26138247-26138247 on Build GRCh38
rs940307753 Chr.X: 26138698-26138698 on Build GRCh38
rs750802375 Chr.X: 26138528-26138528 on Build GRCh38

View Full Product Details