Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 103310226-103310731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEX2
Entrez Gene ID
84707
Gene Name
brain expressed X-linked 2
Gene Aliases
BEX1, DJ79P11.1
UniGene
Hs.398989
Gene Chromosome Location
Chr.X: 103309346-103311046 on Build GRCh38
Transcripts
NM_001168399.1 NM_001168400.1 NM_001168401.1 NM_032621.3

SNP Detail

SNP ID Chromosome Location
rs750769708 Chr.X: 103310412-103310412 on Build GRCh38
rs1035230384 Chr.X: 103310634-103310634 on Build GRCh38
rs962330634 Chr.X: 103310362-103310362 on Build GRCh38
rs763106357 Chr.X: 103310344-103310344 on Build GRCh38
rs2246252 Chr.X: 103310275-103310275 on Build GRCh38
rs970544599 Chr.X: 103310352-103310352 on Build GRCh38
rs1128200 Chr.X: 103310414-103310414 on Build GRCh38
rs1045057 Chr.X: 103310396-103310396 on Build GRCh38
rs1036595448 Chr.X: 103310541-103310541 on Build GRCh38
rs767970181 Chr.X: 103310402-103310402 on Build GRCh38
rs1050980142 Chr.X: 103310436-103310436 on Build GRCh38
rs777469582 Chr.X: 103310688-103310688 on Build GRCh38
rs1045050 Chr.X: 103310408-103310408 on Build GRCh38
rs868457875 Chr.X: 103310614-103310614 on Build GRCh38
rs935009841 Chr.X: 103310401-103310401 on Build GRCh38
rs942663426 Chr.X: 103310512-103310512 on Build GRCh38
rs775789864 Chr.X: 103310343-103310343 on Build GRCh38
rs946674215 Chr.X: 103310532-103310532 on Build GRCh38
rs1045052 Chr.X: 103310399-103310399 on Build GRCh38
rs3184865 Chr.X: 103310415-103310415 on Build GRCh38
rs902560822 Chr.X: 103310562-103310562 on Build GRCh38
rs752638891 Chr.X: 103310607-103310607 on Build GRCh38
rs894559723 Chr.X: 103310628-103310628 on Build GRCh38
rs762194007 Chr.X: 103310364-103310364 on Build GRCh38
rs1024390740 Chr.X: 103310682-103310682 on Build GRCh38
rs770138271 Chr.X: 103310340-103310340 on Build GRCh38
rs898057757 Chr.X: 103310547-103310547 on Build GRCh38
rs759896247 Chr.X: 103310337-103310337 on Build GRCh38
rs1128202 Chr.X: 103310373-103310373 on Build GRCh38
rs776549676 Chr.X: 103310335-103310335 on Build GRCh38
rs1023814076 Chr.X: 103310699-103310699 on Build GRCh38
rs752138391 Chr.X: 103310359-103310359 on Build GRCh38
rs890658000 Chr.X: 103310452-103310452 on Build GRCh38
rs752300447 Chr.X: 103310421-103310421 on Build GRCh38
rs770760954 Chr.X: 103310464-103310464 on Build GRCh38
rs376290170 Chr.X: 103310587-103310587 on Build GRCh38
rs747019825 Chr.X: 103310458-103310458 on Build GRCh38
rs994859787 Chr.X: 103310565-103310565 on Build GRCh38
rs1000016198 Chr.X: 103310599-103310599 on Build GRCh38
rs916270709 Chr.X: 103310255-103310255 on Build GRCh38
rs377532718 Chr.X: 103310257-103310262 on Build GRCh38
rs764029753 Chr.X: 103310348-103310348 on Build GRCh38
rs1045055 Chr.X: 103310398-103310398 on Build GRCh38
rs1040010417 Chr.X: 103310512-103310512 on Build GRCh38
rs74656502 Chr.X: 103310257-103310257 on Build GRCh38
rs759259308 Chr.X: 103310427-103310427 on Build GRCh38
rs185744581 Chr.X: 103310666-103310666 on Build GRCh38
rs1043842693 Chr.X: 103310534-103310534 on Build GRCh38
rs990674887 Chr.X: 103310277-103310277 on Build GRCh38
rs970196933 Chr.X: 103310449-103310449 on Build GRCh38
rs181583504 Chr.X: 103310285-103310285 on Build GRCh38
rs757959751 Chr.X: 103310424-103310424 on Build GRCh38
rs754689445 Chr.X: 103310413-103310413 on Build GRCh38
rs751303856 Chr.X: 103310417-103310417 on Build GRCh38
rs900525449 Chr.X: 103310525-103310525 on Build GRCh38
rs944863520 Chr.X: 103310530-103310530 on Build GRCh38
rs1023431087 Chr.X: 103310288-103310288 on Build GRCh38
rs973701934 Chr.X: 103310511-103310511 on Build GRCh38
rs781199713 Chr.X: 103310570-103310570 on Build GRCh38

View Full Product Details