Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 124380000-124380511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TENM1
Entrez Gene ID
10178
Gene Name
teneurin transmembrane protein 1
Gene Aliases
ODZ1, ODZ3, TEN-M1, TNM, TNM1
UniGene
Hs.23796
Gene Chromosome Location
Chr.X: 124374680-125204305 on Build GRCh38
Transcripts
NM_001163278.1 NM_001163279.1 NM_014253.3 XM_011531230.2 XM_011531236.2 XM_011531237.2 XM_017029208.1 XM_017029209.1 XM_017029210.1 XM_017029211.1 XM_017029212.1 XM_017029213.1 XM_017029214.1

SNP Detail

SNP ID Chromosome Location
rs754424221 Chr.X: 124380110-124380110 on Build GRCh38
rs183787422 Chr.X: 124380250-124380250 on Build GRCh38
rs904342332 Chr.X: 124380053-124380053 on Build GRCh38
rs550445021 Chr.X: 124380150-124380150 on Build GRCh38
rs971309470 Chr.X: 124380022-124380022 on Build GRCh38
rs866243608 Chr.X: 124380052-124380052 on Build GRCh38
rs1028345159 Chr.X: 124380246-124380246 on Build GRCh38
rs751104056 Chr.X: 124380078-124380078 on Build GRCh38
rs912026757 Chr.X: 124380256-124380256 on Build GRCh38
rs986319130 Chr.X: 124380251-124380251 on Build GRCh38
rs998556789 Chr.X: 124380212-124380212 on Build GRCh38
rs188517490 Chr.X: 124380396-124380396 on Build GRCh38
rs775550148 Chr.X: 124380367-124380370 on Build GRCh38
rs1017165751 Chr.X: 124380425-124380425 on Build GRCh38
rs982669897 Chr.X: 124380033-124380033 on Build GRCh38
rs760845129 Chr.X: 124380395-124380395 on Build GRCh38
rs777547690 Chr.X: 124380217-124380217 on Build GRCh38
rs777339025 Chr.X: 124380475-124380475 on Build GRCh38
rs1025984900 Chr.X: 124380485-124380486 on Build GRCh38
rs1042881225 Chr.X: 124380046-124380046 on Build GRCh38
rs1053187235 Chr.X: 124380145-124380145 on Build GRCh38
rs890007244 Chr.X: 124380328-124380328 on Build GRCh38
rs903431575 Chr.X: 124380402-124380402 on Build GRCh38
rs769660883 Chr.X: 124380200-124380202 on Build GRCh38
rs6648597 Chr.X: 124380156-124380156 on Build GRCh38
rs753383953 Chr.X: 124380088-124380088 on Build GRCh38
rs1005789512 Chr.X: 124380355-124380355 on Build GRCh38
rs41312759 Chr.X: 124380195-124380195 on Build GRCh38
rs961522076 Chr.X: 124380473-124380473 on Build GRCh38
rs772359429 Chr.X: 124380323-124380323 on Build GRCh38
rs962396103 Chr.X: 124380068-124380068 on Build GRCh38

View Full Product Details