Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130202618-130203122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF280C
Entrez Gene ID
55609
Gene Name
zinc finger protein 280C
Gene Aliases
SUHW3, ZNF633
UniGene
Hs.308418
Gene Chromosome Location
Chr.X: 130202699-130268948 on Build GRCh38
Transcripts
NM_017666.4 XM_006724765.3

SNP Detail

SNP ID Chromosome Location
rs780507177 Chr.X: 130202792-130202792 on Build GRCh38
rs186693362 Chr.X: 130202845-130202845 on Build GRCh38
rs897488495 Chr.X: 130202668-130202668 on Build GRCh38
rs756392680 Chr.X: 130202770-130202770 on Build GRCh38
rs905122640 Chr.X: 130202798-130202798 on Build GRCh38
rs939831074 Chr.X: 130203020-130203020 on Build GRCh38
rs12848296 Chr.X: 130202758-130202758 on Build GRCh38
rs919983975 Chr.X: 130203067-130203067 on Build GRCh38
rs940213960 Chr.X: 130203082-130203082 on Build GRCh38
rs748169502 Chr.X: 130202877-130202877 on Build GRCh38
rs920945457 Chr.X: 130202867-130202867 on Build GRCh38
rs1009636392 Chr.X: 130202872-130202872 on Build GRCh38
rs1055105442 Chr.X: 130203094-130203094 on Build GRCh38
rs1001969793 Chr.X: 130202818-130202818 on Build GRCh38
rs910034935 Chr.X: 130203033-130203033 on Build GRCh38
rs965398968 Chr.X: 130202900-130202900 on Build GRCh38
rs1001029673 Chr.X: 130202777-130202777 on Build GRCh38
rs966834241 Chr.X: 130202696-130202696 on Build GRCh38
rs1021495836 Chr.X: 130202897-130202897 on Build GRCh38
rs12848294 Chr.X: 130202753-130202753 on Build GRCh38
rs755308503 Chr.X: 130202847-130202847 on Build GRCh38
rs184766256 Chr.X: 130202971-130202971 on Build GRCh38
rs1013705577 Chr.X: 130202780-130202780 on Build GRCh38
rs148988487 Chr.X: 130203066-130203066 on Build GRCh38
rs190314754 Chr.X: 130202976-130202976 on Build GRCh38
rs1028534670 Chr.X: 130202950-130202950 on Build GRCh38
rs761433503 Chr.X: 130202859-130202860 on Build GRCh38
rs766245219 Chr.X: 130202855-130202860 on Build GRCh38
rs192617480 Chr.X: 130202893-130202893 on Build GRCh38
rs1036546903 Chr.X: 130202648-130202648 on Build GRCh38
rs182463199 Chr.X: 130202702-130202702 on Build GRCh38
rs780938060 Chr.X: 130202917-130202917 on Build GRCh38
rs1032716581 Chr.X: 130202785-130202785 on Build GRCh38
rs957354603 Chr.X: 130202830-130202830 on Build GRCh38
rs762842967 Chr.X: 130202861-130202861 on Build GRCh38
rs779128463 Chr.X: 130202868-130202868 on Build GRCh38
rs770788195 Chr.X: 130202869-130202869 on Build GRCh38

View Full Product Details