Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 236806060-236806566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTR
Entrez Gene ID
4548
Gene Name
5-methyltetrahydrofolate-homocysteine methyltransferase
Gene Aliases
HMAG, MS, cblG
UniGene
Hs.498187
Gene Chromosome Location
Chr.1: 236794304-236903981 on Build GRCh38
Transcripts
NM_000254.2 NM_001291939.1 NM_001291940.1 XM_005273141.4 XM_011544194.2 XM_017001329.1 XM_017001330.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
337654 c.298A>T p.S100C Chr.1: 236806192-236806192 on Build GRCh38
4487102 c.481C>T p.Q161* Chr.1: 236806207-236806207 on Build GRCh38
4487101 c.313C>T p.Q105* Chr.1: 236806207-236806207 on Build GRCh38
338252 c.265G>T p.G89W Chr.1: 236806159-236806159 on Build GRCh38
5669461 c.311C>T p.A104V Chr.1: 236806205-236806205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009197414 Chr.1: 236806473-236806473 on Build GRCh38
rs1022140766 Chr.1: 236806500-236806500 on Build GRCh38
rs928871851 Chr.1: 236806289-236806289 on Build GRCh38
rs763900581 Chr.1: 236806170-236806170 on Build GRCh38
rs367730702 Chr.1: 236806202-236806202 on Build GRCh38
rs766800721 Chr.1: 236806148-236806148 on Build GRCh38
rs778513217 Chr.1: 236806178-236806178 on Build GRCh38
rs888842531 Chr.1: 236806383-236806383 on Build GRCh38
rs902177943 Chr.1: 236806394-236806394 on Build GRCh38
rs755806533 Chr.1: 236806167-236806167 on Build GRCh38
rs145939391 Chr.1: 236806143-236806143 on Build GRCh38
rs901741474 Chr.1: 236806408-236806408 on Build GRCh38
rs774911437 Chr.1: 236806272-236806272 on Build GRCh38
rs1056853713 Chr.1: 236806309-236806309 on Build GRCh38
rs756873897 Chr.1: 236806172-236806172 on Build GRCh38
rs997026356 Chr.1: 236806089-236806089 on Build GRCh38
rs956335835 Chr.1: 236806465-236806465 on Build GRCh38
rs767967845 Chr.1: 236806278-236806278 on Build GRCh38
rs771918341 Chr.1: 236806101-236806101 on Build GRCh38
rs759888518 Chr.1: 236806276-236806276 on Build GRCh38
rs984844630 Chr.1: 236806221-236806221 on Build GRCh38
rs946519296 Chr.1: 236806188-236806188 on Build GRCh38
rs763321071 Chr.1: 236806147-236806147 on Build GRCh38
rs547956995 Chr.1: 236806354-236806354 on Build GRCh38
rs768700614 Chr.1: 236806119-236806119 on Build GRCh38
rs766742741 Chr.1: 236806271-236806271 on Build GRCh38
rs1028844347 Chr.1: 236806445-236806445 on Build GRCh38
rs781642325 Chr.1: 236806401-236806401 on Build GRCh38
rs146077730 Chr.1: 236806508-236806508 on Build GRCh38
rs980995246 Chr.1: 236806509-236806509 on Build GRCh38
rs745775089 Chr.1: 236806094-236806094 on Build GRCh38
rs1049940759 Chr.1: 236806362-236806362 on Build GRCh38
rs753563823 Chr.1: 236806285-236806285 on Build GRCh38
rs915162995 Chr.1: 236806179-236806179 on Build GRCh38
rs898754733 Chr.1: 236806345-236806345 on Build GRCh38
rs773235792 Chr.1: 236806263-236806263 on Build GRCh38
rs1043420705 Chr.1: 236806364-236806364 on Build GRCh38
rs552070315 Chr.1: 236806428-236806428 on Build GRCh38
rs753506566 Chr.1: 236806171-236806171 on Build GRCh38
rs996086567 Chr.1: 236806410-236806410 on Build GRCh38
rs751808381 Chr.1: 236806153-236806153 on Build GRCh38
rs745828101 Chr.1: 236806192-236806192 on Build GRCh38
rs199843808 Chr.1: 236806252-236806252 on Build GRCh38
rs956616795 Chr.1: 236806165-236806165 on Build GRCh38
rs765272438 Chr.1: 236806135-236806135 on Build GRCh38
rs961067441 Chr.1: 236806544-236806544 on Build GRCh38
rs778937056 Chr.1: 236806337-236806337 on Build GRCh38
rs533083283 Chr.1: 236806525-236806525 on Build GRCh38
rs543159675 Chr.1: 236806132-236806132 on Build GRCh38
rs940251738 Chr.1: 236806333-236806333 on Build GRCh38
rs758202311 Chr.1: 236806195-236806195 on Build GRCh38
rs748350137 Chr.1: 236806253-236806253 on Build GRCh38
rs763524854 Chr.1: 236806270-236806270 on Build GRCh38
rs766745649 Chr.1: 236806114-236806114 on Build GRCh38
rs908578305 Chr.1: 236806318-236806318 on Build GRCh38
rs751825341 Chr.1: 236806163-236806163 on Build GRCh38
rs768491623 Chr.1: 236806225-236806225 on Build GRCh38
rs769885251 Chr.1: 236806255-236806255 on Build GRCh38
rs1018633815 Chr.1: 236806158-236806158 on Build GRCh38
rs746836137 Chr.1: 236806219-236806219 on Build GRCh38
rs371419394 Chr.1: 236806117-236806117 on Build GRCh38
rs776820025 Chr.1: 236806122-236806122 on Build GRCh38
rs1006314119 Chr.1: 236806524-236806524 on Build GRCh38
rs138252407 Chr.1: 236806356-236806356 on Build GRCh38
rs980644569 Chr.1: 236806196-236806196 on Build GRCh38
rs937553404 Chr.1: 236806313-236806313 on Build GRCh38
rs142874816 Chr.1: 236806402-236806402 on Build GRCh38
rs775106520 Chr.1: 236806113-236806113 on Build GRCh38
rs753252190 Chr.1: 236806506-236806506 on Build GRCh38
rs184332230 Chr.1: 236806137-236806137 on Build GRCh38

View Full Product Details