Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 32011292-32011802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF5B
Entrez Gene ID
3799
Gene Name
kinesin family member 5B
Gene Aliases
HEL-S-61, KINH, KNS, KNS1, UKHC
UniGene
Hs.327736
Gene Chromosome Location
Chr.10: 32009010-32056443 on Build GRCh38
Transcripts
NM_004521.2 XM_017016224.1

SNP Detail

SNP ID Chromosome Location
rs1008466493 Chr.10: 32011494-32011494 on Build GRCh38
rs899484017 Chr.10: 32011666-32011666 on Build GRCh38
rs773034225 Chr.10: 32011480-32011480 on Build GRCh38
rs565679178 Chr.10: 32011409-32011409 on Build GRCh38
rs752462436 Chr.10: 32011500-32011500 on Build GRCh38
rs1035530511 Chr.10: 32011465-32011465 on Build GRCh38
rs1054637940 Chr.10: 32011442-32011443 on Build GRCh38
rs750824262 Chr.10: 32011650-32011650 on Build GRCh38
rs955639255 Chr.10: 32011325-32011325 on Build GRCh38
rs1018226431 Chr.10: 32011499-32011499 on Build GRCh38
rs951998606 Chr.10: 32011398-32011398 on Build GRCh38
rs916615460 Chr.10: 32011336-32011336 on Build GRCh38
rs767245779 Chr.10: 32011388-32011388 on Build GRCh38
rs974174693 Chr.10: 32011532-32011532 on Build GRCh38
rs960701115 Chr.10: 32011477-32011477 on Build GRCh38
rs190556914 Chr.10: 32011507-32011507 on Build GRCh38
rs1042808568 Chr.10: 32011718-32011718 on Build GRCh38
rs890835987 Chr.10: 32011373-32011373 on Build GRCh38
rs867722197 Chr.10: 32011317-32011317 on Build GRCh38
rs1007860149 Chr.10: 32011381-32011381 on Build GRCh38
rs992255874 Chr.10: 32011326-32011326 on Build GRCh38
rs145356849 Chr.10: 32011485-32011486 on Build GRCh38
rs759017238 Chr.10: 32011356-32011356 on Build GRCh38
rs536159818 Chr.10: 32011365-32011365 on Build GRCh38
rs201318035 Chr.10: 32011487-32011487 on Build GRCh38
rs765707067 Chr.10: 32011583-32011583 on Build GRCh38
rs868209002 Chr.10: 32011504-32011504 on Build GRCh38
rs567112865 Chr.10: 32011340-32011340 on Build GRCh38
rs1021906436 Chr.10: 32011418-32011418 on Build GRCh38
rs1038923017 Chr.10: 32011351-32011351 on Build GRCh38
rs1054141796 Chr.10: 32011763-32011764 on Build GRCh38
rs930127277 Chr.10: 32011548-32011548 on Build GRCh38
rs139136383 Chr.10: 32011409-32011410 on Build GRCh38
rs1011774167 Chr.10: 32011711-32011711 on Build GRCh38
rs926928872 Chr.10: 32011736-32011736 on Build GRCh38
rs558759624 Chr.10: 32011606-32011606 on Build GRCh38
rs182779696 Chr.10: 32011752-32011752 on Build GRCh38
rs916686652 Chr.10: 32011501-32011501 on Build GRCh38
rs995589066 Chr.10: 32011525-32011525 on Build GRCh38
rs529680611 Chr.10: 32011417-32011417 on Build GRCh38
rs1047338829 Chr.10: 32011371-32011371 on Build GRCh38
rs111264964 Chr.10: 32011447-32011447 on Build GRCh38
rs1047346943 Chr.10: 32011555-32011555 on Build GRCh38
rs570554357 Chr.10: 32011607-32011607 on Build GRCh38
rs903025827 Chr.10: 32011746-32011746 on Build GRCh38
rs899552847 Chr.10: 32011415-32011415 on Build GRCh38
rs937072219 Chr.10: 32011434-32011434 on Build GRCh38
rs537330034 Chr.10: 32011577-32011577 on Build GRCh38
rs554485629 Chr.10: 32011323-32011323 on Build GRCh38
rs1039800702 Chr.10: 32011649-32011649 on Build GRCh38
rs771564522 Chr.10: 32011457-32011457 on Build GRCh38
rs971298010 Chr.10: 32011715-32011715 on Build GRCh38
rs757680699 Chr.10: 32011535-32011535 on Build GRCh38
rs983652550 Chr.10: 32011612-32011613 on Build GRCh38
rs1010797333 Chr.10: 32011456-32011456 on Build GRCh38
rs907719467 Chr.10: 32011654-32011654 on Build GRCh38
rs1035582601 Chr.10: 32011761-32011761 on Build GRCh38
rs574424008 Chr.10: 32011587-32011587 on Build GRCh38
rs778609440 Chr.10: 32011361-32011363 on Build GRCh38

View Full Product Details