Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80159077-80159615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA11
Entrez Gene ID
311
Gene Name
annexin A11
Gene Aliases
ANX11, CAP50
UniGene
Hs.530291
Gene Chromosome Location
Chr.10: 80153953-80205677 on Build GRCh38
Transcripts
NM_001157.2 NM_001278407.1 NM_001278408.1 NM_001278409.1 NM_145868.1 NM_145869.1 XM_005269741.4 XM_005269742.1 XM_006717813.1 XM_006717814.3 XM_011539735.1 XM_011539736.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5428764 c.1266G>A p.M422I Chr.10: 80159110-80159110 on Build GRCh38
6020854 c.1189G>A p.E397K Chr.10: 80159187-80159187 on Build GRCh38
6243219 c.174+5G>T p.? Chr.10: 80159095-80159095 on Build GRCh38
1745095 c.1265delT p.M422fs*5 Chr.10: 80159111-80159111 on Build GRCh38
4573855 c.143C>G p.S48C Chr.10: 80159131-80159131 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs962957286 Chr.10: 80159185-80159185 on Build GRCh38
rs539615215 Chr.10: 80159161-80159161 on Build GRCh38
rs926487770 Chr.10: 80159461-80159461 on Build GRCh38
rs746042070 Chr.10: 80159144-80159144 on Build GRCh38
rs745786156 Chr.10: 80159115-80159115 on Build GRCh38
rs973267116 Chr.10: 80159403-80159403 on Build GRCh38
rs946873876 Chr.10: 80159290-80159290 on Build GRCh38
rs768088292 Chr.10: 80159238-80159238 on Build GRCh38
rs770785396 Chr.10: 80159104-80159104 on Build GRCh38
rs777173398 Chr.10: 80159360-80159360 on Build GRCh38
rs955249380 Chr.10: 80159484-80159484 on Build GRCh38
rs376783741 Chr.10: 80159203-80159204 on Build GRCh38
rs74666141 Chr.10: 80159152-80159152 on Build GRCh38
rs753071285 Chr.10: 80159148-80159148 on Build GRCh38
rs199860119 Chr.10: 80159218-80159218 on Build GRCh38
rs984046215 Chr.10: 80159334-80159334 on Build GRCh38
rs886862708 Chr.10: 80159374-80159374 on Build GRCh38
rs745864099 Chr.10: 80159166-80159166 on Build GRCh38
rs1056034299 Chr.10: 80159369-80159369 on Build GRCh38
rs779137776 Chr.10: 80159136-80159136 on Build GRCh38
rs919078033 Chr.10: 80159414-80159414 on Build GRCh38
rs61860018 Chr.10: 80159124-80159124 on Build GRCh38
rs778841776 Chr.10: 80159114-80159114 on Build GRCh38
rs1028638418 Chr.10: 80159201-80159201 on Build GRCh38
rs777457189 Chr.10: 80159100-80159100 on Build GRCh38
rs751266462 Chr.10: 80159585-80159585 on Build GRCh38
rs111629385 Chr.10: 80159098-80159098 on Build GRCh38
rs768043334 Chr.10: 80159141-80159141 on Build GRCh38
rs987469979 Chr.10: 80159489-80159489 on Build GRCh38
rs937919823 Chr.10: 80159463-80159463 on Build GRCh38
rs538003091 Chr.10: 80159339-80159339 on Build GRCh38
rs375383859 Chr.10: 80159142-80159142 on Build GRCh38
rs777508467 Chr.10: 80159154-80159154 on Build GRCh38
rs762296563 Chr.10: 80159474-80159474 on Build GRCh38
rs1028449315 Chr.10: 80159563-80159563 on Build GRCh38
rs765643802 Chr.10: 80159556-80159556 on Build GRCh38
rs987440441 Chr.10: 80159222-80159222 on Build GRCh38
rs111749321 Chr.10: 80159099-80159099 on Build GRCh38
rs373734382 Chr.10: 80159123-80159123 on Build GRCh38
rs75810295 Chr.10: 80159200-80159200 on Build GRCh38
rs548189402 Chr.10: 80159427-80159427 on Build GRCh38
rs961473750 Chr.10: 80159364-80159364 on Build GRCh38
rs1009302401 Chr.10: 80159497-80159497 on Build GRCh38
rs900935090 Chr.10: 80159552-80159552 on Build GRCh38
rs771962116 Chr.10: 80159189-80159189 on Build GRCh38
rs956682290 Chr.10: 80159554-80159554 on Build GRCh38
rs561587637 Chr.10: 80159103-80159103 on Build GRCh38
rs1028504937 Chr.10: 80159291-80159291 on Build GRCh38
rs1016888376 Chr.10: 80159593-80159593 on Build GRCh38
rs1046766209 Chr.10: 80159138-80159138 on Build GRCh38
rs139193001 Chr.10: 80159131-80159131 on Build GRCh38
rs556439937 Chr.10: 80159272-80159272 on Build GRCh38
rs905432429 Chr.10: 80159354-80159354 on Build GRCh38
rs774273496 Chr.10: 80159223-80159223 on Build GRCh38
rs746761934 Chr.10: 80159199-80159199 on Build GRCh38
rs80095636 Chr.10: 80159307-80159307 on Build GRCh38
rs1042561914 Chr.10: 80159517-80159517 on Build GRCh38
rs763033566 Chr.10: 80159128-80159128 on Build GRCh38
rs536292340 Chr.10: 80159475-80159475 on Build GRCh38
rs988313968 Chr.10: 80159267-80159267 on Build GRCh38
rs568343281 Chr.10: 80159464-80159464 on Build GRCh38
rs748028317 Chr.10: 80159143-80159145 on Build GRCh38
rs878955930 Chr.10: 80159220-80159220 on Build GRCh38
rs954259140 Chr.10: 80159570-80159570 on Build GRCh38
rs998436486 Chr.10: 80159251-80159251 on Build GRCh38
rs773306833 Chr.10: 80159127-80159127 on Build GRCh38
rs771102449 Chr.10: 80159220-80159220 on Build GRCh38
rs760129121 Chr.10: 80159237-80159237 on Build GRCh38
rs775240100 Chr.10: 80159121-80159121 on Build GRCh38
rs1046275660 Chr.10: 80159443-80159443 on Build GRCh38
rs146683788 Chr.10: 80159130-80159130 on Build GRCh38
rs778270793 Chr.10: 80159163-80159163 on Build GRCh38
rs753290653 Chr.10: 80159158-80159158 on Build GRCh38
rs759947992 Chr.10: 80159133-80159133 on Build GRCh38
rs940100824 Chr.10: 80159119-80159119 on Build GRCh38
rs201948928 Chr.10: 80159118-80159118 on Build GRCh38

View Full Product Details