Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 90895696-90896230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPP30
Entrez Gene ID
10556
Gene Name
ribonuclease P/MRP subunit p30
Gene Aliases
TSG15
UniGene
Hs.139120
Gene Chromosome Location
Chr.10: 90859062-90908558 on Build GRCh38
Transcripts
NM_001104546.1 NM_006413.4 XM_011539120.2 XM_011539121.2 XM_017015475.1 XM_017015476.1 XM_017015477.1 XM_017015478.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5831601 c.673G>T p.V225L Chr.10: 90895907-90895907 on Build GRCh38
7186779 c.602A>G p.Y201C Chr.10: 90895902-90895902 on Build GRCh38
921357 c.607G>A p.V203M Chr.10: 90895907-90895907 on Build GRCh38
7186778 c.602A>G p.Y201C Chr.10: 90895902-90895902 on Build GRCh38
921356 c.607G>A p.V203M Chr.10: 90895907-90895907 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575707559 Chr.10: 90896030-90896030 on Build GRCh38
rs141221334 Chr.10: 90895897-90895897 on Build GRCh38
rs535269986 Chr.10: 90895985-90895985 on Build GRCh38
rs866071217 Chr.10: 90895752-90895752 on Build GRCh38
rs878982517 Chr.10: 90895979-90895979 on Build GRCh38
rs560776633 Chr.10: 90896186-90896186 on Build GRCh38
rs558208750 Chr.10: 90896114-90896114 on Build GRCh38
rs1047987405 Chr.10: 90895991-90895991 on Build GRCh38
rs1020620061 Chr.10: 90895889-90895889 on Build GRCh38
rs927725483 Chr.10: 90896036-90896036 on Build GRCh38
rs777638660 Chr.10: 90896019-90896019 on Build GRCh38
rs1037716677 Chr.10: 90895730-90895730 on Build GRCh38
rs566386266 Chr.10: 90895980-90895980 on Build GRCh38
rs756880890 Chr.10: 90895906-90895906 on Build GRCh38
rs148105825 Chr.10: 90895807-90895807 on Build GRCh38
rs1053965469 Chr.10: 90896099-90896099 on Build GRCh38
rs919278950 Chr.10: 90896169-90896169 on Build GRCh38
rs867177747 Chr.10: 90896107-90896107 on Build GRCh38
rs550568752 Chr.10: 90895811-90895811 on Build GRCh38
rs370064577 Chr.10: 90895872-90895872 on Build GRCh38
rs145521039 Chr.10: 90896203-90896206 on Build GRCh38
rs755762589 Chr.10: 90895840-90895850 on Build GRCh38
rs540623133 Chr.10: 90896155-90896155 on Build GRCh38
rs150762208 Chr.10: 90895812-90895813 on Build GRCh38
rs1017808803 Chr.10: 90895814-90895814 on Build GRCh38
rs917041189 Chr.10: 90896003-90896003 on Build GRCh38
rs570389012 Chr.10: 90895831-90895831 on Build GRCh38
rs191597163 Chr.10: 90895776-90895776 on Build GRCh38
rs763470383 Chr.10: 90895880-90895880 on Build GRCh38
rs753106932 Chr.10: 90895968-90895968 on Build GRCh38
rs555114664 Chr.10: 90895995-90895995 on Build GRCh38
rs779867731 Chr.10: 90895861-90895862 on Build GRCh38
rs767421586 Chr.10: 90895857-90895857 on Build GRCh38
rs781638278 Chr.10: 90895956-90895956 on Build GRCh38
rs747113503 Chr.10: 90895983-90895983 on Build GRCh38
rs73315680 Chr.10: 90895855-90895855 on Build GRCh38
rs536721772 Chr.10: 90895809-90895809 on Build GRCh38
rs981868631 Chr.10: 90896028-90896028 on Build GRCh38
rs552888142 Chr.10: 90895944-90895944 on Build GRCh38
rs1007705866 Chr.10: 90895810-90895810 on Build GRCh38
rs181276001 Chr.10: 90896050-90896050 on Build GRCh38
rs750081883 Chr.10: 90895938-90895938 on Build GRCh38
rs764936639 Chr.10: 90895907-90895907 on Build GRCh38
rs577996094 Chr.10: 90896147-90896147 on Build GRCh38
rs796408937 Chr.10: 90895809-90895812 on Build GRCh38
rs765271378 Chr.10: 90895793-90895796 on Build GRCh38
rs542799720 Chr.10: 90895815-90895815 on Build GRCh38
rs989517333 Chr.10: 90895870-90895870 on Build GRCh38
rs565598458 Chr.10: 90896056-90896056 on Build GRCh38
rs1038086095 Chr.10: 90896119-90896119 on Build GRCh38
rs375055405 Chr.10: 90895751-90895751 on Build GRCh38
rs775230842 Chr.10: 90895869-90895869 on Build GRCh38
rs993561951 Chr.10: 90895790-90895790 on Build GRCh38
rs746758024 Chr.10: 90896072-90896072 on Build GRCh38
rs10881848 Chr.10: 90895802-90895802 on Build GRCh38
rs55809834 Chr.10: 90896121-90896121 on Build GRCh38

View Full Product Details